Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6496688-6497194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRID2IP
Entrez Gene ID
392862
Gene Name
Grid2 interacting protein
Gene Aliases
DELPHILIN
UniGene
Hs.512174
Gene Chromosome Location
Chr.7: 6496778-6551436 on Build GRCh38
Transcripts
NM_001145118.1 XM_011515404.2

SNP Detail

SNP ID Chromosome Location
rs564581501 Chr.7: 6497012-6497012 on Build GRCh38
rs375817913 Chr.7: 6496915-6496915 on Build GRCh38
rs146416970 Chr.7: 6496926-6496926 on Build GRCh38
rs368523010 Chr.7: 6497048-6497048 on Build GRCh38
rs903186556 Chr.7: 6496968-6496968 on Build GRCh38
rs1025011346 Chr.7: 6496977-6496977 on Build GRCh38
rs756167382 Chr.7: 6496744-6496745 on Build GRCh38
rs78961173 Chr.7: 6497173-6497173 on Build GRCh38
rs1038698118 Chr.7: 6496813-6496813 on Build GRCh38
rs937799312 Chr.7: 6497117-6497117 on Build GRCh38
rs114173960 Chr.7: 6496800-6496800 on Build GRCh38
rs927213981 Chr.7: 6497055-6497055 on Build GRCh38
rs952652906 Chr.7: 6496852-6496852 on Build GRCh38
rs544150580 Chr.7: 6497027-6497027 on Build GRCh38
rs937764451 Chr.7: 6496901-6496901 on Build GRCh38
rs1026760245 Chr.7: 6496839-6496839 on Build GRCh38
rs79502733 Chr.7: 6496911-6496911 on Build GRCh38
rs572830407 Chr.7: 6496934-6496934 on Build GRCh38
rs181580376 Chr.7: 6497056-6497056 on Build GRCh38
rs549489384 Chr.7: 6496759-6496761 on Build GRCh38
rs981905011 Chr.7: 6496896-6496896 on Build GRCh38
rs559036622 Chr.7: 6496767-6496767 on Build GRCh38
rs567931772 Chr.7: 6496786-6496786 on Build GRCh38
rs754063375 Chr.7: 6496743-6496745 on Build GRCh38
rs139121791 Chr.7: 6497013-6497013 on Build GRCh38
rs142749037 Chr.7: 6496760-6496761 on Build GRCh38
rs548825875 Chr.7: 6497064-6497064 on Build GRCh38
rs970434616 Chr.7: 6497069-6497069 on Build GRCh38
rs368600971 Chr.7: 6496740-6496741 on Build GRCh38
rs990802838 Chr.7: 6496954-6496954 on Build GRCh38
rs989221871 Chr.7: 6497121-6497121 on Build GRCh38
rs542474445 Chr.7: 6496921-6496921 on Build GRCh38
rs1015444282 Chr.7: 6497057-6497057 on Build GRCh38
rs1019708500 Chr.7: 6496991-6496991 on Build GRCh38
rs908228155 Chr.7: 6496879-6496879 on Build GRCh38
rs796859983 Chr.7: 6496742-6496744 on Build GRCh38
rs982215319 Chr.7: 6497088-6497088 on Build GRCh38
rs751963173 Chr.7: 6497120-6497120 on Build GRCh38
rs535012151 Chr.7: 6496903-6496903 on Build GRCh38
rs773761298 Chr.7: 6497011-6497011 on Build GRCh38
rs755670062 Chr.7: 6497128-6497128 on Build GRCh38
rs765704656 Chr.7: 6496875-6496875 on Build GRCh38
rs766706667 Chr.7: 6496742-6496745 on Build GRCh38
rs538198407 Chr.7: 6496858-6496858 on Build GRCh38
rs540133614 Chr.7: 6496978-6496978 on Build GRCh38
rs149387100 Chr.7: 6497131-6497131 on Build GRCh38
rs763836635 Chr.7: 6497135-6497135 on Build GRCh38
rs763294219 Chr.7: 6497043-6497043 on Build GRCh38
rs534471227 Chr.7: 6496765-6496765 on Build GRCh38
rs55699366 Chr.7: 6496890-6496890 on Build GRCh38
rs943151536 Chr.7: 6497044-6497044 on Build GRCh38
rs926337780 Chr.7: 6496897-6496897 on Build GRCh38
rs919187518 Chr.7: 6497019-6497019 on Build GRCh38
rs902090460 Chr.7: 6496837-6496837 on Build GRCh38
rs77587965 Chr.7: 6496760-6496763 on Build GRCh38
rs528118102 Chr.7: 6496742-6496743 on Build GRCh38
rs562467939 Chr.7: 6497031-6497031 on Build GRCh38
rs79101283 Chr.7: 6497110-6497110 on Build GRCh38

View Full Product Details