Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6591227-6591771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf26
Entrez Gene ID
79034
Gene Name
chromosome 7 open reading frame 26
Gene Aliases
-
UniGene
Hs.487511
Gene Chromosome Location
Chr.7: 6589368-6608726 on Build GRCh38
Transcripts
NM_001303039.1 NM_024067.3 XM_005249845.4 XM_011515518.2 XM_017012614.1 XM_017012615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4534775 c.211G>A p.E71K Chr.7: 6591664-6591664 on Build GRCh38
3085919 c.271C>T p.R91W Chr.7: 6591724-6591724 on Build GRCh38
7151792 c.271C>A p.R91R Chr.7: 6591724-6591724 on Build GRCh38
1755339 c.297C>G p.F99L Chr.7: 6591750-6591750 on Build GRCh38
7116313 c.194G>C p.R65T Chr.7: 6591647-6591647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051221937 Chr.7: 6591452-6591454 on Build GRCh38
rs771996232 Chr.7: 6591679-6591679 on Build GRCh38
rs374676551 Chr.7: 6591481-6591481 on Build GRCh38
rs973940201 Chr.7: 6591366-6591366 on Build GRCh38
rs183961465 Chr.7: 6591554-6591554 on Build GRCh38
rs555309516 Chr.7: 6591426-6591426 on Build GRCh38
rs545954942 Chr.7: 6591519-6591519 on Build GRCh38
rs113895450 Chr.7: 6591454-6591455 on Build GRCh38
rs188590629 Chr.7: 6591623-6591623 on Build GRCh38
rs543531934 Chr.7: 6591415-6591415 on Build GRCh38
rs980517103 Chr.7: 6591565-6591565 on Build GRCh38
rs1051133600 Chr.7: 6591701-6591701 on Build GRCh38
rs796144828 Chr.7: 6591268-6591268 on Build GRCh38
rs753604756 Chr.7: 6591692-6591692 on Build GRCh38
rs995414220 Chr.7: 6591491-6591491 on Build GRCh38
rs763523249 Chr.7: 6591596-6591596 on Build GRCh38
rs899451025 Chr.7: 6591441-6591441 on Build GRCh38
rs898809082 Chr.7: 6591643-6591643 on Build GRCh38
rs1007314135 Chr.7: 6591286-6591286 on Build GRCh38
rs747814413 Chr.7: 6591357-6591357 on Build GRCh38
rs1025280074 Chr.7: 6591377-6591377 on Build GRCh38
rs779953466 Chr.7: 6591603-6591603 on Build GRCh38
rs987498428 Chr.7: 6591336-6591336 on Build GRCh38
rs755940962 Chr.7: 6591601-6591601 on Build GRCh38
rs763372025 Chr.7: 6591639-6591639 on Build GRCh38
rs560395568 Chr.7: 6591497-6591497 on Build GRCh38
rs1013663163 Chr.7: 6591539-6591539 on Build GRCh38
rs775365008 Chr.7: 6591699-6591699 on Build GRCh38
rs751642837 Chr.7: 6591728-6591728 on Build GRCh38
rs554448439 Chr.7: 6591274-6591274 on Build GRCh38
rs11459559 Chr.7: 6591426-6591427 on Build GRCh38
rs1008991617 Chr.7: 6591480-6591480 on Build GRCh38
rs750131690 Chr.7: 6591672-6591672 on Build GRCh38
rs377013109 Chr.7: 6591269-6591269 on Build GRCh38
rs372457145 Chr.7: 6591553-6591553 on Build GRCh38
rs777722001 Chr.7: 6591609-6591609 on Build GRCh38
rs775786469 Chr.7: 6591637-6591637 on Build GRCh38
rs112645978 Chr.7: 6591432-6591432 on Build GRCh38
rs541677641 Chr.7: 6591453-6591453 on Build GRCh38
rs946026380 Chr.7: 6591451-6591451 on Build GRCh38
rs398003548 Chr.7: 6591285-6591285 on Build GRCh38
rs778196249 Chr.7: 6591629-6591629 on Build GRCh38
rs746114214 Chr.7: 6591734-6591734 on Build GRCh38
rs991894176 Chr.7: 6591391-6591391 on Build GRCh38
rs754938039 Chr.7: 6591695-6591695 on Build GRCh38
rs754888906 Chr.7: 6591607-6591607 on Build GRCh38
rs1007127231 Chr.7: 6591276-6591276 on Build GRCh38
rs560539982 Chr.7: 6591506-6591506 on Build GRCh38
rs770325450 Chr.7: 6591582-6591582 on Build GRCh38
rs766291186 Chr.7: 6591686-6591686 on Build GRCh38
rs1030140049 Chr.7: 6591273-6591273 on Build GRCh38
rs771118265 Chr.7: 6591622-6591622 on Build GRCh38
rs368858890 Chr.7: 6591414-6591414 on Build GRCh38
rs756923211 Chr.7: 6591599-6591599 on Build GRCh38
rs569932813 Chr.7: 6591733-6591733 on Build GRCh38
rs536115160 Chr.7: 6591260-6591260 on Build GRCh38
rs769198880 Chr.7: 6591642-6591642 on Build GRCh38
rs576536674 Chr.7: 6591353-6591353 on Build GRCh38
rs111250508 Chr.7: 6591352-6591352 on Build GRCh38
rs568012038 Chr.7: 6591555-6591555 on Build GRCh38
rs756300721 Chr.7: 6591542-6591542 on Build GRCh38
rs566290185 Chr.7: 6591299-6591299 on Build GRCh38
rs1036437411 Chr.7: 6591438-6591438 on Build GRCh38
rs1054700391 Chr.7: 6591440-6591440 on Build GRCh38
rs540015497 Chr.7: 6591347-6591347 on Build GRCh38
rs766197906 Chr.7: 6591286-6591286 on Build GRCh38
rs528891661 Chr.7: 6591556-6591556 on Build GRCh38
rs564145151 Chr.7: 6591525-6591525 on Build GRCh38
rs953518427 Chr.7: 6591275-6591275 on Build GRCh38
rs767261844 Chr.7: 6591600-6591600 on Build GRCh38
rs747107778 Chr.7: 6591621-6591621 on Build GRCh38
rs576106471 Chr.7: 6591386-6591386 on Build GRCh38
rs995484880 Chr.7: 6591445-6591445 on Build GRCh38
rs547118974 Chr.7: 6591563-6591563 on Build GRCh38
rs201524287 Chr.7: 6591710-6591710 on Build GRCh38
rs767317357 Chr.7: 6591658-6591658 on Build GRCh38
rs749259329 Chr.7: 6591605-6591605 on Build GRCh38
rs761361184 Chr.7: 6591657-6591657 on Build GRCh38
rs927202112 Chr.7: 6591586-6591586 on Build GRCh38
rs978645072 Chr.7: 6591408-6591408 on Build GRCh38
rs146220253 Chr.7: 6591652-6591652 on Build GRCh38
rs9690179 Chr.7: 6591530-6591530 on Build GRCh38
rs770177848 Chr.7: 6591630-6591630 on Build GRCh38
rs376154184 Chr.7: 6591594-6591594 on Build GRCh38
rs913187319 Chr.7: 6591380-6591380 on Build GRCh38
rs769933870 Chr.7: 6591744-6591744 on Build GRCh38
rs760352445 Chr.7: 6591676-6591676 on Build GRCh38
rs111552863 Chr.7: 6591593-6591593 on Build GRCh38
rs570249186 Chr.7: 6591416-6591416 on Build GRCh38
rs889300747 Chr.7: 6591466-6591466 on Build GRCh38
rs895536199 Chr.7: 6591431-6591431 on Build GRCh38
rs560804771 Chr.7: 6591724-6591724 on Build GRCh38
rs988191949 Chr.7: 6591533-6591533 on Build GRCh38
rs1039963207 Chr.7: 6591472-6591472 on Build GRCh38
rs377738296 Chr.7: 6591268-6591268 on Build GRCh38
rs751012958 Chr.7: 6591597-6591597 on Build GRCh38

View Full Product Details