Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 12344240-12344742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWDE
Entrez Gene ID
221806
Gene Name
von Willebrand factor D and EGF domains
Gene Aliases
-
UniGene
Hs.669526
Gene Chromosome Location
Chr.7: 12330883-12404226 on Build GRCh38
Transcripts
NM_001135924.1 XM_005249641.1 XM_011515174.1 XM_011515175.1 XM_017011828.1 XM_017011829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4186547 c.1754A>C p.Q585P Chr.7: 12344379-12344379 on Build GRCh38
2767731 c.3991G>T p.D1331Y Chr.7: 12344282-12344282 on Build GRCh38
6030313 c.3908G>C p.G1303A Chr.7: 12344448-12344448 on Build GRCh38
2767729 c.4028C>T p.P1343L Chr.7: 12344245-12344245 on Build GRCh38
5423953 c.1696G>C p.E566Q Chr.7: 12344437-12344437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199577271 Chr.7: 12344487-12344487 on Build GRCh38
rs951906786 Chr.7: 12344662-12344662 on Build GRCh38
rs184450251 Chr.7: 12344357-12344357 on Build GRCh38
rs767752866 Chr.7: 12344450-12344450 on Build GRCh38
rs1014942092 Chr.7: 12344670-12344670 on Build GRCh38
rs756760186 Chr.7: 12344380-12344380 on Build GRCh38
rs760552046 Chr.7: 12344633-12344633 on Build GRCh38
rs772387107 Chr.7: 12344458-12344458 on Build GRCh38
rs17165872 Chr.7: 12344463-12344463 on Build GRCh38
rs61732210 Chr.7: 12344280-12344280 on Build GRCh38
rs983727733 Chr.7: 12344663-12344663 on Build GRCh38
rs538601768 Chr.7: 12344541-12344541 on Build GRCh38
rs983522492 Chr.7: 12344468-12344468 on Build GRCh38
rs754820996 Chr.7: 12344493-12344493 on Build GRCh38
rs765567937 Chr.7: 12344321-12344321 on Build GRCh38
rs536789782 Chr.7: 12344503-12344503 on Build GRCh38
rs1042138202 Chr.7: 12344524-12344524 on Build GRCh38
rs907983926 Chr.7: 12344471-12344471 on Build GRCh38
rs375483470 Chr.7: 12344320-12344320 on Build GRCh38
rs950050279 Chr.7: 12344479-12344479 on Build GRCh38
rs964652057 Chr.7: 12344410-12344410 on Build GRCh38
rs1008607934 Chr.7: 12344348-12344348 on Build GRCh38
rs1018593644 Chr.7: 12344549-12344549 on Build GRCh38
rs763874469 Chr.7: 12344716-12344716 on Build GRCh38
rs73294382 Chr.7: 12344412-12344412 on Build GRCh38
rs1440032 Chr.7: 12344341-12344341 on Build GRCh38
rs866156848 Chr.7: 12344437-12344437 on Build GRCh38
rs553043270 Chr.7: 12344515-12344515 on Build GRCh38
rs561386091 Chr.7: 12344482-12344482 on Build GRCh38
rs557493216 Chr.7: 12344421-12344421 on Build GRCh38
rs905149496 Chr.7: 12344311-12344311 on Build GRCh38
rs754135749 Chr.7: 12344293-12344293 on Build GRCh38
rs550430548 Chr.7: 12344614-12344614 on Build GRCh38
rs73294384 Chr.7: 12344579-12344579 on Build GRCh38
rs543274392 Chr.7: 12344594-12344594 on Build GRCh38
rs774630565 Chr.7: 12344288-12344288 on Build GRCh38
rs374464426 Chr.7: 12344607-12344607 on Build GRCh38
rs893265255 Chr.7: 12344509-12344509 on Build GRCh38
rs768610830 Chr.7: 12344489-12344489 on Build GRCh38
rs902288360 Chr.7: 12344531-12344531 on Build GRCh38
rs866878189 Chr.7: 12344448-12344448 on Build GRCh38
rs370930703 Chr.7: 12344367-12344367 on Build GRCh38
rs941967666 Chr.7: 12344659-12344659 on Build GRCh38
rs368736806 Chr.7: 12344362-12344362 on Build GRCh38
rs773614884 Chr.7: 12344267-12344267 on Build GRCh38
rs771147958 Chr.7: 12344277-12344277 on Build GRCh38
rs767685497 Chr.7: 12344320-12344321 on Build GRCh38
rs572950976 Chr.7: 12344519-12344519 on Build GRCh38
rs981429486 Chr.7: 12344481-12344481 on Build GRCh38
rs978934398 Chr.7: 12344540-12344540 on Build GRCh38
rs563526483 Chr.7: 12344608-12344608 on Build GRCh38
rs750581658 Chr.7: 12344472-12344472 on Build GRCh38
rs1008555571 Chr.7: 12344545-12344545 on Build GRCh38
rs866018656 Chr.7: 12344686-12344686 on Build GRCh38
rs756426822 Chr.7: 12344471-12344472 on Build GRCh38
rs1027468797 Chr.7: 12344443-12344443 on Build GRCh38
rs756009983 Chr.7: 12344453-12344453 on Build GRCh38
rs557640963 Chr.7: 12344572-12344572 on Build GRCh38
rs926515679 Chr.7: 12344640-12344640 on Build GRCh38
rs528082668 Chr.7: 12344285-12344285 on Build GRCh38
rs1018855528 Chr.7: 12344359-12344359 on Build GRCh38

View Full Product Details