Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16604481-16605006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKMY2
Entrez Gene ID
57037
Gene Name
ankyrin repeat and MYND domain containing 2
Gene Aliases
ZMYND20
UniGene
Hs.157378
Gene Chromosome Location
Chr.7: 16599776-16645817 on Build GRCh38
Transcripts
NM_020319.2 XM_011515458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1449755 c.883-1G>C p.? Chr.7: 16604850-16604850 on Build GRCh38
253666 c.1000G>C p.V334L Chr.7: 16604732-16604732 on Build GRCh38
3923290 c.940T>G p.F314V Chr.7: 16604792-16604792 on Build GRCh38
1192085 c.998C>T p.S333L Chr.7: 16604734-16604734 on Build GRCh38
5887967 c.887C>T p.S296F Chr.7: 16604845-16604845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142708506 Chr.7: 16604759-16604759 on Build GRCh38
rs759041539 Chr.7: 16604765-16604765 on Build GRCh38
rs546491556 Chr.7: 16604923-16604923 on Build GRCh38
rs764746423 Chr.7: 16604878-16604878 on Build GRCh38
rs772631537 Chr.7: 16604846-16604846 on Build GRCh38
rs867926925 Chr.7: 16604857-16604857 on Build GRCh38
rs756177251 Chr.7: 16604972-16604974 on Build GRCh38
rs760314156 Chr.7: 16604730-16604730 on Build GRCh38
rs747989042 Chr.7: 16604930-16604930 on Build GRCh38
rs901662045 Chr.7: 16604645-16604645 on Build GRCh38
rs530415888 Chr.7: 16604639-16604639 on Build GRCh38
rs887693019 Chr.7: 16604718-16604718 on Build GRCh38
rs974578700 Chr.7: 16604661-16604661 on Build GRCh38
rs2108137 Chr.7: 16604595-16604595 on Build GRCh38
rs1010404429 Chr.7: 16604592-16604592 on Build GRCh38
rs778869600 Chr.7: 16604826-16604826 on Build GRCh38
rs76610649 Chr.7: 16604583-16604583 on Build GRCh38
rs117183796 Chr.7: 16604695-16604695 on Build GRCh38
rs946703153 Chr.7: 16604604-16604604 on Build GRCh38
rs746032254 Chr.7: 16604675-16604675 on Build GRCh38
rs748636586 Chr.7: 16604829-16604829 on Build GRCh38
rs377609221 Chr.7: 16604586-16604586 on Build GRCh38
rs767867521 Chr.7: 16604803-16604804 on Build GRCh38
rs779964987 Chr.7: 16604689-16604689 on Build GRCh38
rs17169559 Chr.7: 16604516-16604516 on Build GRCh38
rs369924009 Chr.7: 16604644-16604644 on Build GRCh38
rs368284594 Chr.7: 16604787-16604787 on Build GRCh38
rs929800272 Chr.7: 16604717-16604717 on Build GRCh38
rs758423349 Chr.7: 16604785-16604785 on Build GRCh38
rs773622347 Chr.7: 16604727-16604727 on Build GRCh38
rs368790215 Chr.7: 16604723-16604723 on Build GRCh38
rs372146581 Chr.7: 16604810-16604810 on Build GRCh38
rs754489577 Chr.7: 16604739-16604739 on Build GRCh38
rs1015016553 Chr.7: 16604752-16604752 on Build GRCh38
rs762186011 Chr.7: 16604714-16604714 on Build GRCh38
rs528357449 Chr.7: 16604774-16604774 on Build GRCh38
rs1005850235 Chr.7: 16604744-16604744 on Build GRCh38
rs115594911 Chr.7: 16604937-16604937 on Build GRCh38
rs762309616 Chr.7: 16604721-16604721 on Build GRCh38
rs147344401 Chr.7: 16604806-16604806 on Build GRCh38
rs199673236 Chr.7: 16604671-16604671 on Build GRCh38
rs936395993 Chr.7: 16604856-16604856 on Build GRCh38
rs768714189 Chr.7: 16604825-16604825 on Build GRCh38
rs774880964 Chr.7: 16604711-16604711 on Build GRCh38
rs771224788 Chr.7: 16604865-16604865 on Build GRCh38
rs916859031 Chr.7: 16604598-16604598 on Build GRCh38
rs897520336 Chr.7: 16604766-16604766 on Build GRCh38
rs976154736 Chr.7: 16604933-16604933 on Build GRCh38
rs778888872 Chr.7: 16604786-16604786 on Build GRCh38
rs766098958 Chr.7: 16604733-16604733 on Build GRCh38
rs770199938 Chr.7: 16604678-16604678 on Build GRCh38
rs756381245 Chr.7: 16604807-16604807 on Build GRCh38
rs935419440 Chr.7: 16604629-16604629 on Build GRCh38
rs748461116 Chr.7: 16604893-16604893 on Build GRCh38
rs117794925 Chr.7: 16604637-16604637 on Build GRCh38
rs373553702 Chr.7: 16604877-16604877 on Build GRCh38
rs747426006 Chr.7: 16604849-16604849 on Build GRCh38
rs772167970 Chr.7: 16604950-16604950 on Build GRCh38
rs557489717 Chr.7: 16604943-16604943 on Build GRCh38
rs924022945 Chr.7: 16604615-16604615 on Build GRCh38
rs570656569 Chr.7: 16604701-16604701 on Build GRCh38
rs768641049 Chr.7: 16604708-16604708 on Build GRCh38
rs775199035 Chr.7: 16604887-16604887 on Build GRCh38
rs187223211 Chr.7: 16604551-16604551 on Build GRCh38
rs118106780 Chr.7: 16604605-16604605 on Build GRCh38
rs776310352 Chr.7: 16604866-16604866 on Build GRCh38
rs935345821 Chr.7: 16604925-16604925 on Build GRCh38
rs1034318249 Chr.7: 16604968-16604968 on Build GRCh38
rs137921647 Chr.7: 16604934-16604934 on Build GRCh38
rs541502353 Chr.7: 16604625-16604625 on Build GRCh38
rs780272104 Chr.7: 16604808-16604808 on Build GRCh38
rs753268272 Chr.7: 16604734-16604734 on Build GRCh38
rs891935829 Chr.7: 16604873-16604873 on Build GRCh38
rs764688085 Chr.7: 16604767-16604767 on Build GRCh38

View Full Product Details