Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75534826-75535366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIP1
Entrez Gene ID
3092
Gene Name
huntingtin interacting protein 1
Gene Aliases
HIP-I, ILWEQ, SHON, SHONbeta, SHONgamma
UniGene
Hs.329266
Gene Chromosome Location
Chr.7: 75533298-75738976 on Build GRCh38
Transcripts
NM_001243198.2 NM_005338.6 XM_005250304.2 XM_005250305.2 XM_011516116.2 XM_017012099.1

SNP Detail

SNP ID Chromosome Location
rs367974587 Chr.7: 75534898-75534898 on Build GRCh38
rs782416678 Chr.7: 75535002-75535002 on Build GRCh38
rs371553830 Chr.7: 75534882-75534882 on Build GRCh38
rs587617715 Chr.7: 75535086-75535086 on Build GRCh38
rs782683785 Chr.7: 75534962-75534962 on Build GRCh38
rs926505569 Chr.7: 75535270-75535270 on Build GRCh38
rs782069943 Chr.7: 75535342-75535342 on Build GRCh38
rs919652879 Chr.7: 75534883-75534883 on Build GRCh38
rs1009644627 Chr.7: 75535322-75535322 on Build GRCh38
rs931046620 Chr.7: 75535190-75535190 on Build GRCh38
rs781908236 Chr.7: 75535278-75535278 on Build GRCh38
rs1042829712 Chr.7: 75535326-75535326 on Build GRCh38
rs949329111 Chr.7: 75534935-75534935 on Build GRCh38
rs1002000316 Chr.7: 75534996-75534996 on Build GRCh38
rs1020950225 Chr.7: 75535077-75535077 on Build GRCh38
rs988166130 Chr.7: 75534859-75534859 on Build GRCh38
rs910710205 Chr.7: 75534862-75534862 on Build GRCh38
rs973513131 Chr.7: 75535170-75535170 on Build GRCh38
rs781798994 Chr.7: 75534969-75534969 on Build GRCh38
rs1046069168 Chr.7: 75535253-75535253 on Build GRCh38
rs116104889 Chr.7: 75535273-75535273 on Build GRCh38
rs782667457 Chr.7: 75535215-75535215 on Build GRCh38
rs964949490 Chr.7: 75535169-75535169 on Build GRCh38
rs782758895 Chr.7: 75535047-75535047 on Build GRCh38
rs1009677348 Chr.7: 75535049-75535049 on Build GRCh38
rs11976516 Chr.7: 75535189-75535189 on Build GRCh38
rs982978351 Chr.7: 75535193-75535193 on Build GRCh38
rs908195010 Chr.7: 75535242-75535242 on Build GRCh38
rs931075980 Chr.7: 75534893-75534893 on Build GRCh38
rs779498072 Chr.7: 75534997-75534997 on Build GRCh38
rs781791803 Chr.7: 75535301-75535301 on Build GRCh38
rs782156967 Chr.7: 75534889-75534889 on Build GRCh38
rs987497137 Chr.7: 75535095-75535095 on Build GRCh38
rs746484138 Chr.7: 75535117-75535117 on Build GRCh38
rs943506444 Chr.7: 75534871-75534871 on Build GRCh38
rs886798537 Chr.7: 75534930-75534930 on Build GRCh38
rs949040005 Chr.7: 75535247-75535247 on Build GRCh38
rs1018234276 Chr.7: 75535153-75535153 on Build GRCh38
rs1046781598 Chr.7: 75534968-75534968 on Build GRCh38

View Full Product Details