Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75881749-75882260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBDD2
Entrez Gene ID
57414
Gene Name
rhomboid domain containing 2
Gene Aliases
NPD007, RHBDL7
UniGene
Hs.488827
Gene Chromosome Location
Chr.7: 75878999-75888926 on Build GRCh38
Transcripts
NM_001040456.1 NM_001040457.1 XM_005250511.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6222240 c.565G>A p.G189R Chr.7: 75882215-75882215 on Build GRCh38
5873082 c.442G>C p.V148L Chr.7: 75882092-75882092 on Build GRCh38
1264132 c.510C>A p.L170L Chr.7: 75882160-75882160 on Build GRCh38
3995700 c.310G>A p.V104M Chr.7: 75881960-75881960 on Build GRCh38
4149347 c.445C>T p.R149C Chr.7: 75882095-75882095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782792779 Chr.7: 75881812-75881812 on Build GRCh38
rs184419820 Chr.7: 75881815-75881815 on Build GRCh38
rs368904185 Chr.7: 75881920-75881920 on Build GRCh38
rs781994261 Chr.7: 75881946-75881946 on Build GRCh38
rs782567905 Chr.7: 75881940-75881940 on Build GRCh38
rs782242747 Chr.7: 75881952-75881952 on Build GRCh38
rs576016970 Chr.7: 75882210-75882210 on Build GRCh38
rs112869815 Chr.7: 75882226-75882226 on Build GRCh38
rs782324035 Chr.7: 75882204-75882204 on Build GRCh38
rs999199888 Chr.7: 75881775-75881775 on Build GRCh38
rs782677037 Chr.7: 75882002-75882002 on Build GRCh38
rs782484425 Chr.7: 75881990-75881990 on Build GRCh38
rs781795798 Chr.7: 75881987-75881987 on Build GRCh38
rs782025133 Chr.7: 75881876-75881876 on Build GRCh38
rs782336247 Chr.7: 75882035-75882035 on Build GRCh38
rs782435874 Chr.7: 75882163-75882163 on Build GRCh38
rs376681954 Chr.7: 75882121-75882121 on Build GRCh38
rs916590734 Chr.7: 75882160-75882160 on Build GRCh38
rs375439958 Chr.7: 75881839-75881839 on Build GRCh38
rs201516311 Chr.7: 75881885-75881885 on Build GRCh38
rs782418470 Chr.7: 75881861-75881861 on Build GRCh38
rs11547498 Chr.7: 75881904-75881904 on Build GRCh38
rs782319607 Chr.7: 75882174-75882174 on Build GRCh38
rs781825583 Chr.7: 75882233-75882233 on Build GRCh38
rs199992690 Chr.7: 75881903-75881903 on Build GRCh38
rs782653770 Chr.7: 75881859-75881859 on Build GRCh38
rs11547500 Chr.7: 75881869-75881869 on Build GRCh38
rs558475589 Chr.7: 75881968-75881968 on Build GRCh38
rs571647764 Chr.7: 75882006-75882006 on Build GRCh38
rs782769048 Chr.7: 75882091-75882091 on Build GRCh38
rs782255329 Chr.7: 75882158-75882158 on Build GRCh38
rs928236647 Chr.7: 75881888-75881888 on Build GRCh38
rs11547499 Chr.7: 75882125-75882125 on Build GRCh38
rs200252846 Chr.7: 75881887-75881887 on Build GRCh38
rs781826201 Chr.7: 75882092-75882092 on Build GRCh38
rs529140175 Chr.7: 75881775-75881775 on Build GRCh38
rs781801731 Chr.7: 75882214-75882214 on Build GRCh38
rs1005737713 Chr.7: 75882019-75882019 on Build GRCh38
rs1028026846 Chr.7: 75882010-75882010 on Build GRCh38
rs370440703 Chr.7: 75882164-75882164 on Build GRCh38
rs782471564 Chr.7: 75881939-75881939 on Build GRCh38
rs782633523 Chr.7: 75881822-75881822 on Build GRCh38
rs781904491 Chr.7: 75881980-75881980 on Build GRCh38
rs372506880 Chr.7: 75881984-75881984 on Build GRCh38
rs782325685 Chr.7: 75882009-75882009 on Build GRCh38
rs377724977 Chr.7: 75882154-75882154 on Build GRCh38
rs782507703 Chr.7: 75881819-75881819 on Build GRCh38
rs782428802 Chr.7: 75881905-75881905 on Build GRCh38
rs564955340 Chr.7: 75881793-75881793 on Build GRCh38
rs782795317 Chr.7: 75882059-75882059 on Build GRCh38
rs782584692 Chr.7: 75881841-75881841 on Build GRCh38
rs782079348 Chr.7: 75881892-75881892 on Build GRCh38
rs782530824 Chr.7: 75882109-75882109 on Build GRCh38
rs189062618 Chr.7: 75882145-75882145 on Build GRCh38
rs782153688 Chr.7: 75881898-75881898 on Build GRCh38
rs781939054 Chr.7: 75882087-75882087 on Build GRCh38
rs782358081 Chr.7: 75881853-75881853 on Build GRCh38
rs782615316 Chr.7: 75881994-75881994 on Build GRCh38
rs201342904 Chr.7: 75881798-75881798 on Build GRCh38
rs782152623 Chr.7: 75882208-75882208 on Build GRCh38
rs267601569 Chr.7: 75882114-75882114 on Build GRCh38
rs782180895 Chr.7: 75881843-75881843 on Build GRCh38
rs782231378 Chr.7: 75881943-75881943 on Build GRCh38
rs782776001 Chr.7: 75881894-75881894 on Build GRCh38
rs375029327 Chr.7: 75882016-75882016 on Build GRCh38
rs782392840 Chr.7: 75882192-75882192 on Build GRCh38
rs782764704 Chr.7: 75881979-75881979 on Build GRCh38
rs782223102 Chr.7: 75882022-75882022 on Build GRCh38
rs368034799 Chr.7: 75881860-75881860 on Build GRCh38
rs781820156 Chr.7: 75881818-75881818 on Build GRCh38
rs781980649 Chr.7: 75882203-75882203 on Build GRCh38
rs1044173390 Chr.7: 75882184-75882184 on Build GRCh38
rs368023673 Chr.7: 75882227-75882227 on Build GRCh38
rs928185849 Chr.7: 75882142-75882142 on Build GRCh38
rs781895731 Chr.7: 75881834-75881834 on Build GRCh38
rs566315161 Chr.7: 75882102-75882102 on Build GRCh38
rs782251279 Chr.7: 75882001-75882001 on Build GRCh38
rs782749616 Chr.7: 75882096-75882096 on Build GRCh38
rs782071802 Chr.7: 75881884-75881884 on Build GRCh38
rs375383259 Chr.7: 75881960-75881960 on Build GRCh38
rs782161902 Chr.7: 75881802-75881802 on Build GRCh38
rs782325362 Chr.7: 75881796-75881796 on Build GRCh38
rs782579148 Chr.7: 75882111-75882111 on Build GRCh38
rs1031330739 Chr.7: 75881777-75881777 on Build GRCh38
rs372131603 Chr.7: 75881929-75881929 on Build GRCh38
rs782412900 Chr.7: 75881959-75881959 on Build GRCh38
rs202239956 Chr.7: 75881981-75881981 on Build GRCh38
rs371547840 Chr.7: 75882130-75882130 on Build GRCh38
rs782349445 Chr.7: 75881945-75881945 on Build GRCh38
rs369162750 Chr.7: 75882047-75882047 on Build GRCh38
rs782272787 Chr.7: 75882005-75882005 on Build GRCh38
rs370960130 Chr.7: 75881795-75881795 on Build GRCh38
rs781851797 Chr.7: 75881896-75881896 on Build GRCh38
rs895222390 Chr.7: 75881928-75881928 on Build GRCh38
rs374702528 Chr.7: 75882175-75882175 on Build GRCh38
rs10255642 Chr.7: 75881816-75881816 on Build GRCh38
rs782079047 Chr.7: 75881971-75881971 on Build GRCh38
rs782170497 Chr.7: 75882051-75882051 on Build GRCh38
rs782092640 Chr.7: 75882186-75882186 on Build GRCh38
rs782275756 Chr.7: 75881930-75881930 on Build GRCh38
rs374653359 Chr.7: 75882094-75882094 on Build GRCh38
rs782392008 Chr.7: 75881914-75881914 on Build GRCh38
rs782059741 Chr.7: 75882088-75882088 on Build GRCh38
rs782246048 Chr.7: 75881891-75881891 on Build GRCh38
rs782203002 Chr.7: 75882170-75882170 on Build GRCh38
rs782186764 Chr.7: 75881814-75881814 on Build GRCh38
rs554427510 Chr.7: 75882167-75882167 on Build GRCh38
rs782250773 Chr.7: 75881787-75881787 on Build GRCh38

View Full Product Details