Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 80662850-80663344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD36
Entrez Gene ID
948
Gene Name
CD36 molecule
Gene Aliases
BDPLT10, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
UniGene
Hs.120949
Gene Chromosome Location
Chr.7: 80602188-80679277 on Build GRCh38
Transcripts
NM_000072.3 NM_001001547.2 NM_001001548.2 NM_001127443.1 NM_001127444.1 NM_001289908.1 NM_001289909.1 NM_001289911.1 NR_110501.1 XM_005250713.1 XM_005250714.1 XM_005250715.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4971957 c.522T>C p.T174T Chr.7: 80663082-80663082 on Build GRCh38
4171687 c.482T>G p.L161R Chr.7: 80663042-80663042 on Build GRCh38
1092060 c.437C>T p.S146F Chr.7: 80662997-80662997 on Build GRCh38
1092066 c.543C>T p.G181G Chr.7: 80663103-80663103 on Build GRCh38
4670475 c.526A>C p.R176R Chr.7: 80663086-80663086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770779882 Chr.7: 80663115-80663116 on Build GRCh38
rs750284750 Chr.7: 80662978-80662978 on Build GRCh38
rs141680676 Chr.7: 80663151-80663151 on Build GRCh38
rs746071954 Chr.7: 80663100-80663100 on Build GRCh38
rs543785881 Chr.7: 80663016-80663016 on Build GRCh38
rs774363575 Chr.7: 80662955-80662955 on Build GRCh38
rs761625379 Chr.7: 80663029-80663029 on Build GRCh38
rs746995788 Chr.7: 80663160-80663160 on Build GRCh38
rs369405900 Chr.7: 80663054-80663054 on Build GRCh38
rs771055875 Chr.7: 80663176-80663176 on Build GRCh38
rs765206579 Chr.7: 80663169-80663170 on Build GRCh38
rs766936015 Chr.7: 80663060-80663060 on Build GRCh38
rs138897347 Chr.7: 80663110-80663110 on Build GRCh38
rs752295941 Chr.7: 80663073-80663073 on Build GRCh38
rs751206342 Chr.7: 80663211-80663214 on Build GRCh38
rs1048378540 Chr.7: 80663175-80663175 on Build GRCh38
rs762544085 Chr.7: 80663208-80663209 on Build GRCh38
rs774272468 Chr.7: 80663116-80663116 on Build GRCh38
rs780355823 Chr.7: 80663159-80663159 on Build GRCh38
rs1043394636 Chr.7: 80663297-80663297 on Build GRCh38
rs1011360452 Chr.7: 80663318-80663319 on Build GRCh38
rs762295437 Chr.7: 80663221-80663221 on Build GRCh38
rs779110338 Chr.7: 80663006-80663006 on Build GRCh38
rs759083137 Chr.7: 80663120-80663120 on Build GRCh38
rs778280648 Chr.7: 80663087-80663087 on Build GRCh38
rs764024527 Chr.7: 80662962-80662962 on Build GRCh38
rs368967839 Chr.7: 80663009-80663009 on Build GRCh38
rs779975352 Chr.7: 80663104-80663104 on Build GRCh38
rs569959776 Chr.7: 80663166-80663166 on Build GRCh38
rs3212168 Chr.7: 80662919-80662919 on Build GRCh38
rs756813190 Chr.7: 80663219-80663219 on Build GRCh38
rs552727703 Chr.7: 80663260-80663260 on Build GRCh38
rs745684384 Chr.7: 80663196-80663196 on Build GRCh38
rs758101980 Chr.7: 80662979-80662979 on Build GRCh38
rs3173801 Chr.7: 80663243-80663243 on Build GRCh38
rs1057086208 Chr.7: 80663155-80663155 on Build GRCh38
rs759293657 Chr.7: 80663059-80663059 on Build GRCh38
rs746411946 Chr.7: 80662977-80662977 on Build GRCh38
rs534737948 Chr.7: 80663047-80663047 on Build GRCh38
rs201759307 Chr.7: 80663098-80663098 on Build GRCh38
rs775232747 Chr.7: 80663202-80663202 on Build GRCh38
rs376421152 Chr.7: 80663107-80663107 on Build GRCh38
rs903634379 Chr.7: 80663315-80663315 on Build GRCh38
rs753135305 Chr.7: 80663000-80663000 on Build GRCh38
rs771187661 Chr.7: 80662953-80662953 on Build GRCh38
rs774524792 Chr.7: 80663095-80663096 on Build GRCh38
rs760371584 Chr.7: 80663203-80663203 on Build GRCh38
rs5956 Chr.7: 80663133-80663133 on Build GRCh38
rs773093201 Chr.7: 80663034-80663034 on Build GRCh38
rs55921427 Chr.7: 80662990-80662990 on Build GRCh38
rs749477115 Chr.7: 80662952-80662952 on Build GRCh38
rs5957 Chr.7: 80663020-80663020 on Build GRCh38
rs966002749 Chr.7: 80663094-80663094 on Build GRCh38
rs758501276 Chr.7: 80663157-80663157 on Build GRCh38
rs146120263 Chr.7: 80663084-80663084 on Build GRCh38
rs199530093 Chr.7: 80662989-80662989 on Build GRCh38
rs763882436 Chr.7: 80663210-80663211 on Build GRCh38
rs754454509 Chr.7: 80662991-80662991 on Build GRCh38
rs568503917 Chr.7: 80663102-80663102 on Build GRCh38
rs780114238 Chr.7: 80663005-80663006 on Build GRCh38
rs1003803903 Chr.7: 80663291-80663291 on Build GRCh38
rs781684471 Chr.7: 80662994-80662994 on Build GRCh38
rs781603007 Chr.7: 80662950-80662954 on Build GRCh38
rs772030218 Chr.7: 80663022-80663022 on Build GRCh38
rs760233317 Chr.7: 80663140-80663140 on Build GRCh38
rs982794413 Chr.7: 80663105-80663105 on Build GRCh38
rs749204094 Chr.7: 80663173-80663173 on Build GRCh38
rs760436425 Chr.7: 80663096-80663097 on Build GRCh38
rs1007288161 Chr.7: 80663317-80663317 on Build GRCh38
rs756525492 Chr.7: 80663080-80663080 on Build GRCh38
rs202075359 Chr.7: 80663099-80663099 on Build GRCh38
rs142698840 Chr.7: 80663113-80663113 on Build GRCh38
rs377172957 Chr.7: 80663005-80663005 on Build GRCh38
rs3211900 Chr.7: 80662959-80662959 on Build GRCh38
rs373217513 Chr.7: 80663023-80663023 on Build GRCh38
rs796832659 Chr.7: 80663138-80663139 on Build GRCh38
rs776248038 Chr.7: 80663138-80663139 on Build GRCh38
rs770247327 Chr.7: 80662989-80662999 on Build GRCh38
rs113887411 Chr.7: 80663097-80663097 on Build GRCh38
rs566211840 Chr.7: 80663290-80663290 on Build GRCh38
rs868174973 Chr.7: 80663070-80663071 on Build GRCh38
rs573439520 Chr.7: 80663003-80663003 on Build GRCh38
rs905474415 Chr.7: 80662908-80662908 on Build GRCh38
rs1033691658 Chr.7: 80663088-80663088 on Build GRCh38
rs773964544 Chr.7: 80663190-80663190 on Build GRCh38
rs866211307 Chr.7: 80663137-80663138 on Build GRCh38
rs768926049 Chr.7: 80663071-80663072 on Build GRCh38
rs757827176 Chr.7: 80663150-80663150 on Build GRCh38
rs770839462 Chr.7: 80663111-80663111 on Build GRCh38
rs755124649 Chr.7: 80663170-80663170 on Build GRCh38
rs762850371 Chr.7: 80662909-80662909 on Build GRCh38
rs35398731 Chr.7: 80663002-80663002 on Build GRCh38
rs771588340 Chr.7: 80663198-80663198 on Build GRCh38
rs754132784 Chr.7: 80663091-80663091 on Build GRCh38
rs117684347 Chr.7: 80662974-80662974 on Build GRCh38
rs143150225 Chr.7: 80663132-80663132 on Build GRCh38
rs866508813 Chr.7: 80662929-80662929 on Build GRCh38
rs3211901 Chr.7: 80662988-80662988 on Build GRCh38
rs138108769 Chr.7: 80662891-80662891 on Build GRCh38
rs372107829 Chr.7: 80662986-80662986 on Build GRCh38
rs530761892 Chr.7: 80663145-80663145 on Build GRCh38
rs201801344 Chr.7: 80663051-80663051 on Build GRCh38
rs750848298 Chr.7: 80663156-80663156 on Build GRCh38
rs752309104 Chr.7: 80663194-80663196 on Build GRCh38
rs749308216 Chr.7: 80663030-80663031 on Build GRCh38
rs769902176 Chr.7: 80663030-80663030 on Build GRCh38
rs776192053 Chr.7: 80663212-80663212 on Build GRCh38
rs757614191 Chr.7: 80662940-80662941 on Build GRCh38
rs776445181 Chr.7: 80662966-80662966 on Build GRCh38

View Full Product Details