Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87411790-87412333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB4
Entrez Gene ID
5244
Gene Name
ATP binding cassette subfamily B member 4
Gene Aliases
ABC21, GBD1, ICP3, MDR2, MDR2/3, MDR3, PFIC-3, PGY3
UniGene
Hs.654403
Gene Chromosome Location
Chr.7: 87399461-87475864 on Build GRCh38
Transcripts
NM_000443.3 NM_018849.2 NM_018850.2 XM_011516308.2 XM_011516309.2 XM_011516310.2 XM_011516311.2 XM_011516312.2 XM_011516313.2 XM_011516315.2 XM_017012323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6456477 c.2793G>A p.V931V Chr.7: 87412024-87412024 on Build GRCh38
1092573 c.2914G>T p.D972Y Chr.7: 87411903-87411903 on Build GRCh38
2155474 c.2921T>C p.I974T Chr.7: 87411896-87411896 on Build GRCh38
4782524 c.2898A>G p.G966G Chr.7: 87411919-87411919 on Build GRCh38
3032543 c.2906G>A p.R969H Chr.7: 87411911-87411911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761037382 Chr.7: 87412063-87412063 on Build GRCh38
rs980645407 Chr.7: 87412301-87412301 on Build GRCh38
rs761780866 Chr.7: 87412067-87412067 on Build GRCh38
rs899969157 Chr.7: 87412152-87412152 on Build GRCh38
rs1000983835 Chr.7: 87412218-87412218 on Build GRCh38
rs891398461 Chr.7: 87412099-87412099 on Build GRCh38
rs764407509 Chr.7: 87412065-87412065 on Build GRCh38
rs562473167 Chr.7: 87412206-87412206 on Build GRCh38
rs747730303 Chr.7: 87411980-87411980 on Build GRCh38
rs565187074 Chr.7: 87412104-87412104 on Build GRCh38
rs771509127 Chr.7: 87412007-87412007 on Build GRCh38
rs756310973 Chr.7: 87412221-87412221 on Build GRCh38
rs61730509 Chr.7: 87412017-87412017 on Build GRCh38
rs1049204734 Chr.7: 87412286-87412286 on Build GRCh38
rs181222840 Chr.7: 87412260-87412260 on Build GRCh38
rs577423887 Chr.7: 87412106-87412106 on Build GRCh38
rs748571017 Chr.7: 87411912-87411912 on Build GRCh38
rs956645578 Chr.7: 87412227-87412227 on Build GRCh38
rs776247303 Chr.7: 87411843-87411843 on Build GRCh38
rs779096226 Chr.7: 87411958-87411958 on Build GRCh38
rs573038020 Chr.7: 87412061-87412061 on Build GRCh38
rs560280155 Chr.7: 87412281-87412281 on Build GRCh38
rs45458995 Chr.7: 87412241-87412241 on Build GRCh38
rs769304314 Chr.7: 87411989-87411989 on Build GRCh38
rs1025037904 Chr.7: 87412250-87412250 on Build GRCh38
rs949265606 Chr.7: 87411836-87411836 on Build GRCh38
rs748739226 Chr.7: 87412000-87412000 on Build GRCh38
rs45550235 Chr.7: 87411868-87411868 on Build GRCh38
rs8187805 Chr.7: 87411869-87411869 on Build GRCh38
rs780856994 Chr.7: 87411973-87411973 on Build GRCh38
rs754722032 Chr.7: 87411963-87411963 on Build GRCh38
rs772833262 Chr.7: 87411993-87411993 on Build GRCh38
rs570755181 Chr.7: 87411833-87411833 on Build GRCh38
rs771067939 Chr.7: 87412282-87412282 on Build GRCh38
rs755126615 Chr.7: 87412290-87412292 on Build GRCh38
rs121918440 Chr.7: 87411948-87411948 on Build GRCh38
rs774846029 Chr.7: 87411886-87411886 on Build GRCh38
rs769251359 Chr.7: 87411855-87411855 on Build GRCh38
rs779829759 Chr.7: 87411957-87411957 on Build GRCh38
rs368692582 Chr.7: 87412083-87412083 on Build GRCh38
rs544592566 Chr.7: 87412121-87412121 on Build GRCh38
rs17149542 Chr.7: 87411828-87411828 on Build GRCh38
rs201498350 Chr.7: 87412045-87412045 on Build GRCh38
rs1028092904 Chr.7: 87411981-87411981 on Build GRCh38
rs372187960 Chr.7: 87412084-87412084 on Build GRCh38
rs575565764 Chr.7: 87411918-87411918 on Build GRCh38
rs752916287 Chr.7: 87411911-87411911 on Build GRCh38
rs767899972 Chr.7: 87411906-87411906 on Build GRCh38
rs537754815 Chr.7: 87411846-87411846 on Build GRCh38
rs966038646 Chr.7: 87412170-87412170 on Build GRCh38
rs759787957 Chr.7: 87411894-87411894 on Build GRCh38
rs886042562 Chr.7: 87411984-87411984 on Build GRCh38
rs766779358 Chr.7: 87411874-87411874 on Build GRCh38
rs767055170 Chr.7: 87411892-87411893 on Build GRCh38
rs774697455 Chr.7: 87412008-87412008 on Build GRCh38
rs758428251 Chr.7: 87411922-87411922 on Build GRCh38

View Full Product Details