Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87835979-87836519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A40
Entrez Gene ID
55972
Gene Name
solute carrier family 25 member 40
Gene Aliases
MCFP
UniGene
Hs.208414
Gene Chromosome Location
Chr.7: 87834499-87876377 on Build GRCh38
Transcripts
NM_018843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1092661 c.940T>G p.C314G Chr.7: 87836326-87836326 on Build GRCh38
1092660 c.1000C>A p.R334R Chr.7: 87836266-87836266 on Build GRCh38
1092659 c.1000C>T p.R334* Chr.7: 87836266-87836266 on Build GRCh38
5962521 c.922A>G p.I308V Chr.7: 87836344-87836344 on Build GRCh38
5737984 c.932C>T p.A311V Chr.7: 87836334-87836334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376423146 Chr.7: 87836241-87836241 on Build GRCh38
rs1025153333 Chr.7: 87836023-87836023 on Build GRCh38
rs752371624 Chr.7: 87836445-87836445 on Build GRCh38
rs760581054 Chr.7: 87836394-87836394 on Build GRCh38
rs771749379 Chr.7: 87836278-87836278 on Build GRCh38
rs984570654 Chr.7: 87836434-87836434 on Build GRCh38
rs542218076 Chr.7: 87836216-87836216 on Build GRCh38
rs748567367 Chr.7: 87836322-87836322 on Build GRCh38
rs777777850 Chr.7: 87836237-87836237 on Build GRCh38
rs763297221 Chr.7: 87836365-87836369 on Build GRCh38
rs926672582 Chr.7: 87836067-87836067 on Build GRCh38
rs950794753 Chr.7: 87836468-87836468 on Build GRCh38
rs917737054 Chr.7: 87836218-87836218 on Build GRCh38
rs367840643 Chr.7: 87836031-87836031 on Build GRCh38
rs764550764 Chr.7: 87836303-87836303 on Build GRCh38
rs987314443 Chr.7: 87836174-87836174 on Build GRCh38
rs761270470 Chr.7: 87836021-87836021 on Build GRCh38
rs537357458 Chr.7: 87836123-87836123 on Build GRCh38
rs1039104000 Chr.7: 87836214-87836214 on Build GRCh38
rs371373274 Chr.7: 87836423-87836423 on Build GRCh38
rs766686631 Chr.7: 87836223-87836223 on Build GRCh38
rs545972367 Chr.7: 87836357-87836357 on Build GRCh38
rs953631890 Chr.7: 87836063-87836063 on Build GRCh38
rs980719546 Chr.7: 87836061-87836061 on Build GRCh38
rs778524158 Chr.7: 87836262-87836262 on Build GRCh38
rs769957787 Chr.7: 87836232-87836238 on Build GRCh38
rs759663615 Chr.7: 87836350-87836350 on Build GRCh38
rs1032923165 Chr.7: 87836012-87836012 on Build GRCh38
rs987291585 Chr.7: 87836487-87836487 on Build GRCh38
rs1000161306 Chr.7: 87836325-87836325 on Build GRCh38
rs146277655 Chr.7: 87836265-87836265 on Build GRCh38
rs919335914 Chr.7: 87836151-87836151 on Build GRCh38
rs753835874 Chr.7: 87836360-87836360 on Build GRCh38
rs200456051 Chr.7: 87836333-87836333 on Build GRCh38
rs779673319 Chr.7: 87836279-87836279 on Build GRCh38
rs766223845 Chr.7: 87836395-87836395 on Build GRCh38
rs746577980 Chr.7: 87836293-87836293 on Build GRCh38
rs576144775 Chr.7: 87836120-87836120 on Build GRCh38
rs779693303 Chr.7: 87836389-87836389 on Build GRCh38
rs531765370 Chr.7: 87836416-87836416 on Build GRCh38
rs753649991 Chr.7: 87836246-87836246 on Build GRCh38
rs1000616551 Chr.7: 87836033-87836033 on Build GRCh38
rs774650967 Chr.7: 87836393-87836395 on Build GRCh38
rs757106666 Chr.7: 87836255-87836255 on Build GRCh38
rs1041774938 Chr.7: 87836290-87836290 on Build GRCh38
rs35067684 Chr.7: 87836227-87836227 on Build GRCh38
rs764240725 Chr.7: 87836381-87836381 on Build GRCh38
rs3216917 Chr.7: 87836221-87836223 on Build GRCh38
rs777202468 Chr.7: 87836318-87836318 on Build GRCh38
rs763222864 Chr.7: 87836222-87836222 on Build GRCh38
rs775429538 Chr.7: 87836349-87836349 on Build GRCh38
rs973817328 Chr.7: 87836427-87836427 on Build GRCh38
rs776988636 Chr.7: 87836202-87836202 on Build GRCh38
rs373485583 Chr.7: 87836376-87836376 on Build GRCh38
rs201263895 Chr.7: 87836256-87836256 on Build GRCh38
rs971827442 Chr.7: 87836119-87836119 on Build GRCh38
rs192706021 Chr.7: 87836212-87836212 on Build GRCh38
rs770221466 Chr.7: 87836328-87836328 on Build GRCh38
rs943298258 Chr.7: 87836208-87836208 on Build GRCh38
rs1026368081 Chr.7: 87836459-87836459 on Build GRCh38
rs937901560 Chr.7: 87836195-87836195 on Build GRCh38
rs755009887 Chr.7: 87836235-87836235 on Build GRCh38
rs963785016 Chr.7: 87836076-87836076 on Build GRCh38
rs929088072 Chr.7: 87836170-87836170 on Build GRCh38
rs951896191 Chr.7: 87836425-87836425 on Build GRCh38
rs951724367 Chr.7: 87836137-87836137 on Build GRCh38
rs956857002 Chr.7: 87836013-87836013 on Build GRCh38
rs751730872 Chr.7: 87836227-87836227 on Build GRCh38
rs753193173 Chr.7: 87836006-87836006 on Build GRCh38
rs557964714 Chr.7: 87836099-87836099 on Build GRCh38
rs555661586 Chr.7: 87836203-87836203 on Build GRCh38
rs778604953 Chr.7: 87836306-87836306 on Build GRCh38
rs763379005 Chr.7: 87836347-87836347 on Build GRCh38
rs369225044 Chr.7: 87836373-87836373 on Build GRCh38
rs201654319 Chr.7: 87836374-87836374 on Build GRCh38
rs542153689 Chr.7: 87836266-87836266 on Build GRCh38
rs746578670 Chr.7: 87836395-87836395 on Build GRCh38
rs766776279 Chr.7: 87836349-87836349 on Build GRCh38

View Full Product Details