Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 95307854-95308384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PON1
Entrez Gene ID
5444
Gene Name
paraoxonase 1
Gene Aliases
ESA, MVCD5, PON
UniGene
Hs.370995
Gene Chromosome Location
Chr.7: 95298357-95324572 on Build GRCh38
Transcripts
NM_000446.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
150330 c.575A>G p.Q192R Chr.7: 95308134-95308134 on Build GRCh38
5738019 c.539G>A p.G180D Chr.7: 95308170-95308170 on Build GRCh38
4956648 c.609G>A p.S203S Chr.7: 95308100-95308100 on Build GRCh38
3412511 c.597T>A p.G199G Chr.7: 95308112-95308112 on Build GRCh38
5645270 c.595G>T p.G199C Chr.7: 95308114-95308114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187549855 Chr.7: 95308328-95308328 on Build GRCh38
rs895263743 Chr.7: 95308298-95308298 on Build GRCh38
rs766568477 Chr.7: 95308143-95308143 on Build GRCh38
rs3917594 Chr.7: 95308127-95308127 on Build GRCh38
rs1009794708 Chr.7: 95307958-95307958 on Build GRCh38
rs769851777 Chr.7: 95308128-95308128 on Build GRCh38
rs755022126 Chr.7: 95308322-95308322 on Build GRCh38
rs1024000122 Chr.7: 95308146-95308146 on Build GRCh38
rs374067811 Chr.7: 95308019-95308019 on Build GRCh38
rs767932117 Chr.7: 95308178-95308178 on Build GRCh38
rs772121280 Chr.7: 95307962-95307962 on Build GRCh38
rs984109911 Chr.7: 95308037-95308037 on Build GRCh38
rs778676808 Chr.7: 95308254-95308254 on Build GRCh38
rs761921244 Chr.7: 95308016-95308016 on Build GRCh38
rs1012981993 Chr.7: 95308145-95308145 on Build GRCh38
rs755586417 Chr.7: 95308216-95308216 on Build GRCh38
rs763222419 Chr.7: 95308135-95308135 on Build GRCh38
rs75997226 Chr.7: 95308212-95308212 on Build GRCh38
rs921194093 Chr.7: 95308295-95308295 on Build GRCh38
rs904066390 Chr.7: 95308126-95308126 on Build GRCh38
rs3917540 Chr.7: 95308334-95308334 on Build GRCh38
rs962865343 Chr.7: 95308231-95308231 on Build GRCh38
rs1033834050 Chr.7: 95308234-95308234 on Build GRCh38
rs763317810 Chr.7: 95308035-95308035 on Build GRCh38
rs908414061 Chr.7: 95307880-95307880 on Build GRCh38
rs912635040 Chr.7: 95308101-95308101 on Build GRCh38
rs778909584 Chr.7: 95308090-95308090 on Build GRCh38
rs370355032 Chr.7: 95308081-95308081 on Build GRCh38
rs372509316 Chr.7: 95308260-95308260 on Build GRCh38
rs753190870 Chr.7: 95308202-95308202 on Build GRCh38
rs755603793 Chr.7: 95308077-95308077 on Build GRCh38
rs141624867 Chr.7: 95308100-95308100 on Build GRCh38
rs751911649 Chr.7: 95308054-95308054 on Build GRCh38
rs143592425 Chr.7: 95308073-95308073 on Build GRCh38
rs971132600 Chr.7: 95308183-95308183 on Build GRCh38
rs778908135 Chr.7: 95308187-95308202 on Build GRCh38
rs375312050 Chr.7: 95307945-95307945 on Build GRCh38
rs550240333 Chr.7: 95308253-95308253 on Build GRCh38
rs759858395 Chr.7: 95308171-95308171 on Build GRCh38
rs961287183 Chr.7: 95308078-95308078 on Build GRCh38
rs961317879 Chr.7: 95308330-95308330 on Build GRCh38
rs147993776 Chr.7: 95308088-95308088 on Build GRCh38
rs112536291 Chr.7: 95308224-95308224 on Build GRCh38
rs182868805 Chr.7: 95308289-95308289 on Build GRCh38
rs757017065 Chr.7: 95308229-95308229 on Build GRCh38
rs148452713 Chr.7: 95308106-95308106 on Build GRCh38
rs1054106813 Chr.7: 95307902-95307902 on Build GRCh38
rs752182948 Chr.7: 95308074-95308074 on Build GRCh38
rs111313397 Chr.7: 95308243-95308243 on Build GRCh38
rs375899405 Chr.7: 95308256-95308256 on Build GRCh38
rs1043000099 Chr.7: 95308167-95308167 on Build GRCh38
rs1003639150 Chr.7: 95308011-95308011 on Build GRCh38
rs564064745 Chr.7: 95308018-95308018 on Build GRCh38
rs80019660 Chr.7: 95308107-95308107 on Build GRCh38
rs763727586 Chr.7: 95308217-95308217 on Build GRCh38
rs766660411 Chr.7: 95308048-95308048 on Build GRCh38
rs529138557 Chr.7: 95308007-95308007 on Build GRCh38
rs3917541 Chr.7: 95308312-95308312 on Build GRCh38
rs570987833 Chr.7: 95308327-95308327 on Build GRCh38
rs750607463 Chr.7: 95308027-95308027 on Build GRCh38
rs755532074 Chr.7: 95308068-95308068 on Build GRCh38
rs1003998564 Chr.7: 95308203-95308203 on Build GRCh38
rs527346006 Chr.7: 95307930-95307930 on Build GRCh38
rs777303791 Chr.7: 95308079-95308079 on Build GRCh38
rs662 Chr.7: 95308134-95308134 on Build GRCh38
rs906656805 Chr.7: 95308009-95308009 on Build GRCh38
rs898078377 Chr.7: 95308358-95308358 on Build GRCh38

View Full Product Details