Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 97733467-97733974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAC1
Entrez Gene ID
6863
Gene Name
tachykinin precursor 1
Gene Aliases
Hs.2563, NK2, NKNA, NPK, TAC2
UniGene
Hs.2563
Gene Chromosome Location
Chr.7: 97731959-97740472 on Build GRCh38
Transcripts
NM_003182.2 NM_013996.2 NM_013997.2 NM_013998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4421223 c.204G>T p.M68I Chr.7: 97733803-97733803 on Build GRCh38
748388 c.126G>A p.E42E Chr.7: 97733725-97733725 on Build GRCh38
7274941 c.159G>T p.Q53H Chr.7: 97733758-97733758 on Build GRCh38
1623340 c.152_161del10 p.L51fs*14 Chr.7: 97733751-97733760 on Build GRCh38
1496704 c.145G>C p.E49Q Chr.7: 97733744-97733744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755654238 Chr.7: 97733720-97733720 on Build GRCh38
rs952397520 Chr.7: 97733944-97733944 on Build GRCh38
rs774214302 Chr.7: 97733676-97733676 on Build GRCh38
rs985424586 Chr.7: 97733834-97733834 on Build GRCh38
rs970529401 Chr.7: 97733531-97733531 on Build GRCh38
rs199513408 Chr.7: 97733866-97733866 on Build GRCh38
rs865867818 Chr.7: 97733831-97733831 on Build GRCh38
rs547564259 Chr.7: 97733943-97733943 on Build GRCh38
rs777362022 Chr.7: 97733832-97733832 on Build GRCh38
rs940077330 Chr.7: 97733557-97733557 on Build GRCh38
rs1023457683 Chr.7: 97733517-97733517 on Build GRCh38
rs1041207121 Chr.7: 97733629-97733629 on Build GRCh38
rs766687646 Chr.7: 97733770-97733770 on Build GRCh38
rs911490642 Chr.7: 97733909-97733909 on Build GRCh38
rs753573932 Chr.7: 97733851-97733851 on Build GRCh38
rs778727252 Chr.7: 97733737-97733737 on Build GRCh38
rs556506847 Chr.7: 97733577-97733577 on Build GRCh38
rs148136566 Chr.7: 97733788-97733788 on Build GRCh38
rs564214397 Chr.7: 97733647-97733647 on Build GRCh38
rs766706281 Chr.7: 97733699-97733699 on Build GRCh38
rs971094584 Chr.7: 97733635-97733635 on Build GRCh38
rs994145838 Chr.7: 97733933-97733933 on Build GRCh38
rs970583110 Chr.7: 97733651-97733651 on Build GRCh38
rs1014685305 Chr.7: 97733661-97733661 on Build GRCh38
rs771238137 Chr.7: 97733768-97733768 on Build GRCh38
rs756883577 Chr.7: 97733853-97733853 on Build GRCh38
rs201586107 Chr.7: 97733749-97733749 on Build GRCh38
rs750635590 Chr.7: 97733691-97733691 on Build GRCh38
rs527929288 Chr.7: 97733906-97733906 on Build GRCh38
rs1034479870 Chr.7: 97733594-97733594 on Build GRCh38
rs752015298 Chr.7: 97733705-97733705 on Build GRCh38
rs763363233 Chr.7: 97733695-97733695 on Build GRCh38
rs745814826 Chr.7: 97733739-97733739 on Build GRCh38
rs528839463 Chr.7: 97733754-97733754 on Build GRCh38
rs780592590 Chr.7: 97733709-97733709 on Build GRCh38
rs768647367 Chr.7: 97733681-97733681 on Build GRCh38
rs370538589 Chr.7: 97733728-97733728 on Build GRCh38
rs752134155 Chr.7: 97733847-97733848 on Build GRCh38
rs1043683232 Chr.7: 97733714-97733714 on Build GRCh38
rs181650984 Chr.7: 97733641-97733641 on Build GRCh38
rs897860011 Chr.7: 97733928-97733928 on Build GRCh38
rs1012615054 Chr.7: 97733559-97733559 on Build GRCh38
rs772094189 Chr.7: 97733744-97733744 on Build GRCh38
rs1014368806 Chr.7: 97733895-97733895 on Build GRCh38
rs902283379 Chr.7: 97733632-97733632 on Build GRCh38
rs894250013 Chr.7: 97733554-97733554 on Build GRCh38
rs769751868 Chr.7: 97733942-97733942 on Build GRCh38
rs376044942 Chr.7: 97733865-97733865 on Build GRCh38
rs759604110 Chr.7: 97733916-97733916 on Build GRCh38
rs150265957 Chr.7: 97733670-97733670 on Build GRCh38
rs533035697 Chr.7: 97733659-97733659 on Build GRCh38
rs2189306 Chr.7: 97733625-97733625 on Build GRCh38
rs972836064 Chr.7: 97733755-97733755 on Build GRCh38
rs919926631 Chr.7: 97733578-97733578 on Build GRCh38
rs770635838 Chr.7: 97733736-97733736 on Build GRCh38
rs753253189 Chr.7: 97733797-97733797 on Build GRCh38
rs1023508317 Chr.7: 97733648-97733648 on Build GRCh38
rs761943477 Chr.7: 97733752-97733752 on Build GRCh38
rs752304994 Chr.7: 97733719-97733719 on Build GRCh38
rs755744209 Chr.7: 97733830-97733830 on Build GRCh38
rs973170382 Chr.7: 97733558-97733558 on Build GRCh38
rs776642530 Chr.7: 97733683-97733683 on Build GRCh38
rs371142680 Chr.7: 97733675-97733675 on Build GRCh38
rs755474751 Chr.7: 97733706-97733706 on Build GRCh38
rs763171892 Chr.7: 97733764-97733764 on Build GRCh38
rs539478451 Chr.7: 97733542-97733542 on Build GRCh38
rs11554502 Chr.7: 97733771-97733771 on Build GRCh38
rs375904849 Chr.7: 97733687-97733687 on Build GRCh38
rs1024424510 Chr.7: 97733581-97733581 on Build GRCh38
rs759207877 Chr.7: 97733765-97733765 on Build GRCh38
rs893464675 Chr.7: 97733646-97733646 on Build GRCh38
rs1045745840 Chr.7: 97733899-97733899 on Build GRCh38
rs768180091 Chr.7: 97733791-97733791 on Build GRCh38
rs748936107 Chr.7: 97733733-97733733 on Build GRCh38
rs373629494 Chr.7: 97733859-97733859 on Build GRCh38
rs552916415 Chr.7: 97733638-97733638 on Build GRCh38
rs911438136 Chr.7: 97733622-97733622 on Build GRCh38
rs200641731 Chr.7: 97733779-97733779 on Build GRCh38
rs537118160 Chr.7: 97733613-97733613 on Build GRCh38
rs928730121 Chr.7: 97733547-97733547 on Build GRCh38
rs576323666 Chr.7: 97733590-97733590 on Build GRCh38
rs541216858 Chr.7: 97733645-97733645 on Build GRCh38

View Full Product Details