Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 132124006-132124517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXNA4
Entrez Gene ID
91584
Gene Name
plexin A4
Gene Aliases
FAYV2820, PLEXA4, PLXNA4A, PLXNA4B, PRO34003
UniGene
Hs.511454
Gene Chromosome Location
Chr.7: 132123332-132648688 on Build GRCh38
Transcripts
NM_020911.1 XM_005250686.4 XM_006716171.3 XM_017012779.1

SNP Detail

SNP ID Chromosome Location
rs11542101 Chr.7: 132124415-132124415 on Build GRCh38
rs865809312 Chr.7: 132124127-132124127 on Build GRCh38
rs898703888 Chr.7: 132124144-132124144 on Build GRCh38
rs923699238 Chr.7: 132124260-132124260 on Build GRCh38
rs981162868 Chr.7: 132124239-132124239 on Build GRCh38
rs7795466 Chr.7: 132124068-132124068 on Build GRCh38
rs986530510 Chr.7: 132124267-132124267 on Build GRCh38
rs559531062 Chr.7: 132124293-132124293 on Build GRCh38
rs1030858798 Chr.7: 132124460-132124460 on Build GRCh38
rs759206410 Chr.7: 132124428-132124428 on Build GRCh38
rs750325452 Chr.7: 132124303-132124304 on Build GRCh38
rs990338827 Chr.7: 132124429-132124429 on Build GRCh38
rs570174894 Chr.7: 132124347-132124347 on Build GRCh38
rs559743455 Chr.7: 132124206-132124206 on Build GRCh38
rs1012832799 Chr.7: 132124399-132124399 on Build GRCh38
rs1025246917 Chr.7: 132124378-132124378 on Build GRCh38
rs897241835 Chr.7: 132124309-132124309 on Build GRCh38
rs979693628 Chr.7: 132124457-132124457 on Build GRCh38
rs191047005 Chr.7: 132124275-132124275 on Build GRCh38
rs543283590 Chr.7: 132124150-132124150 on Build GRCh38
rs375575755 Chr.7: 132124345-132124345 on Build GRCh38
rs961496167 Chr.7: 132124124-132124124 on Build GRCh38
rs143771503 Chr.7: 132124295-132124295 on Build GRCh38
rs924367830 Chr.7: 132124249-132124249 on Build GRCh38
rs894521502 Chr.7: 132124380-132124380 on Build GRCh38
rs990000763 Chr.7: 132124268-132124268 on Build GRCh38
rs996035466 Chr.7: 132124330-132124330 on Build GRCh38
rs981110448 Chr.7: 132124406-132124406 on Build GRCh38
rs144973917 Chr.7: 132124176-132124176 on Build GRCh38
rs113801565 Chr.7: 132124221-132124221 on Build GRCh38
rs116421992 Chr.7: 132124225-132124225 on Build GRCh38
rs1008252653 Chr.7: 132124105-132124105 on Build GRCh38
rs749552370 Chr.7: 132124065-132124065 on Build GRCh38
rs771250759 Chr.7: 132124290-132124290 on Build GRCh38
rs7795964 Chr.7: 132124167-132124167 on Build GRCh38
rs1050722093 Chr.7: 132124042-132124042 on Build GRCh38
rs914398348 Chr.7: 132124474-132124474 on Build GRCh38
rs1023506766 Chr.7: 132124245-132124245 on Build GRCh38
rs773582591 Chr.7: 132124276-132124276 on Build GRCh38
rs879047353 Chr.7: 132124379-132124379 on Build GRCh38
rs7795977 Chr.7: 132124197-132124197 on Build GRCh38
rs956733701 Chr.7: 132124469-132124469 on Build GRCh38
rs1037044324 Chr.7: 132124300-132124300 on Build GRCh38
rs997211414 Chr.7: 132124062-132124062 on Build GRCh38
rs986586251 Chr.7: 132124475-132124475 on Build GRCh38
rs934527778 Chr.7: 132124027-132124027 on Build GRCh38
rs375960501 Chr.7: 132124187-132124188 on Build GRCh38
rs548374357 Chr.7: 132124335-132124335 on Build GRCh38
rs931486121 Chr.7: 132124320-132124320 on Build GRCh38
rs968291550 Chr.7: 132124248-132124248 on Build GRCh38
rs887253652 Chr.7: 132124346-132124346 on Build GRCh38
rs1012483706 Chr.7: 132124363-132124363 on Build GRCh38
rs893059314 Chr.7: 132124060-132124060 on Build GRCh38
rs1047202079 Chr.7: 132124344-132124344 on Build GRCh38
rs866146853 Chr.7: 132124259-132124259 on Build GRCh38
rs13247270 Chr.7: 132124417-132124417 on Build GRCh38
rs1015650511 Chr.7: 132124110-132124110 on Build GRCh38
rs1054749815 Chr.7: 132124041-132124041 on Build GRCh38
rs1023518016 Chr.7: 132124416-132124416 on Build GRCh38
rs149047301 Chr.7: 132124289-132124289 on Build GRCh38
rs866131744 Chr.7: 132124484-132124484 on Build GRCh38
rs1049714702 Chr.7: 132124080-132124080 on Build GRCh38
rs751820036 Chr.7: 132124329-132124329 on Build GRCh38
rs530894929 Chr.7: 132124362-132124362 on Build GRCh38
rs1026156686 Chr.7: 132124171-132124171 on Build GRCh38
rs1035675258 Chr.7: 132124207-132124207 on Build GRCh38
rs891160583 Chr.7: 132124087-132124087 on Build GRCh38
rs935072553 Chr.7: 132124262-132124262 on Build GRCh38
rs1027148987 Chr.7: 132124340-132124340 on Build GRCh38

View Full Product Details