Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138734055-138734521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0A4
Entrez Gene ID
50617
Gene Name
ATPase H+ transporting V0 subunit a4
Gene Aliases
A4, ATP6N1B, ATP6N2, RDRTA2, RTA1C, RTADR, STV1, VPH1, VPP2
UniGene
Hs.98967
Gene Chromosome Location
Chr.7: 138706294-138799839 on Build GRCh38
Transcripts
NM_020632.2 NM_130840.2 NM_130841.2 XM_005250393.1 XM_005250394.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1673586 c.1631C>T p.S544L Chr.7: 138734196-138734196 on Build GRCh38
3633900 c.1577G>A p.W526* Chr.7: 138734250-138734250 on Build GRCh38
6608048 c.1613C>T p.S538L Chr.7: 138734214-138734214 on Build GRCh38
5777713 c.1650C>A p.V550V Chr.7: 138734177-138734177 on Build GRCh38
3878511 c.1614G>A p.S538S Chr.7: 138734213-138734213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746972170 Chr.7: 138734241-138734241 on Build GRCh38
rs778560567 Chr.7: 138734185-138734185 on Build GRCh38
rs776521583 Chr.7: 138734244-138734244 on Build GRCh38
rs781358295 Chr.7: 138734109-138734109 on Build GRCh38
rs61747675 Chr.7: 138734186-138734186 on Build GRCh38
rs915479851 Chr.7: 138734171-138734171 on Build GRCh38
rs770905239 Chr.7: 138734242-138734242 on Build GRCh38
rs201362655 Chr.7: 138734086-138734086 on Build GRCh38
rs369979838 Chr.7: 138734324-138734324 on Build GRCh38
rs201166384 Chr.7: 138734112-138734112 on Build GRCh38
rs764402160 Chr.7: 138734273-138734273 on Build GRCh38
rs753053757 Chr.7: 138734089-138734089 on Build GRCh38
rs1024191682 Chr.7: 138734463-138734463 on Build GRCh38
rs968723621 Chr.7: 138734386-138734386 on Build GRCh38
rs200419423 Chr.7: 138734301-138734301 on Build GRCh38
rs762603854 Chr.7: 138734263-138734263 on Build GRCh38
rs200131246 Chr.7: 138734195-138734195 on Build GRCh38
rs757803112 Chr.7: 138734178-138734178 on Build GRCh38
rs771895516 Chr.7: 138734127-138734127 on Build GRCh38
rs756319723 Chr.7: 138734299-138734299 on Build GRCh38
rs778278013 Chr.7: 138734298-138734300 on Build GRCh38
rs772991958 Chr.7: 138734226-138734226 on Build GRCh38
rs1037692 Chr.7: 138734340-138734340 on Build GRCh38
rs768176257 Chr.7: 138734280-138734280 on Build GRCh38
rs753113224 Chr.7: 138734139-138734139 on Build GRCh38
rs756228070 Chr.7: 138734194-138734194 on Build GRCh38
rs761550045 Chr.7: 138734120-138734120 on Build GRCh38
rs565186611 Chr.7: 138734100-138734100 on Build GRCh38
rs1031284159 Chr.7: 138734421-138734421 on Build GRCh38
rs776229594 Chr.7: 138734387-138734387 on Build GRCh38
rs778213721 Chr.7: 138734093-138734093 on Build GRCh38
rs753232747 Chr.7: 138734133-138734134 on Build GRCh38
rs1030198868 Chr.7: 138734336-138734336 on Build GRCh38
rs201215397 Chr.7: 138734285-138734285 on Build GRCh38
rs369486474 Chr.7: 138734152-138734152 on Build GRCh38
rs759024783 Chr.7: 138734145-138734145 on Build GRCh38
rs199690679 Chr.7: 138734302-138734302 on Build GRCh38
rs778982140 Chr.7: 138734306-138734306 on Build GRCh38
rs764717102 Chr.7: 138734251-138734251 on Build GRCh38
rs759114660 Chr.7: 138734247-138734247 on Build GRCh38
rs764911849 Chr.7: 138734146-138734146 on Build GRCh38
rs115616210 Chr.7: 138734322-138734322 on Build GRCh38
rs781376064 Chr.7: 138734116-138734116 on Build GRCh38
rs775830752 Chr.7: 138734114-138734114 on Build GRCh38
rs760603032 Chr.7: 138734138-138734138 on Build GRCh38
rs377704642 Chr.7: 138734213-138734213 on Build GRCh38
rs961469487 Chr.7: 138734143-138734143 on Build GRCh38
rs992535827 Chr.7: 138734453-138734453 on Build GRCh38
rs147301634 Chr.7: 138734158-138734158 on Build GRCh38
rs545593553 Chr.7: 138734092-138734092 on Build GRCh38
rs746120812 Chr.7: 138734110-138734110 on Build GRCh38
rs1026435 Chr.7: 138734165-138734165 on Build GRCh38
rs547777494 Chr.7: 138734153-138734153 on Build GRCh38
rs759013681 Chr.7: 138734272-138734273 on Build GRCh38
rs745991559 Chr.7: 138734187-138734187 on Build GRCh38
rs954197513 Chr.7: 138734338-138734338 on Build GRCh38
rs771776708 Chr.7: 138734214-138734214 on Build GRCh38
rs371935296 Chr.7: 138734088-138734088 on Build GRCh38
rs916988874 Chr.7: 138734493-138734493 on Build GRCh38
rs369693858 Chr.7: 138734136-138734136 on Build GRCh38
rs955739300 Chr.7: 138734451-138734451 on Build GRCh38
rs576600988 Chr.7: 138734078-138734078 on Build GRCh38
rs752015489 Chr.7: 138734170-138734170 on Build GRCh38
rs185271815 Chr.7: 138734323-138734323 on Build GRCh38
rs182512951 Chr.7: 138734257-138734257 on Build GRCh38
rs202011589 Chr.7: 138734279-138734279 on Build GRCh38
rs199546745 Chr.7: 138734107-138734107 on Build GRCh38
rs587776615 Chr.7: 138734135-138734135 on Build GRCh38
rs755186596 Chr.7: 138734305-138734305 on Build GRCh38
rs991745432 Chr.7: 138734348-138734348 on Build GRCh38
rs772341312 Chr.7: 138734349-138734349 on Build GRCh38
rs751899624 Chr.7: 138734275-138734275 on Build GRCh38
rs748420920 Chr.7: 138734300-138734300 on Build GRCh38
rs150027159 Chr.7: 138734156-138734156 on Build GRCh38

View Full Product Details