Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 141723838-141724350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WEE2
Entrez Gene ID
494551
Gene Name
WEE1 homolog 2 (S. pombe)
Gene Aliases
WEE1B
UniGene
Hs.657927
Gene Chromosome Location
Chr.7: 141708353-141731271 on Build GRCh38
Transcripts
NM_001105558.1 NR_015392.1

Gene Symbol
WEE2-AS1
Entrez Gene ID
285962
Gene Name
WEE2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.17589
Gene Chromosome Location
Chr.7: 141704338-141738230 on Build GRCh38
Transcripts
NM_001105558.1 NR_015392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1086308 c.1150G>A p.A384T Chr.7: 141724204-141724204 on Build GRCh38
1086309 c.1192C>T p.R398C Chr.7: 141724246-141724246 on Build GRCh38
3634302 c.1184G>A p.G395E Chr.7: 141724238-141724238 on Build GRCh38
5779279 c.1039A>T p.I347F Chr.7: 141723952-141723952 on Build GRCh38
6822479 c.1164C>T p.N388N Chr.7: 141724218-141724218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560462527 Chr.7: 141724177-141724177 on Build GRCh38
rs992329984 Chr.7: 141723985-141723985 on Build GRCh38
rs149484954 Chr.7: 141724312-141724312 on Build GRCh38
rs754254300 Chr.7: 141724037-141724037 on Build GRCh38
rs766854560 Chr.7: 141723923-141723923 on Build GRCh38
rs746850260 Chr.7: 141724252-141724252 on Build GRCh38
rs772905831 Chr.7: 141724031-141724031 on Build GRCh38
rs865776217 Chr.7: 141723915-141723915 on Build GRCh38
rs755667095 Chr.7: 141723919-141723919 on Build GRCh38
rs763577677 Chr.7: 141724315-141724315 on Build GRCh38
rs776207184 Chr.7: 141724274-141724274 on Build GRCh38
rs759019678 Chr.7: 141724179-141724179 on Build GRCh38
rs770597791 Chr.7: 141724269-141724269 on Build GRCh38
rs1006918455 Chr.7: 141723908-141723908 on Build GRCh38
rs751782234 Chr.7: 141724197-141724197 on Build GRCh38
rs761551942 Chr.7: 141724021-141724021 on Build GRCh38
rs768605870 Chr.7: 141724098-141724098 on Build GRCh38
rs760169334 Chr.7: 141724032-141724032 on Build GRCh38
rs753086761 Chr.7: 141724055-141724055 on Build GRCh38
rs758665518 Chr.7: 141723972-141723972 on Build GRCh38
rs996543323 Chr.7: 141724225-141724225 on Build GRCh38
rs969094301 Chr.7: 141724046-141724046 on Build GRCh38
rs1028282877 Chr.7: 141724287-141724287 on Build GRCh38
rs767130803 Chr.7: 141724022-141724022 on Build GRCh38
rs765762621 Chr.7: 141724180-141724180 on Build GRCh38
rs755545031 Chr.7: 141724045-141724045 on Build GRCh38
rs550050383 Chr.7: 141724204-141724204 on Build GRCh38
rs373240057 Chr.7: 141723932-141723932 on Build GRCh38
rs769194846 Chr.7: 141724293-141724293 on Build GRCh38
rs771794268 Chr.7: 141724246-141724246 on Build GRCh38
rs531482231 Chr.7: 141724155-141724155 on Build GRCh38
rs137863922 Chr.7: 141723994-141723994 on Build GRCh38
rs748281292 Chr.7: 141723947-141723947 on Build GRCh38
rs374188394 Chr.7: 141723958-141723958 on Build GRCh38
rs775224888 Chr.7: 141723986-141723986 on Build GRCh38
rs756202556 Chr.7: 141724214-141724214 on Build GRCh38
rs543032135 Chr.7: 141723946-141723946 on Build GRCh38
rs747239391 Chr.7: 141724076-141724076 on Build GRCh38
rs749261746 Chr.7: 141723989-141723989 on Build GRCh38
rs552800129 Chr.7: 141724036-141724036 on Build GRCh38
rs747074087 Chr.7: 141723981-141723981 on Build GRCh38
rs535510834 Chr.7: 141724284-141724284 on Build GRCh38
rs376828374 Chr.7: 141724152-141724152 on Build GRCh38
rs778139045 Chr.7: 141724069-141724069 on Build GRCh38
rs778903115 Chr.7: 141724223-141724223 on Build GRCh38
rs764567068 Chr.7: 141724195-141724195 on Build GRCh38
rs759084206 Chr.7: 141724276-141724276 on Build GRCh38
rs753213041 Chr.7: 141724184-141724184 on Build GRCh38
rs771728907 Chr.7: 141724179-141724180 on Build GRCh38
rs762129319 Chr.7: 141724132-141724132 on Build GRCh38
rs760346923 Chr.7: 141724161-141724161 on Build GRCh38
rs769075825 Chr.7: 141723913-141723913 on Build GRCh38
rs748078513 Chr.7: 141724230-141724230 on Build GRCh38
rs757353305 Chr.7: 141724086-141724086 on Build GRCh38
rs774050095 Chr.7: 141724165-141724166 on Build GRCh38
rs749088512 Chr.7: 141723989-141723990 on Build GRCh38
rs757584609 Chr.7: 141724203-141724203 on Build GRCh38
rs761308738 Chr.7: 141724173-141724178 on Build GRCh38
rs903802302 Chr.7: 141723930-141723930 on Build GRCh38
rs199758946 Chr.7: 141723928-141723929 on Build GRCh38
rs750492987 Chr.7: 141724209-141724209 on Build GRCh38
rs749138708 Chr.7: 141724097-141724097 on Build GRCh38
rs545352124 Chr.7: 141723982-141723982 on Build GRCh38
rs765705974 Chr.7: 141724049-141724049 on Build GRCh38
rs770392359 Chr.7: 141724170-141724170 on Build GRCh38
rs754431807 Chr.7: 141723929-141723929 on Build GRCh38
rs774231384 Chr.7: 141724007-141724007 on Build GRCh38
rs200610853 Chr.7: 141724247-141724247 on Build GRCh38
rs369697892 Chr.7: 141723952-141723952 on Build GRCh38
rs758892614 Chr.7: 141724056-141724056 on Build GRCh38
rs372745103 Chr.7: 141724150-141724150 on Build GRCh38
rs768395840 Chr.7: 141724024-141724033 on Build GRCh38
rs972971631 Chr.7: 141724130-141724130 on Build GRCh38
rs377075519 Chr.7: 141724156-141724156 on Build GRCh38
rs908877653 Chr.7: 141724077-141724077 on Build GRCh38
rs763342257 Chr.7: 141724190-141724190 on Build GRCh38
rs749901624 Chr.7: 141723906-141723906 on Build GRCh38
rs377624307 Chr.7: 141723978-141723978 on Build GRCh38
rs766973200 Chr.7: 141723890-141723890 on Build GRCh38
rs779897429 Chr.7: 141723929-141723929 on Build GRCh38
rs185059891 Chr.7: 141724088-141724088 on Build GRCh38
rs772664320 Chr.7: 141724235-141724235 on Build GRCh38

View Full Product Details