Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142252002-142252532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS58
Entrez Gene ID
136541
Gene Name
protease, serine 58
Gene Aliases
PRSS1, TRY1, TRYX3, UNQ2540
UniGene
Hs.98609
Gene Chromosome Location
Chr.7: 142252138-142258058 on Build GRCh38
Transcripts
NM_001001317.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4153919 c.636G>A p.A212A Chr.7: 142252311-142252311 on Build GRCh38
4153918 c.641G>C p.G214A Chr.7: 142252306-142252306 on Build GRCh38
3431327 c.678A>C p.K226N Chr.7: 142252269-142252269 on Build GRCh38
7223149 c.689A>G p.Y230C Chr.7: 142252258-142252258 on Build GRCh38
5670666 c.576G>T p.K192N Chr.7: 142252472-142252472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016299430 Chr.7: 142252075-142252075 on Build GRCh38
rs1052410 Chr.7: 142252178-142252178 on Build GRCh38
rs748494546 Chr.7: 142252460-142252460 on Build GRCh38
rs559607569 Chr.7: 142252297-142252297 on Build GRCh38
rs750137185 Chr.7: 142252411-142252411 on Build GRCh38
rs774540040 Chr.7: 142252348-142252348 on Build GRCh38
rs751330531 Chr.7: 142252392-142252392 on Build GRCh38
rs756249398 Chr.7: 142252384-142252386 on Build GRCh38
rs1052408 Chr.7: 142252203-142252203 on Build GRCh38
rs767883860 Chr.7: 142252268-142252268 on Build GRCh38
rs562026650 Chr.7: 142252269-142252269 on Build GRCh38
rs780869245 Chr.7: 142252312-142252312 on Build GRCh38
rs749703175 Chr.7: 142252336-142252336 on Build GRCh38
rs773633290 Chr.7: 142252353-142252353 on Build GRCh38
rs761634635 Chr.7: 142252393-142252393 on Build GRCh38
rs767155189 Chr.7: 142252400-142252400 on Build GRCh38
rs765214367 Chr.7: 142252289-142252289 on Build GRCh38
rs1005749536 Chr.7: 142252072-142252072 on Build GRCh38
rs759706485 Chr.7: 142252496-142252496 on Build GRCh38
rs758368511 Chr.7: 142252293-142252293 on Build GRCh38
rs576669566 Chr.7: 142252184-142252184 on Build GRCh38
rs572297660 Chr.7: 142252274-142252274 on Build GRCh38
rs755391867 Chr.7: 142252429-142252429 on Build GRCh38
rs753419916 Chr.7: 142252427-142252427 on Build GRCh38
rs376777288 Chr.7: 142252252-142252252 on Build GRCh38
rs1031777642 Chr.7: 142252058-142252058 on Build GRCh38
rs4726506 Chr.7: 142252035-142252035 on Build GRCh38
rs758436988 Chr.7: 142252122-142252122 on Build GRCh38
rs772403916 Chr.7: 142252352-142252352 on Build GRCh38
rs866583795 Chr.7: 142252327-142252327 on Build GRCh38
rs762061297 Chr.7: 142252255-142252255 on Build GRCh38
rs767714641 Chr.7: 142252198-142252199 on Build GRCh38
rs768963947 Chr.7: 142252238-142252238 on Build GRCh38
rs1033660197 Chr.7: 142252045-142252045 on Build GRCh38
rs764111941 Chr.7: 142252386-142252386 on Build GRCh38
rs779368969 Chr.7: 142252422-142252422 on Build GRCh38
rs766659724 Chr.7: 142252389-142252390 on Build GRCh38
rs778294511 Chr.7: 142252475-142252475 on Build GRCh38
rs371359169 Chr.7: 142252208-142252208 on Build GRCh38
rs913165939 Chr.7: 142252234-142252234 on Build GRCh38
rs777467342 Chr.7: 142252182-142252182 on Build GRCh38
rs192538414 Chr.7: 142252092-142252092 on Build GRCh38
rs774796918 Chr.7: 142252253-142252253 on Build GRCh38
rs533423282 Chr.7: 142252370-142252370 on Build GRCh38
rs373877236 Chr.7: 142252311-142252311 on Build GRCh38
rs925618651 Chr.7: 142252194-142252194 on Build GRCh38
rs545498317 Chr.7: 142252096-142252096 on Build GRCh38
rs1052406 Chr.7: 142252290-142252290 on Build GRCh38
rs750821582 Chr.7: 142252370-142252370 on Build GRCh38
rs780600981 Chr.7: 142252325-142252325 on Build GRCh38
rs780116090 Chr.7: 142252132-142252132 on Build GRCh38
rs199744203 Chr.7: 142252488-142252488 on Build GRCh38
rs1055171184 Chr.7: 142252313-142252313 on Build GRCh38
rs747132754 Chr.7: 142252483-142252483 on Build GRCh38
rs368783544 Chr.7: 142252199-142252199 on Build GRCh38
rs753942561 Chr.7: 142252450-142252451 on Build GRCh38
rs745348715 Chr.7: 142252317-142252317 on Build GRCh38
rs370140173 Chr.7: 142252373-142252373 on Build GRCh38
rs868572253 Chr.7: 142252138-142252138 on Build GRCh38
rs755649417 Chr.7: 142252322-142252322 on Build GRCh38
rs758192404 Chr.7: 142252181-142252181 on Build GRCh38
rs753839883 Chr.7: 142252106-142252106 on Build GRCh38
rs748547499 Chr.7: 142252349-142252349 on Build GRCh38
rs749662654 Chr.7: 142252217-142252217 on Build GRCh38
rs775904047 Chr.7: 142252209-142252209 on Build GRCh38
rs772622142 Chr.7: 142252473-142252473 on Build GRCh38
rs769214940 Chr.7: 142252346-142252346 on Build GRCh38
rs553938243 Chr.7: 142252066-142252066 on Build GRCh38
rs535629821 Chr.7: 142252040-142252040 on Build GRCh38
rs755808296 Chr.7: 142252414-142252414 on Build GRCh38
rs751381841 Chr.7: 142252301-142252301 on Build GRCh38
rs369875290 Chr.7: 142252367-142252367 on Build GRCh38
rs985083656 Chr.7: 142252192-142252192 on Build GRCh38
rs998850950 Chr.7: 142252041-142252041 on Build GRCh38
rs1015831051 Chr.7: 142252382-142252382 on Build GRCh38
rs184524600 Chr.7: 142252457-142252457 on Build GRCh38
rs963038812 Chr.7: 142252242-142252242 on Build GRCh38
rs760822493 Chr.7: 142252354-142252354 on Build GRCh38
rs144200781 Chr.7: 142252493-142252493 on Build GRCh38
rs1023572331 Chr.7: 142252158-142252158 on Build GRCh38
rs765111680 Chr.7: 142252177-142252177 on Build GRCh38

View Full Product Details