Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142854795-142855299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB6
Entrez Gene ID
2051
Gene Name
EPH receptor B6
Gene Aliases
HEP
UniGene
Hs.380089
Gene Chromosome Location
Chr.7: 142854640-142871093 on Build GRCh38
Transcripts
NM_001280794.2 XM_006715881.2 XM_011515879.1 XM_011515880.1 XM_011515881.2 XM_011515882.2

SNP Detail

SNP ID Chromosome Location
rs949689923 Chr.7: 142855070-142855070 on Build GRCh38
rs908202710 Chr.7: 142855075-142855075 on Build GRCh38
rs551730976 Chr.7: 142855073-142855073 on Build GRCh38
rs1033886530 Chr.7: 142855028-142855028 on Build GRCh38
rs1034366184 Chr.7: 142855129-142855129 on Build GRCh38
rs934305821 Chr.7: 142855053-142855053 on Build GRCh38
rs569990722 Chr.7: 142855112-142855112 on Build GRCh38
rs959588034 Chr.7: 142855153-142855153 on Build GRCh38
rs543072092 Chr.7: 142854836-142854836 on Build GRCh38
rs535623063 Chr.7: 142855160-142855160 on Build GRCh38
rs899709601 Chr.7: 142855098-142855098 on Build GRCh38
rs868148834 Chr.7: 142854922-142854922 on Build GRCh38
rs943707139 Chr.7: 142855196-142855196 on Build GRCh38
rs528988947 Chr.7: 142854971-142854971 on Build GRCh38
rs916403599 Chr.7: 142854844-142854844 on Build GRCh38
rs947131251 Chr.7: 142855068-142855068 on Build GRCh38
rs912221459 Chr.7: 142855030-142855030 on Build GRCh38
rs184491739 Chr.7: 142855157-142855157 on Build GRCh38
rs904040581 Chr.7: 142855078-142855078 on Build GRCh38
rs992531542 Chr.7: 142855058-142855058 on Build GRCh38
rs914175842 Chr.7: 142854881-142854881 on Build GRCh38
rs893870686 Chr.7: 142854987-142854987 on Build GRCh38
rs982716320 Chr.7: 142855210-142855210 on Build GRCh38
rs8177100 Chr.7: 142854984-142854984 on Build GRCh38
rs561787566 Chr.7: 142854912-142854912 on Build GRCh38
rs575660202 Chr.7: 142854832-142854832 on Build GRCh38
rs930999026 Chr.7: 142854936-142854936 on Build GRCh38
rs1055462482 Chr.7: 142854928-142854928 on Build GRCh38
rs867154177 Chr.7: 142855090-142855090 on Build GRCh38
rs921193594 Chr.7: 142855238-142855238 on Build GRCh38
rs919816393 Chr.7: 142854880-142854880 on Build GRCh38
rs971356843 Chr.7: 142854887-142854887 on Build GRCh38
rs899662164 Chr.7: 142854977-142854977 on Build GRCh38
rs1027762596 Chr.7: 142855162-142855162 on Build GRCh38
rs114390654 Chr.7: 142855211-142855211 on Build GRCh38
rs906519469 Chr.7: 142855123-142855123 on Build GRCh38
rs146087273 Chr.7: 142854986-142854986 on Build GRCh38
rs964186234 Chr.7: 142854830-142854830 on Build GRCh38
rs944004938 Chr.7: 142855087-142855087 on Build GRCh38
rs908254177 Chr.7: 142855186-142855186 on Build GRCh38
rs919691014 Chr.7: 142855060-142855060 on Build GRCh38
rs931052873 Chr.7: 142855061-142855061 on Build GRCh38
rs932489602 Chr.7: 142855106-142855106 on Build GRCh38
rs982932730 Chr.7: 142855185-142855185 on Build GRCh38
rs192288549 Chr.7: 142855015-142855015 on Build GRCh38
rs959535569 Chr.7: 142855044-142855044 on Build GRCh38
rs943756365 Chr.7: 142854921-142854921 on Build GRCh38
rs988099332 Chr.7: 142854880-142854880 on Build GRCh38
rs887060272 Chr.7: 142854946-142854946 on Build GRCh38
rs1040925326 Chr.7: 142854975-142854975 on Build GRCh38
rs1003511924 Chr.7: 142855125-142855125 on Build GRCh38
rs1044290117 Chr.7: 142854951-142854951 on Build GRCh38
rs973519681 Chr.7: 142855045-142855045 on Build GRCh38
rs1053588057 Chr.7: 142855107-142855107 on Build GRCh38
rs1021360124 Chr.7: 142855007-142855007 on Build GRCh38
rs114110119 Chr.7: 142855246-142855246 on Build GRCh38
rs975077208 Chr.7: 142855050-142855050 on Build GRCh38
rs920979306 Chr.7: 142855066-142855066 on Build GRCh38
rs896489709 Chr.7: 142855099-142855099 on Build GRCh38
rs1039507094 Chr.7: 142854927-142854927 on Build GRCh38
rs1009582979 Chr.7: 142854992-142854992 on Build GRCh38
rs982330424 Chr.7: 142854902-142854902 on Build GRCh38
rs958029883 Chr.7: 142854871-142854871 on Build GRCh38
rs112612791 Chr.7: 142855092-142855092 on Build GRCh38
rs1000202562 Chr.7: 142854967-142854967 on Build GRCh38
rs1047142178 Chr.7: 142854940-142854940 on Build GRCh38
rs929891051 Chr.7: 142854901-142854901 on Build GRCh38
rs920948100 Chr.7: 142855272-142855272 on Build GRCh38
rs1026801881 Chr.7: 142855173-142855173 on Build GRCh38
rs755828058 Chr.7: 142855156-142855156 on Build GRCh38
rs943723767 Chr.7: 142855033-142855034 on Build GRCh38
rs8177101 Chr.7: 142855114-142855114 on Build GRCh38
rs926764709 Chr.7: 142854910-142854910 on Build GRCh38
rs952504692 Chr.7: 142855183-142855183 on Build GRCh38
rs918145213 Chr.7: 142855064-142855064 on Build GRCh38
rs115063624 Chr.7: 142855241-142855241 on Build GRCh38
rs531699317 Chr.7: 142855113-142855113 on Build GRCh38
rs1008545242 Chr.7: 142854962-142854962 on Build GRCh38
rs386718773 Chr.7: 142855241-142855246 on Build GRCh38
rs1019854971 Chr.7: 142854825-142854825 on Build GRCh38
rs1041370331 Chr.7: 142855095-142855095 on Build GRCh38
rs919638154 Chr.7: 142854935-142854935 on Build GRCh38
rs1032380665 Chr.7: 142854857-142854857 on Build GRCh38
rs971026308 Chr.7: 142855014-142855014 on Build GRCh38

View Full Product Details