Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143727778-143728267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCAF2
Entrez Gene ID
285966
Gene Name
TRPM8 channel associated factor 2
Gene Aliases
FAM115C, FAM139A
UniGene
Hs.743565
Gene Chromosome Location
Chr.7: 143620952-143732576 on Build GRCh38
Transcripts
NM_001130026.2 XM_006715927.3 XM_011516059.2

SNP Detail

SNP ID Chromosome Location
rs533794222 Chr.7: 143727899-143727899 on Build GRCh38
rs545669604 Chr.7: 143728238-143728238 on Build GRCh38
rs1047313287 Chr.7: 143728226-143728226 on Build GRCh38
rs1010787597 Chr.7: 143727932-143727932 on Build GRCh38
rs1045903146 Chr.7: 143727933-143727933 on Build GRCh38
rs150090419 Chr.7: 143728224-143728224 on Build GRCh38
rs558527210 Chr.7: 143727940-143727940 on Build GRCh38
rs907426104 Chr.7: 143727977-143727977 on Build GRCh38
rs566312475 Chr.7: 143727891-143727891 on Build GRCh38
rs770522254 Chr.7: 143728178-143728178 on Build GRCh38
rs576726239 Chr.7: 143728015-143728015 on Build GRCh38
rs555924180 Chr.7: 143728066-143728066 on Build GRCh38
rs922476977 Chr.7: 143728227-143728227 on Build GRCh38
rs1054450322 Chr.7: 143727852-143727852 on Build GRCh38
rs748990148 Chr.7: 143728234-143728234 on Build GRCh38
rs574321829 Chr.7: 143728081-143728081 on Build GRCh38
rs542728413 Chr.7: 143728169-143728169 on Build GRCh38
rs919570987 Chr.7: 143728214-143728214 on Build GRCh38
rs748712590 Chr.7: 143728171-143728171 on Build GRCh38
rs959011302 Chr.7: 143728121-143728121 on Build GRCh38
rs1041599774 Chr.7: 143728240-143728240 on Build GRCh38
rs569503974 Chr.7: 143727801-143727801 on Build GRCh38
rs936032032 Chr.7: 143727835-143727835 on Build GRCh38
rs1001807489 Chr.7: 143728090-143728090 on Build GRCh38
rs944256763 Chr.7: 143728239-143728239 on Build GRCh38
rs1035046386 Chr.7: 143728091-143728091 on Build GRCh38
rs544141628 Chr.7: 143728021-143728021 on Build GRCh38
rs554669224 Chr.7: 143727861-143727861 on Build GRCh38
rs34379232 Chr.7: 143727840-143727840 on Build GRCh38

View Full Product Details