Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39160808-39161316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM32
Entrez Gene ID
203102
Gene Name
ADAM metallopeptidase domain 32
Gene Aliases
-
UniGene
Hs.521545
Gene Chromosome Location
Chr.8: 39107529-39284917 on Build GRCh38
Transcripts
NM_001313994.1 NM_145004.6 XM_006716298.2 XM_006716299.1 XM_011544433.1 XM_011544434.1 XM_017013183.1 XM_017013184.1 XM_017013185.1 XM_017013186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880172 c.532C>T p.P178S Chr.8: 39160903-39160903 on Build GRCh38
6412090 c.533C>A p.P178Q Chr.8: 39160904-39160904 on Build GRCh38
1244259 c.559T>C p.Y187H Chr.8: 39160930-39160930 on Build GRCh38
2960810 c.580G>T p.V194L Chr.8: 39160951-39160951 on Build GRCh38
5409561 c.562C>T p.L188L Chr.8: 39160933-39160933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201112324 Chr.8: 39161253-39161253 on Build GRCh38
rs752727462 Chr.8: 39160903-39160903 on Build GRCh38
rs902201184 Chr.8: 39161196-39161196 on Build GRCh38
rs555698152 Chr.8: 39161258-39161258 on Build GRCh38
rs1043385912 Chr.8: 39161193-39161193 on Build GRCh38
rs878872533 Chr.8: 39161175-39161175 on Build GRCh38
rs150109740 Chr.8: 39160988-39160988 on Build GRCh38
rs765754639 Chr.8: 39160859-39160859 on Build GRCh38
rs565942062 Chr.8: 39160844-39160845 on Build GRCh38
rs555607293 Chr.8: 39161133-39161133 on Build GRCh38
rs866183919 Chr.8: 39160955-39160955 on Build GRCh38
rs763315046 Chr.8: 39161008-39161008 on Build GRCh38
rs769256281 Chr.8: 39160965-39160965 on Build GRCh38
rs953140398 Chr.8: 39160936-39160936 on Build GRCh38
rs747170493 Chr.8: 39161142-39161142 on Build GRCh38
rs921674359 Chr.8: 39160921-39160921 on Build GRCh38
rs760944244 Chr.8: 39160983-39160993 on Build GRCh38
rs778145139 Chr.8: 39161063-39161066 on Build GRCh38
rs766704268 Chr.8: 39161008-39161009 on Build GRCh38
rs4288342 Chr.8: 39161264-39161264 on Build GRCh38
rs1055008180 Chr.8: 39161146-39161146 on Build GRCh38
rs751934244 Chr.8: 39160889-39160889 on Build GRCh38
rs1055587074 Chr.8: 39161112-39161115 on Build GRCh38
rs201473947 Chr.8: 39160951-39160951 on Build GRCh38
rs183201534 Chr.8: 39161009-39161009 on Build GRCh38
rs756186164 Chr.8: 39160907-39160907 on Build GRCh38
rs74393611 Chr.8: 39161279-39161280 on Build GRCh38
rs781144059 Chr.8: 39161162-39161162 on Build GRCh38
rs1010573444 Chr.8: 39160915-39160915 on Build GRCh38
rs573126392 Chr.8: 39161158-39161158 on Build GRCh38
rs773771882 Chr.8: 39160874-39160874 on Build GRCh38
rs750190341 Chr.8: 39160860-39160860 on Build GRCh38
rs567988260 Chr.8: 39160844-39160844 on Build GRCh38
rs374255858 Chr.8: 39160875-39160875 on Build GRCh38
rs567394417 Chr.8: 39161224-39161224 on Build GRCh38
rs781425044 Chr.8: 39160894-39160894 on Build GRCh38
rs939224994 Chr.8: 39160873-39160873 on Build GRCh38
rs939174281 Chr.8: 39161127-39161127 on Build GRCh38
rs946659401 Chr.8: 39161179-39161179 on Build GRCh38
rs535406136 Chr.8: 39161251-39161251 on Build GRCh38
rs374537005 Chr.8: 39161060-39161060 on Build GRCh38
rs750936152 Chr.8: 39160866-39160866 on Build GRCh38
rs1055057859 Chr.8: 39160900-39160900 on Build GRCh38
rs755127722 Chr.8: 39160890-39160890 on Build GRCh38
rs766716123 Chr.8: 39160888-39160888 on Build GRCh38
rs954893721 Chr.8: 39160981-39160981 on Build GRCh38
rs200851034 Chr.8: 39160941-39160941 on Build GRCh38
rs770729373 Chr.8: 39161206-39161206 on Build GRCh38
rs370655802 Chr.8: 39160964-39160964 on Build GRCh38
rs766291932 Chr.8: 39161052-39161052 on Build GRCh38
rs890590326 Chr.8: 39161239-39161239 on Build GRCh38
rs927751555 Chr.8: 39161120-39161120 on Build GRCh38
rs1033659578 Chr.8: 39161252-39161252 on Build GRCh38
rs772059462 Chr.8: 39160939-39160939 on Build GRCh38
rs548123592 Chr.8: 39160841-39160841 on Build GRCh38
rs370499409 Chr.8: 39161055-39161055 on Build GRCh38
rs911526763 Chr.8: 39160969-39160969 on Build GRCh38
rs776474227 Chr.8: 39160953-39160953 on Build GRCh38
rs965033185 Chr.8: 39161038-39161038 on Build GRCh38
rs983551117 Chr.8: 39161064-39161064 on Build GRCh38

View Full Product Details