Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41497375-41497888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA7
Entrez Gene ID
51125
Gene Name
golgin A7
Gene Aliases
GCP16, GOLGA3AP1, GOLGA7A, HSPC041
UniGene
Hs.654773
Gene Chromosome Location
Chr.8: 41490562-41510980 on Build GRCh38
Transcripts
NM_001002296.1 NM_001174124.1 NM_016099.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672675 c.203G>A p.G68D Chr.8: 41497600-41497600 on Build GRCh38
2961233 c.142C>T p.R48* Chr.8: 41497539-41497539 on Build GRCh38
6672674 c.143G>A p.R48Q Chr.8: 41497540-41497540 on Build GRCh38
6579266 c.206G>T p.C69F Chr.8: 41497603-41497603 on Build GRCh38
7248234 c.180C>T p.L60L Chr.8: 41497577-41497577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751771811 Chr.8: 41497693-41497694 on Build GRCh38
rs780203269 Chr.8: 41497479-41497479 on Build GRCh38
rs1009601371 Chr.8: 41497683-41497683 on Build GRCh38
rs768390042 Chr.8: 41497595-41497595 on Build GRCh38
rs991441343 Chr.8: 41497715-41497715 on Build GRCh38
rs762518399 Chr.8: 41497523-41497523 on Build GRCh38
rs145142983 Chr.8: 41497580-41497580 on Build GRCh38
rs747952167 Chr.8: 41497649-41497649 on Build GRCh38
rs572050337 Chr.8: 41497752-41497752 on Build GRCh38
rs991098806 Chr.8: 41497404-41497404 on Build GRCh38
rs986476506 Chr.8: 41497431-41497431 on Build GRCh38
rs140679190 Chr.8: 41497692-41497692 on Build GRCh38
rs772905664 Chr.8: 41497517-41497517 on Build GRCh38
rs939732650 Chr.8: 41497475-41497475 on Build GRCh38
rs199720920 Chr.8: 41497499-41497499 on Build GRCh38
rs768596699 Chr.8: 41497495-41497495 on Build GRCh38
rs573833038 Chr.8: 41497578-41497578 on Build GRCh38
rs958415832 Chr.8: 41497701-41497701 on Build GRCh38
rs752937284 Chr.8: 41497572-41497572 on Build GRCh38
rs146085726 Chr.8: 41497540-41497540 on Build GRCh38
rs773808599 Chr.8: 41497693-41497693 on Build GRCh38
rs949535322 Chr.8: 41497446-41497446 on Build GRCh38
rs545979226 Chr.8: 41497676-41497676 on Build GRCh38
rs763329840 Chr.8: 41497462-41497463 on Build GRCh38
rs538587388 Chr.8: 41497620-41497620 on Build GRCh38
rs144603409 Chr.8: 41497828-41497828 on Build GRCh38
rs1028543183 Chr.8: 41497769-41497769 on Build GRCh38
rs767955805 Chr.8: 41497547-41497547 on Build GRCh38
rs769658033 Chr.8: 41497640-41497640 on Build GRCh38
rs759985523 Chr.8: 41497530-41497530 on Build GRCh38
rs576188594 Chr.8: 41497672-41497672 on Build GRCh38
rs558267265 Chr.8: 41497710-41497710 on Build GRCh38
rs748918663 Chr.8: 41497679-41497679 on Build GRCh38
rs987072643 Chr.8: 41497840-41497840 on Build GRCh38
rs879698825 Chr.8: 41497702-41497702 on Build GRCh38
rs779089607 Chr.8: 41497581-41497581 on Build GRCh38
rs770447674 Chr.8: 41497687-41497687 on Build GRCh38
rs746910123 Chr.8: 41497491-41497491 on Build GRCh38
rs769276847 Chr.8: 41497510-41497510 on Build GRCh38
rs566111363 Chr.8: 41497464-41497464 on Build GRCh38
rs1025240194 Chr.8: 41497737-41497737 on Build GRCh38
rs747844513 Chr.8: 41497505-41497505 on Build GRCh38
rs970878498 Chr.8: 41497738-41497740 on Build GRCh38
rs769627888 Chr.8: 41497652-41497652 on Build GRCh38
rs758331680 Chr.8: 41497426-41497426 on Build GRCh38
rs781406337 Chr.8: 41497799-41497799 on Build GRCh38
rs764413392 Chr.8: 41497546-41497549 on Build GRCh38
rs903983847 Chr.8: 41497717-41497717 on Build GRCh38
rs372071364 Chr.8: 41497622-41497622 on Build GRCh38
rs201213146 Chr.8: 41497637-41497637 on Build GRCh38
rs754105702 Chr.8: 41497579-41497579 on Build GRCh38
rs978295284 Chr.8: 41497767-41497767 on Build GRCh38
rs200463324 Chr.8: 41497529-41497529 on Build GRCh38
rs898942488 Chr.8: 41497589-41497589 on Build GRCh38
rs542115229 Chr.8: 41497607-41497607 on Build GRCh38
rs757395910 Chr.8: 41497472-41497472 on Build GRCh38
rs765741491 Chr.8: 41497460-41497462 on Build GRCh38
rs1015254281 Chr.8: 41497844-41497844 on Build GRCh38
rs17656952 Chr.8: 41497577-41497577 on Build GRCh38
rs924006306 Chr.8: 41497846-41497846 on Build GRCh38
rs775980013 Chr.8: 41497463-41497463 on Build GRCh38

View Full Product Details