Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41510292-41510804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA7
Entrez Gene ID
51125
Gene Name
golgin A7
Gene Aliases
GCP16, GOLGA3AP1, GOLGA7A, HSPC041
UniGene
Hs.654773
Gene Chromosome Location
Chr.8: 41490562-41510980 on Build GRCh38
Transcripts
NM_001002296.1 NM_001174124.1 NM_016099.2

SNP Detail

SNP ID Chromosome Location
rs917536894 Chr.8: 41510712-41510712 on Build GRCh38
rs572701217 Chr.8: 41510703-41510703 on Build GRCh38
rs977521200 Chr.8: 41510750-41510750 on Build GRCh38
rs926312453 Chr.8: 41510442-41510443 on Build GRCh38
rs936105169 Chr.8: 41510569-41510569 on Build GRCh38
rs189900604 Chr.8: 41510775-41510775 on Build GRCh38
rs899844724 Chr.8: 41510628-41510628 on Build GRCh38
rs1016785303 Chr.8: 41510677-41510677 on Build GRCh38
rs11337 Chr.8: 41510767-41510767 on Build GRCh38
rs759868873 Chr.8: 41510718-41510718 on Build GRCh38
rs776752230 Chr.8: 41510477-41510477 on Build GRCh38
rs771229040 Chr.8: 41510367-41510367 on Build GRCh38
rs980564617 Chr.8: 41510690-41510690 on Build GRCh38
rs770364962 Chr.8: 41510675-41510675 on Build GRCh38
rs768679735 Chr.8: 41510448-41510448 on Build GRCh38
rs564154653 Chr.8: 41510779-41510779 on Build GRCh38
rs1033733091 Chr.8: 41510699-41510699 on Build GRCh38
rs768329435 Chr.8: 41510514-41510514 on Build GRCh38
rs541494234 Chr.8: 41510719-41510719 on Build GRCh38
rs115198548 Chr.8: 41510771-41510771 on Build GRCh38
rs993105370 Chr.8: 41510448-41510449 on Build GRCh38
rs777091950 Chr.8: 41510657-41510657 on Build GRCh38
rs937563878 Chr.8: 41510442-41510444 on Build GRCh38
rs998498053 Chr.8: 41510645-41510645 on Build GRCh38
rs1040088782 Chr.8: 41510601-41510601 on Build GRCh38
rs904584164 Chr.8: 41510524-41510524 on Build GRCh38
rs559080258 Chr.8: 41510680-41510680 on Build GRCh38
rs545731362 Chr.8: 41510571-41510571 on Build GRCh38
rs181993229 Chr.8: 41510409-41510409 on Build GRCh38
rs7442 Chr.8: 41510363-41510363 on Build GRCh38
rs186223423 Chr.8: 41510482-41510482 on Build GRCh38
rs1039819492 Chr.8: 41510773-41510773 on Build GRCh38
rs558105177 Chr.8: 41510700-41510700 on Build GRCh38
rs769414238 Chr.8: 41510382-41510382 on Build GRCh38
rs1029604434 Chr.8: 41510652-41510652 on Build GRCh38

View Full Product Details