Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41665088-41665587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK1
Entrez Gene ID
286
Gene Name
ankyrin 1
Gene Aliases
ANK, SPH1, SPH2
UniGene
Hs.654438
Gene Chromosome Location
Chr.8: 41653225-41896762 on Build GRCh38
Transcripts
NM_000037.3 NM_001142445.1 NM_001142446.1 NM_020475.2 NM_020476.2 NM_020477.2 NM_020478.4 NM_020480.4 XM_005273476.4 XM_011544490.2 XM_011544491.2 XM_011544494.2 XM_011544495.2 XM_011544496.2 XM_011544500.2 XM_011544501.2 XM_011544502.2 XM_011544503.2 XM_011544504.2 XM_011544505.2 XM_017013319.1 XM_017013320.1 XM_017013321.1 XM_017013322.1 XM_017013323.1 XM_017013324.1 XM_017013325.1 XM_017013326.1 XM_017013327.1 XM_017013328.1 XM_017013329.1

SNP Detail

SNP ID Chromosome Location
rs913334796 Chr.8: 41665417-41665417 on Build GRCh38
rs956867126 Chr.8: 41665434-41665434 on Build GRCh38
rs975111753 Chr.8: 41665488-41665488 on Build GRCh38
rs1035615900 Chr.8: 41665433-41665433 on Build GRCh38
rs528911704 Chr.8: 41665243-41665243 on Build GRCh38
rs185298631 Chr.8: 41665562-41665562 on Build GRCh38
rs1023704360 Chr.8: 41665204-41665204 on Build GRCh38
rs914924189 Chr.8: 41665280-41665280 on Build GRCh38
rs969425689 Chr.8: 41665216-41665216 on Build GRCh38
rs901005693 Chr.8: 41665267-41665267 on Build GRCh38
rs577748398 Chr.8: 41665122-41665122 on Build GRCh38
rs763871919 Chr.8: 41665500-41665500 on Build GRCh38
rs1019444933 Chr.8: 41665232-41665232 on Build GRCh38
rs1040927831 Chr.8: 41665479-41665479 on Build GRCh38
rs989326202 Chr.8: 41665413-41665413 on Build GRCh38
rs759914594 Chr.8: 41665497-41665497 on Build GRCh38
rs776397611 Chr.8: 41665217-41665217 on Build GRCh38
rs1051376451 Chr.8: 41665326-41665326 on Build GRCh38
rs905294701 Chr.8: 41665419-41665419 on Build GRCh38
rs887056609 Chr.8: 41665113-41665113 on Build GRCh38
rs534245933 Chr.8: 41665340-41665340 on Build GRCh38
rs957181845 Chr.8: 41665274-41665274 on Build GRCh38
rs889746251 Chr.8: 41665472-41665472 on Build GRCh38
rs185242397 Chr.8: 41665252-41665252 on Build GRCh38
rs150848796 Chr.8: 41665342-41665342 on Build GRCh38
rs1022579345 Chr.8: 41665452-41665452 on Build GRCh38
rs975059458 Chr.8: 41665305-41665305 on Build GRCh38
rs749758122 Chr.8: 41665356-41665356 on Build GRCh38
rs564436110 Chr.8: 41665118-41665118 on Build GRCh38
rs551770894 Chr.8: 41665323-41665323 on Build GRCh38
rs747285023 Chr.8: 41665134-41665134 on Build GRCh38
rs117222764 Chr.8: 41665325-41665325 on Build GRCh38
rs1025868290 Chr.8: 41665524-41665524 on Build GRCh38
rs982726417 Chr.8: 41665230-41665230 on Build GRCh38
rs540391507 Chr.8: 41665139-41665139 on Build GRCh38
rs536736750 Chr.8: 41665465-41665465 on Build GRCh38
rs986541754 Chr.8: 41665511-41665511 on Build GRCh38
rs142183303 Chr.8: 41665279-41665279 on Build GRCh38
rs1025373522 Chr.8: 41665291-41665291 on Build GRCh38
rs1003591740 Chr.8: 41665426-41665426 on Build GRCh38
rs527275351 Chr.8: 41665137-41665138 on Build GRCh38
rs938790409 Chr.8: 41665162-41665162 on Build GRCh38
rs994894801 Chr.8: 41665148-41665148 on Build GRCh38
rs886904375 Chr.8: 41665328-41665328 on Build GRCh38
rs994819947 Chr.8: 41665369-41665369 on Build GRCh38
rs1023588344 Chr.8: 41665418-41665418 on Build GRCh38
rs755203264 Chr.8: 41665176-41665176 on Build GRCh38
rs948918918 Chr.8: 41665115-41665115 on Build GRCh38
rs994032560 Chr.8: 41665501-41665501 on Build GRCh38
rs1053203530 Chr.8: 41665171-41665171 on Build GRCh38
rs908383574 Chr.8: 41665557-41665557 on Build GRCh38
rs768382043 Chr.8: 41665174-41665174 on Build GRCh38
rs28411394 Chr.8: 41665292-41665292 on Build GRCh38
rs1015340339 Chr.8: 41665137-41665137 on Build GRCh38
rs938820947 Chr.8: 41665350-41665350 on Build GRCh38
rs891821572 Chr.8: 41665181-41665184 on Build GRCh38
rs508419 Chr.8: 41665473-41665473 on Build GRCh38
rs888208159 Chr.8: 41665544-41665544 on Build GRCh38
rs181204187 Chr.8: 41665448-41665448 on Build GRCh38
rs939949416 Chr.8: 41665339-41665339 on Build GRCh38
rs921056193 Chr.8: 41665315-41665315 on Build GRCh38
rs7815433 Chr.8: 41665231-41665231 on Build GRCh38

View Full Product Details