Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42178819-42179267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAT
Entrez Gene ID
5327
Gene Name
plasminogen activator, tissue type
Gene Aliases
T-PA, TPA
UniGene
Hs.491582
Gene Chromosome Location
Chr.8: 42174718-42207724 on Build GRCh38
Transcripts
NM_000930.4 NM_001319189.1 NM_033011.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3763349 c.1368T>C p.S456S Chr.8: 42179059-42179059 on Build GRCh38
6377821 c.1411T>C p.Y471H Chr.8: 42179016-42179016 on Build GRCh38
5430475 c.1364-1G>A p.? Chr.8: 42179064-42179064 on Build GRCh38
1099980 c.1379C>T p.S460L Chr.8: 42179048-42179048 on Build GRCh38
4588112 c.1507G>A p.A503T Chr.8: 42178920-42178920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564988743 Chr.8: 42179048-42179048 on Build GRCh38
rs540959015 Chr.8: 42179190-42179190 on Build GRCh38
rs762064626 Chr.8: 42179204-42179204 on Build GRCh38
rs549342605 Chr.8: 42178929-42178929 on Build GRCh38
rs547475202 Chr.8: 42178867-42178867 on Build GRCh38
rs540835879 Chr.8: 42179119-42179119 on Build GRCh38
rs774851580 Chr.8: 42178905-42178905 on Build GRCh38
rs752630879 Chr.8: 42179017-42179017 on Build GRCh38
rs763934243 Chr.8: 42179083-42179083 on Build GRCh38
rs887226344 Chr.8: 42178930-42178930 on Build GRCh38
rs8178788 Chr.8: 42179245-42179245 on Build GRCh38
rs898555520 Chr.8: 42179212-42179212 on Build GRCh38
rs770984249 Chr.8: 42179062-42179062 on Build GRCh38
rs184239040 Chr.8: 42178909-42178909 on Build GRCh38
rs200252970 Chr.8: 42178966-42178966 on Build GRCh38
rs748426960 Chr.8: 42179051-42179051 on Build GRCh38
rs757373453 Chr.8: 42178858-42178858 on Build GRCh38
rs974355536 Chr.8: 42179128-42179128 on Build GRCh38
rs780841805 Chr.8: 42178870-42178870 on Build GRCh38
rs1038866680 Chr.8: 42179211-42179211 on Build GRCh38
rs933172127 Chr.8: 42178914-42178914 on Build GRCh38
rs762002120 Chr.8: 42178926-42178926 on Build GRCh38
rs900820899 Chr.8: 42179240-42179240 on Build GRCh38
rs770378279 Chr.8: 42178884-42178884 on Build GRCh38
rs778389030 Chr.8: 42178941-42178941 on Build GRCh38
rs1136159 Chr.8: 42179059-42179059 on Build GRCh38
rs552849606 Chr.8: 42179005-42179005 on Build GRCh38
rs753553474 Chr.8: 42179026-42179026 on Build GRCh38
rs776948300 Chr.8: 42178919-42178919 on Build GRCh38
rs753599533 Chr.8: 42179084-42179084 on Build GRCh38
rs764442424 Chr.8: 42178912-42178912 on Build GRCh38
rs757853131 Chr.8: 42179113-42179113 on Build GRCh38
rs767393072 Chr.8: 42178931-42178931 on Build GRCh38
rs745318874 Chr.8: 42179036-42179045 on Build GRCh38
rs557377668 Chr.8: 42179025-42179025 on Build GRCh38
rs765255848 Chr.8: 42179124-42179127 on Build GRCh38
rs114534719 Chr.8: 42179016-42179016 on Build GRCh38
rs753938978 Chr.8: 42178856-42178856 on Build GRCh38
rs371252148 Chr.8: 42178864-42178864 on Build GRCh38
rs923213394 Chr.8: 42179163-42179163 on Build GRCh38
rs62001886 Chr.8: 42178948-42178948 on Build GRCh38
rs538933951 Chr.8: 42178938-42178938 on Build GRCh38
rs1052744690 Chr.8: 42178846-42178846 on Build GRCh38
rs778527103 Chr.8: 42179033-42179033 on Build GRCh38
rs762124946 Chr.8: 42178976-42178976 on Build GRCh38
rs374401465 Chr.8: 42178869-42178869 on Build GRCh38
rs946863418 Chr.8: 42179189-42179189 on Build GRCh38
rs892734585 Chr.8: 42179155-42179155 on Build GRCh38
rs567211487 Chr.8: 42178880-42178880 on Build GRCh38
rs781771661 Chr.8: 42178988-42178990 on Build GRCh38
rs777001661 Chr.8: 42178970-42178970 on Build GRCh38
rs61755432 Chr.8: 42178946-42178946 on Build GRCh38
rs62001885 Chr.8: 42179052-42179052 on Build GRCh38
rs760600716 Chr.8: 42179069-42179069 on Build GRCh38
rs527444834 Chr.8: 42178844-42178844 on Build GRCh38
rs777301185 Chr.8: 42178881-42178881 on Build GRCh38
rs1011060263 Chr.8: 42178868-42178868 on Build GRCh38
rs545287158 Chr.8: 42179043-42179043 on Build GRCh38
rs764319839 Chr.8: 42178850-42178850 on Build GRCh38
rs529724417 Chr.8: 42179198-42179198 on Build GRCh38
rs1021334987 Chr.8: 42179103-42179103 on Build GRCh38
rs756882853 Chr.8: 42179030-42179030 on Build GRCh38
rs760475942 Chr.8: 42179023-42179023 on Build GRCh38
rs747889671 Chr.8: 42178872-42178872 on Build GRCh38
rs769212083 Chr.8: 42179047-42179047 on Build GRCh38
rs750035977 Chr.8: 42179104-42179104 on Build GRCh38
rs755859607 Chr.8: 42178933-42178933 on Build GRCh38
rs199540133 Chr.8: 42179042-42179042 on Build GRCh38
rs764821085 Chr.8: 42179093-42179093 on Build GRCh38
rs761556900 Chr.8: 42179086-42179086 on Build GRCh38
rs751584975 Chr.8: 42178928-42178928 on Build GRCh38
rs76464116 Chr.8: 42178922-42178922 on Build GRCh38
rs776677837 Chr.8: 42179203-42179203 on Build GRCh38
rs748826704 Chr.8: 42178882-42178882 on Build GRCh38
rs765343785 Chr.8: 42178927-42178927 on Build GRCh38
rs770054356 Chr.8: 42178997-42178997 on Build GRCh38
rs116109429 Chr.8: 42179018-42179018 on Build GRCh38
rs775294699 Chr.8: 42179068-42179068 on Build GRCh38
rs368496765 Chr.8: 42178908-42178908 on Build GRCh38
rs534863394 Chr.8: 42178965-42178965 on Build GRCh38
rs773140572 Chr.8: 42179003-42179003 on Build GRCh38
rs746292560 Chr.8: 42178906-42178906 on Build GRCh38
rs549403695 Chr.8: 42179235-42179235 on Build GRCh38
rs757749145 Chr.8: 42178959-42178961 on Build GRCh38
rs759553227 Chr.8: 42179010-42179010 on Build GRCh38
rs560675440 Chr.8: 42179129-42179129 on Build GRCh38
rs201002244 Chr.8: 42178937-42178937 on Build GRCh38
rs555186256 Chr.8: 42178932-42178932 on Build GRCh38

View Full Product Details