Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42344837-42345350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLB
Entrez Gene ID
5423
Gene Name
DNA polymerase beta
Gene Aliases
-
UniGene
Hs.654484
Gene Chromosome Location
Chr.8: 42338452-42371813 on Build GRCh38
Transcripts
NM_002690.2 XM_005273535.3 XM_005273536.3 XM_005273537.3 XM_005273538.2 XM_005273539.2 XM_005273540.4 XM_006716353.2 XM_017013583.1 XM_017013584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
246700 c.149C>T p.P50L Chr.8: 42344982-42344982 on Build GRCh38
3900284 c.131C>G p.S44C Chr.8: 42344964-42344964 on Build GRCh38
95727 c.120-1G>C p.? Chr.8: 42344952-42344952 on Build GRCh38
6410641 c.186G>T p.L62F Chr.8: 42345019-42345019 on Build GRCh38
5997922 c.146A>C p.Y49S Chr.8: 42344979-42344979 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs556645499 Chr.8: 42345263-42345263 on Build GRCh38
rs147550866 Chr.8: 42344922-42344922 on Build GRCh38
rs913756670 Chr.8: 42345260-42345260 on Build GRCh38
rs1049399928 Chr.8: 42345301-42345301 on Build GRCh38
rs767197743 Chr.8: 42345070-42345071 on Build GRCh38
rs771511811 Chr.8: 42344943-42344943 on Build GRCh38
rs951305998 Chr.8: 42345275-42345275 on Build GRCh38
rs759976060 Chr.8: 42344949-42344949 on Build GRCh38
rs142575633 Chr.8: 42344906-42344906 on Build GRCh38
rs980888040 Chr.8: 42344880-42344880 on Build GRCh38
rs754679153 Chr.8: 42344911-42344911 on Build GRCh38
rs188201499 Chr.8: 42345289-42345289 on Build GRCh38
rs943690367 Chr.8: 42344866-42344866 on Build GRCh38
rs536519771 Chr.8: 42345192-42345192 on Build GRCh38
rs554257288 Chr.8: 42345044-42345044 on Build GRCh38
rs913777714 Chr.8: 42344928-42344928 on Build GRCh38
rs770478325 Chr.8: 42345237-42345237 on Build GRCh38
rs926674783 Chr.8: 42345178-42345178 on Build GRCh38
rs576592256 Chr.8: 42345288-42345288 on Build GRCh38
rs762341217 Chr.8: 42345133-42345133 on Build GRCh38
rs951994230 Chr.8: 42345151-42345151 on Build GRCh38
rs747649753 Chr.8: 42344913-42344913 on Build GRCh38
rs780891094 Chr.8: 42344912-42344912 on Build GRCh38
rs1007066869 Chr.8: 42345226-42345226 on Build GRCh38
rs965422352 Chr.8: 42345238-42345238 on Build GRCh38
rs980772991 Chr.8: 42345173-42345173 on Build GRCh38
rs930714716 Chr.8: 42344879-42344879 on Build GRCh38
rs570223385 Chr.8: 42344871-42344871 on Build GRCh38
rs945340443 Chr.8: 42344947-42344947 on Build GRCh38
rs79251622 Chr.8: 42344994-42344994 on Build GRCh38
rs755659583 Chr.8: 42345070-42345070 on Build GRCh38
rs767161530 Chr.8: 42345294-42345294 on Build GRCh38
rs563686679 Chr.8: 42345128-42345128 on Build GRCh38
rs993789800 Chr.8: 42345069-42345069 on Build GRCh38
rs939144141 Chr.8: 42344915-42344915 on Build GRCh38
rs747992233 Chr.8: 42345034-42345034 on Build GRCh38
rs892757349 Chr.8: 42345170-42345170 on Build GRCh38
rs948158277 Chr.8: 42344964-42344964 on Build GRCh38
rs890858054 Chr.8: 42345312-42345312 on Build GRCh38
rs143763357 Chr.8: 42344962-42344962 on Build GRCh38
rs185498497 Chr.8: 42345273-42345273 on Build GRCh38
rs1027651750 Chr.8: 42345117-42345117 on Build GRCh38
rs747868852 Chr.8: 42345057-42345057 on Build GRCh38
rs141971948 Chr.8: 42345235-42345235 on Build GRCh38
rs773270626 Chr.8: 42345056-42345056 on Build GRCh38
rs117229828 Chr.8: 42345043-42345043 on Build GRCh38
rs774914651 Chr.8: 42344946-42344946 on Build GRCh38
rs996544429 Chr.8: 42344980-42344980 on Build GRCh38
rs367961767 Chr.8: 42345052-42345052 on Build GRCh38
rs558241585 Chr.8: 42344869-42344869 on Build GRCh38
rs749821103 Chr.8: 42344936-42344936 on Build GRCh38
rs1011104416 Chr.8: 42345172-42345172 on Build GRCh38
rs774360153 Chr.8: 42345254-42345254 on Build GRCh38
rs766286109 Chr.8: 42345041-42345041 on Build GRCh38
rs960833191 Chr.8: 42345204-42345204 on Build GRCh38
rs772560551 Chr.8: 42344933-42344933 on Build GRCh38
rs764264750 Chr.8: 42344999-42344999 on Build GRCh38
rs775309622 Chr.8: 42345113-42345113 on Build GRCh38
rs111586415 Chr.8: 42344987-42344987 on Build GRCh38
rs962590244 Chr.8: 42345064-42345064 on Build GRCh38
rs920617858 Chr.8: 42344863-42344863 on Build GRCh38
rs138219401 Chr.8: 42344951-42344951 on Build GRCh38
rs769610290 Chr.8: 42345053-42345053 on Build GRCh38
rs568092686 Chr.8: 42345072-42345072 on Build GRCh38
rs758344693 Chr.8: 42345040-42345040 on Build GRCh38

View Full Product Details