Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42437470-42437978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC20A2
Entrez Gene ID
6575
Gene Name
solute carrier family 20 member 2
Gene Aliases
GLVR-2, GLVR2, IBGC1, IBGC3, MLVAR, PIT-2, PIT2, RAM1, Ram-1
UniGene
Hs.653173
Gene Chromosome Location
Chr.8: 42416462-42542213 on Build GRCh38
Transcripts
NM_001257180.1 NM_001257181.1 NM_006749.4 XM_005273613.3 XM_005273615.3 XM_006716390.3 XM_006716391.3 XM_017013748.1 XM_017013749.1 XM_017013750.1 XM_017013751.1 XM_017013752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6348871 c.947C>A p.S316Y Chr.8: 42437565-42437565 on Build GRCh38
5409631 c.948C>T p.S316S Chr.8: 42437564-42437564 on Build GRCh38
3834752 c.999C>T p.F333F Chr.8: 42437513-42437513 on Build GRCh38
323365 c.994G>T p.G332C Chr.8: 42437518-42437518 on Build GRCh38
1100038 c.971C>T p.S324L Chr.8: 42437541-42437541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938212015 Chr.8: 42437713-42437713 on Build GRCh38
rs1054850410 Chr.8: 42437672-42437672 on Build GRCh38
rs977234305 Chr.8: 42437863-42437863 on Build GRCh38
rs142610044 Chr.8: 42437894-42437894 on Build GRCh38
rs972531844 Chr.8: 42437558-42437558 on Build GRCh38
rs569791992 Chr.8: 42437518-42437518 on Build GRCh38
rs886041752 Chr.8: 42437541-42437541 on Build GRCh38
rs773231316 Chr.8: 42437612-42437613 on Build GRCh38
rs530131172 Chr.8: 42437907-42437907 on Build GRCh38
rs1025411420 Chr.8: 42437924-42437924 on Build GRCh38
rs529109194 Chr.8: 42437674-42437674 on Build GRCh38
rs753165725 Chr.8: 42437858-42437858 on Build GRCh38
rs879148358 Chr.8: 42437581-42437581 on Build GRCh38
rs1027599667 Chr.8: 42437537-42437537 on Build GRCh38
rs952531070 Chr.8: 42437933-42437933 on Build GRCh38
rs78608637 Chr.8: 42437625-42437625 on Build GRCh38
rs879009551 Chr.8: 42437598-42437598 on Build GRCh38
rs753556815 Chr.8: 42437627-42437627 on Build GRCh38
rs1036049635 Chr.8: 42437715-42437715 on Build GRCh38
rs777755692 Chr.8: 42437624-42437624 on Build GRCh38
rs756119373 Chr.8: 42437623-42437623 on Build GRCh38
rs116359869 Chr.8: 42437501-42437501 on Build GRCh38
rs762332674 Chr.8: 42437496-42437496 on Build GRCh38
rs755987724 Chr.8: 42437513-42437513 on Build GRCh38
rs891262176 Chr.8: 42437712-42437712 on Build GRCh38
rs200863955 Chr.8: 42437596-42437596 on Build GRCh38
rs188821846 Chr.8: 42437676-42437676 on Build GRCh38
rs750447282 Chr.8: 42437502-42437502 on Build GRCh38
rs754056948 Chr.8: 42437613-42437618 on Build GRCh38
rs565986549 Chr.8: 42437844-42437844 on Build GRCh38
rs141611151 Chr.8: 42437492-42437492 on Build GRCh38
rs984986535 Chr.8: 42437940-42437940 on Build GRCh38
rs534577752 Chr.8: 42437883-42437883 on Build GRCh38
rs182674447 Chr.8: 42437787-42437787 on Build GRCh38
rs902570641 Chr.8: 42437804-42437804 on Build GRCh38
rs776949497 Chr.8: 42437534-42437534 on Build GRCh38
rs763972652 Chr.8: 42437613-42437613 on Build GRCh38
rs766521602 Chr.8: 42437613-42437614 on Build GRCh38
rs769073655 Chr.8: 42437532-42437532 on Build GRCh38
rs111553899 Chr.8: 42437504-42437504 on Build GRCh38
rs1031784270 Chr.8: 42437893-42437893 on Build GRCh38
rs144236532 Chr.8: 42437589-42437589 on Build GRCh38
rs372758064 Chr.8: 42437611-42437611 on Build GRCh38
rs866596728 Chr.8: 42437508-42437508 on Build GRCh38
rs559628836 Chr.8: 42437857-42437857 on Build GRCh38
rs554622605 Chr.8: 42437622-42437622 on Build GRCh38
rs775986808 Chr.8: 42437528-42437528 on Build GRCh38
rs965011450 Chr.8: 42437556-42437556 on Build GRCh38
rs918318786 Chr.8: 42437587-42437587 on Build GRCh38
rs563362126 Chr.8: 42437586-42437586 on Build GRCh38
rs762919038 Chr.8: 42437610-42437611 on Build GRCh38
rs762948291 Chr.8: 42437597-42437597 on Build GRCh38
rs889607103 Chr.8: 42437864-42437864 on Build GRCh38
rs563334782 Chr.8: 42437679-42437679 on Build GRCh38
rs369778536 Chr.8: 42437512-42437512 on Build GRCh38
rs994541616 Chr.8: 42437761-42437761 on Build GRCh38
rs1000705331 Chr.8: 42437808-42437808 on Build GRCh38
rs548201896 Chr.8: 42437495-42437495 on Build GRCh38
rs767994198 Chr.8: 42437829-42437829 on Build GRCh38
rs949349690 Chr.8: 42437771-42437771 on Build GRCh38
rs956013625 Chr.8: 42437632-42437632 on Build GRCh38
rs574546551 Chr.8: 42437642-42437642 on Build GRCh38
rs781309493 Chr.8: 42437509-42437509 on Build GRCh38
rs149134052 Chr.8: 42437525-42437525 on Build GRCh38
rs372443583 Chr.8: 42437531-42437531 on Build GRCh38
rs921223446 Chr.8: 42437917-42437917 on Build GRCh38
rs1027726535 Chr.8: 42437785-42437785 on Build GRCh38
rs762143787 Chr.8: 42437569-42437569 on Build GRCh38
rs115961682 Chr.8: 42437527-42437527 on Build GRCh38
rs909415661 Chr.8: 42437947-42437947 on Build GRCh38
rs560949693 Chr.8: 42437896-42437896 on Build GRCh38
rs752617509 Chr.8: 42437510-42437510 on Build GRCh38
rs994210543 Chr.8: 42437922-42437922 on Build GRCh38
rs990203771 Chr.8: 42437902-42437902 on Build GRCh38
rs749040792 Chr.8: 42437522-42437522 on Build GRCh38
rs147775311 Chr.8: 42437882-42437882 on Build GRCh38
rs917964596 Chr.8: 42437755-42437755 on Build GRCh38
rs192268724 Chr.8: 42437747-42437747 on Build GRCh38
rs371592937 Chr.8: 42437787-42437788 on Build GRCh38
rs770462505 Chr.8: 42437523-42437523 on Build GRCh38
rs909469408 Chr.8: 42437687-42437687 on Build GRCh38
rs774129042 Chr.8: 42437546-42437546 on Build GRCh38
rs986986224 Chr.8: 42437934-42437934 on Build GRCh38
rs762327751 Chr.8: 42437916-42437916 on Build GRCh38

View Full Product Details