Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 7848647-7849146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPAG11A
Entrez Gene ID
653423
Gene Name
sperm associated antigen 11A
Gene Aliases
EDDM2A, HE2
UniGene
Hs.559506
Gene Chromosome Location
Chr.8: 7847880-7863797 on Build GRCh38
Transcripts
NM_001081552.2 XM_005272391.4 XM_005272393.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3367405 c.178C>T p.R60W Chr.8: 7848807-7848807 on Build GRCh38
3367406 c.178C>T p.R60W Chr.8: 7848807-7848807 on Build GRCh38
7391528 c.148G>A p.G50R Chr.8: 7848777-7848777 on Build GRCh38
3951879 c.141C>A p.G47G Chr.8: 7848770-7848770 on Build GRCh38
3951878 c.141C>A p.G47G Chr.8: 7848770-7848770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767505896 Chr.8: 7848839-7848839 on Build GRCh38
rs879137007 Chr.8: 7849082-7849082 on Build GRCh38
rs761648248 Chr.8: 7848824-7848824 on Build GRCh38
rs979886597 Chr.8: 7848971-7848971 on Build GRCh38
rs764132932 Chr.8: 7848777-7848777 on Build GRCh38
rs553117862 Chr.8: 7848910-7848910 on Build GRCh38
rs990458123 Chr.8: 7848848-7848848 on Build GRCh38
rs955962933 Chr.8: 7848826-7848826 on Build GRCh38
rs755972411 Chr.8: 7848890-7848890 on Build GRCh38
rs572943943 Chr.8: 7848925-7848925 on Build GRCh38
rs928409341 Chr.8: 7849007-7849007 on Build GRCh38
rs78408317 Chr.8: 7848665-7848665 on Build GRCh38
rs375429255 Chr.8: 7848755-7848755 on Build GRCh38
rs202056068 Chr.8: 7848807-7848807 on Build GRCh38
rs763201280 Chr.8: 7848776-7848776 on Build GRCh38
rs746254868 Chr.8: 7848723-7848723 on Build GRCh38
rs750139031 Chr.8: 7848862-7848862 on Build GRCh38
rs970258937 Chr.8: 7848916-7848916 on Build GRCh38
rs774543778 Chr.8: 7848792-7848792 on Build GRCh38
rs959781153 Chr.8: 7849026-7849026 on Build GRCh38
rs769952590 Chr.8: 7848738-7848738 on Build GRCh38
rs1021469095 Chr.8: 7848859-7848859 on Build GRCh38
rs192249995 Chr.8: 7849104-7849104 on Build GRCh38
rs536383050 Chr.8: 7848833-7848833 on Build GRCh38

View Full Product Details