Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 8328040-8328531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGK223
Entrez Gene ID
157285
Gene Name
homolog of rat pragma of Rnd2
Gene Aliases
PRAGMIN
UniGene
Hs.583863
Gene Chromosome Location
Chr.8: 8317731-8386444 on Build GRCh38
Transcripts
NM_001080826.2 XM_005272369.4 XM_005272370.4

SNP Detail

SNP ID Chromosome Location
rs992409119 Chr.8: 8328178-8328178 on Build GRCh38
rs150737128 Chr.8: 8328248-8328248 on Build GRCh38
rs756519764 Chr.8: 8328207-8328207 on Build GRCh38
rs376695924 Chr.8: 8328188-8328188 on Build GRCh38
rs757966361 Chr.8: 8328443-8328443 on Build GRCh38
rs757841886 Chr.8: 8328497-8328497 on Build GRCh38
rs370291180 Chr.8: 8328161-8328161 on Build GRCh38
rs754620559 Chr.8: 8328437-8328437 on Build GRCh38
rs189604957 Chr.8: 8328086-8328086 on Build GRCh38
rs758462451 Chr.8: 8328268-8328268 on Build GRCh38
rs529056598 Chr.8: 8328431-8328431 on Build GRCh38
rs1038446642 Chr.8: 8328062-8328062 on Build GRCh38
rs745527714 Chr.8: 8328234-8328234 on Build GRCh38
rs201115647 Chr.8: 8328398-8328398 on Build GRCh38
rs376611892 Chr.8: 8328488-8328488 on Build GRCh38
rs755872843 Chr.8: 8328426-8328426 on Build GRCh38
rs757695897 Chr.8: 8328082-8328082 on Build GRCh38
rs373723255 Chr.8: 8328257-8328257 on Build GRCh38
rs1033941671 Chr.8: 8328176-8328176 on Build GRCh38
rs781629031 Chr.8: 8328149-8328149 on Build GRCh38
rs756545845 Chr.8: 8328499-8328499 on Build GRCh38
rs778580985 Chr.8: 8328439-8328439 on Build GRCh38
rs748898707 Chr.8: 8328367-8328367 on Build GRCh38
rs777284626 Chr.8: 8328498-8328498 on Build GRCh38
rs771709111 Chr.8: 8328171-8328171 on Build GRCh38
rs780128658 Chr.8: 8328090-8328090 on Build GRCh38
rs759862258 Chr.8: 8328323-8328323 on Build GRCh38
rs749185233 Chr.8: 8328094-8328094 on Build GRCh38
rs748179534 Chr.8: 8328149-8328151 on Build GRCh38
rs201258121 Chr.8: 8328315-8328315 on Build GRCh38
rs753483464 Chr.8: 8328436-8328436 on Build GRCh38
rs751544456 Chr.8: 8328276-8328276 on Build GRCh38
rs56207906 Chr.8: 8328341-8328341 on Build GRCh38
rs769135662 Chr.8: 8328237-8328237 on Build GRCh38
rs777459493 Chr.8: 8328481-8328488 on Build GRCh38
rs1043881793 Chr.8: 8328280-8328280 on Build GRCh38
rs200130163 Chr.8: 8328066-8328066 on Build GRCh38
rs892490300 Chr.8: 8328174-8328174 on Build GRCh38
rs760554883 Chr.8: 8328472-8328472 on Build GRCh38
rs746783519 Chr.8: 8328218-8328218 on Build GRCh38
rs985891311 Chr.8: 8328221-8328221 on Build GRCh38
rs758077308 Chr.8: 8328344-8328344 on Build GRCh38
rs779753721 Chr.8: 8328359-8328359 on Build GRCh38
rs750394600 Chr.8: 8328424-8328424 on Build GRCh38
rs781265350 Chr.8: 8328084-8328084 on Build GRCh38
rs369769327 Chr.8: 8328177-8328177 on Build GRCh38
rs55844905 Chr.8: 8328298-8328298 on Build GRCh38
rs774323145 Chr.8: 8328100-8328100 on Build GRCh38
rs930459935 Chr.8: 8328068-8328068 on Build GRCh38
rs773767338 Chr.8: 8328249-8328249 on Build GRCh38
rs564448328 Chr.8: 8328077-8328077 on Build GRCh38
rs199990185 Chr.8: 8328455-8328455 on Build GRCh38
rs746104751 Chr.8: 8328085-8328085 on Build GRCh38
rs200340792 Chr.8: 8328262-8328262 on Build GRCh38
rs753548402 Chr.8: 8328483-8328483 on Build GRCh38
rs764227798 Chr.8: 8328405-8328405 on Build GRCh38
rs750770300 Chr.8: 8328203-8328203 on Build GRCh38
rs767711445 Chr.8: 8328464-8328464 on Build GRCh38
rs779066377 Chr.8: 8328214-8328214 on Build GRCh38
rs1009762697 Chr.8: 8328128-8328128 on Build GRCh38
rs764681581 Chr.8: 8328138-8328138 on Build GRCh38
rs555914877 Chr.8: 8328098-8328098 on Build GRCh38
rs749348322 Chr.8: 8328166-8328166 on Build GRCh38
rs770055358 Chr.8: 8328399-8328399 on Build GRCh38
rs765480115 Chr.8: 8328328-8328328 on Build GRCh38
rs750148724 Chr.8: 8328067-8328067 on Build GRCh38
rs757290219 Chr.8: 8328347-8328347 on Build GRCh38
rs751657674 Chr.8: 8328416-8328416 on Build GRCh38
rs943217810 Chr.8: 8328159-8328159 on Build GRCh38
rs752413297 Chr.8: 8328268-8328268 on Build GRCh38
rs564201535 Chr.8: 8328349-8328349 on Build GRCh38
rs780296797 Chr.8: 8328165-8328165 on Build GRCh38
rs910417479 Chr.8: 8328400-8328400 on Build GRCh38
rs781608984 Chr.8: 8328444-8328444 on Build GRCh38
rs768156378 Chr.8: 8328372-8328372 on Build GRCh38
rs778460085 Chr.8: 8328078-8328078 on Build GRCh38
rs764842853 Chr.8: 8328187-8328187 on Build GRCh38
rs557554757 Chr.8: 8328261-8328261 on Build GRCh38
rs201657603 Chr.8: 8328199-8328199 on Build GRCh38
rs765195604 Chr.8: 8328263-8328263 on Build GRCh38
rs768976970 Chr.8: 8328458-8328458 on Build GRCh38
rs751845225 Chr.8: 8328080-8328080 on Build GRCh38
rs747639107 Chr.8: 8328441-8328441 on Build GRCh38
rs373620049 Chr.8: 8328334-8328334 on Build GRCh38
rs1027034154 Chr.8: 8328386-8328386 on Build GRCh38
rs773145180 Chr.8: 8328470-8328470 on Build GRCh38
rs759109904 Chr.8: 8328183-8328183 on Build GRCh38
rs776481865 Chr.8: 8328181-8328181 on Build GRCh38
rs904043290 Chr.8: 8328156-8328156 on Build GRCh38
rs201882445 Chr.8: 8328373-8328373 on Build GRCh38
rs752347316 Chr.8: 8328492-8328492 on Build GRCh38
rs768390231 Chr.8: 8328060-8328060 on Build GRCh38
rs867447479 Chr.8: 8328198-8328198 on Build GRCh38
rs762410274 Chr.8: 8328245-8328245 on Build GRCh38
rs769209986 Chr.8: 8328505-8328505 on Build GRCh38
rs777759386 Chr.8: 8328275-8328275 on Build GRCh38
rs768119796 Chr.8: 8328313-8328313 on Build GRCh38
rs372709302 Chr.8: 8328476-8328476 on Build GRCh38
rs745477097 Chr.8: 8328299-8328299 on Build GRCh38
rs752172969 Chr.8: 8328144-8328144 on Build GRCh38
rs747477543 Chr.8: 8328375-8328375 on Build GRCh38
rs919998593 Chr.8: 8328213-8328213 on Build GRCh38
rs773193946 Chr.8: 8328322-8328322 on Build GRCh38
rs1009641331 Chr.8: 8328175-8328175 on Build GRCh38
rs757059242 Chr.8: 8328296-8328296 on Build GRCh38
rs369967222 Chr.8: 8328423-8328423 on Build GRCh38
rs200699182 Chr.8: 8328329-8328329 on Build GRCh38
rs199973377 Chr.8: 8328210-8328210 on Build GRCh38
rs199707534 Chr.8: 8328309-8328309 on Build GRCh38
rs761979952 Chr.8: 8328463-8328463 on Build GRCh38
rs374993110 Chr.8: 8328072-8328072 on Build GRCh38
rs748819754 Chr.8: 8328312-8328312 on Build GRCh38
rs56215812 Chr.8: 8328230-8328230 on Build GRCh38
rs747991043 Chr.8: 8328216-8328216 on Build GRCh38
rs750561679 Chr.8: 8328153-8328153 on Build GRCh38
rs746372270 Chr.8: 8328447-8328447 on Build GRCh38
rs200047944 Chr.8: 8328226-8328226 on Build GRCh38
rs1033843662 Chr.8: 8328365-8328365 on Build GRCh38
rs371954058 Chr.8: 8328129-8328129 on Build GRCh38
rs771973266 Chr.8: 8328109-8328109 on Build GRCh38
rs373954477 Chr.8: 8328380-8328380 on Build GRCh38
rs374641477 Chr.8: 8328314-8328314 on Build GRCh38
rs762032938 Chr.8: 8328421-8328421 on Build GRCh38
rs896679874 Chr.8: 8328477-8328477 on Build GRCh38
rs375255385 Chr.8: 8328146-8328146 on Build GRCh38
rs772676848 Chr.8: 8328111-8328111 on Build GRCh38
rs981017632 Chr.8: 8328064-8328064 on Build GRCh38
rs752794570 Chr.8: 8328264-8328264 on Build GRCh38
rs13272376 Chr.8: 8328075-8328075 on Build GRCh38
rs55764095 Chr.8: 8328217-8328217 on Build GRCh38
rs116369577 Chr.8: 8328397-8328397 on Build GRCh38
rs548800292 Chr.8: 8328457-8328457 on Build GRCh38
rs549591581 Chr.8: 8328097-8328097 on Build GRCh38
rs548677577 Chr.8: 8328196-8328196 on Build GRCh38
rs761501061 Chr.8: 8328101-8328101 on Build GRCh38
rs929054832 Chr.8: 8328105-8328105 on Build GRCh38
rs976425806 Chr.8: 8328403-8328403 on Build GRCh38
rs775095768 Chr.8: 8328241-8328241 on Build GRCh38
rs755920024 Chr.8: 8328351-8328351 on Build GRCh38
rs754986834 Chr.8: 8328211-8328211 on Build GRCh38
rs751491457 Chr.8: 8328343-8328343 on Build GRCh38
rs1016046256 Chr.8: 8328179-8328179 on Build GRCh38
rs202114232 Chr.8: 8328402-8328402 on Build GRCh38
rs765941718 Chr.8: 8328117-8328117 on Build GRCh38
rs773085902 Chr.8: 8328172-8328172 on Build GRCh38
rs760392743 Chr.8: 8328112-8328112 on Build GRCh38
rs766160293 Chr.8: 8328479-8328479 on Build GRCh38
rs367674951 Chr.8: 8328368-8328368 on Build GRCh38
rs765994600 Chr.8: 8328073-8328073 on Build GRCh38
rs754820192 Chr.8: 8328485-8328485 on Build GRCh38
rs766736132 Chr.8: 8328259-8328259 on Build GRCh38
rs749602747 Chr.8: 8328503-8328503 on Build GRCh38
rs778712638 Chr.8: 8328170-8328170 on Build GRCh38
rs377667569 Chr.8: 8328258-8328258 on Build GRCh38
rs757494861 Chr.8: 8328145-8328145 on Build GRCh38
rs771400118 Chr.8: 8328390-8328390 on Build GRCh38
rs769456312 Chr.8: 8328307-8328307 on Build GRCh38
rs763104094 Chr.8: 8328404-8328404 on Build GRCh38

View Full Product Details