Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 10674209-10674714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf74
Entrez Gene ID
203076
Gene Name
chromosome 8 open reading frame 74
Gene Aliases
-
UniGene
Hs.371776
Gene Chromosome Location
Chr.8: 10672557-10700616 on Build GRCh38
Transcripts
NM_001040032.1 XM_011543816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5358794 c.113A>G p.D38G Chr.8: 10674710-10674710 on Build GRCh38
7220889 c.55C>G p.Q19E Chr.8: 10674652-10674652 on Build GRCh38
3715928 c.68G>T p.R23L Chr.8: 10674665-10674665 on Build GRCh38
7294374 c.73C>T p.R25W Chr.8: 10674670-10674670 on Build GRCh38
3270808 c.103G>A p.E35K Chr.8: 10674700-10674700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752047048 Chr.8: 10674496-10674497 on Build GRCh38
rs750146299 Chr.8: 10674655-10674655 on Build GRCh38
rs113006569 Chr.8: 10674477-10674477 on Build GRCh38
rs373039547 Chr.8: 10674244-10674244 on Build GRCh38
rs534946694 Chr.8: 10674671-10674671 on Build GRCh38
rs902786442 Chr.8: 10674618-10674618 on Build GRCh38
rs111841369 Chr.8: 10674444-10674444 on Build GRCh38
rs893520673 Chr.8: 10674643-10674643 on Build GRCh38
rs556558731 Chr.8: 10674532-10674532 on Build GRCh38
rs112897246 Chr.8: 10674540-10674540 on Build GRCh38
rs112336844 Chr.8: 10674271-10674271 on Build GRCh38
rs112231451 Chr.8: 10674452-10674452 on Build GRCh38
rs111865913 Chr.8: 10674410-10674410 on Build GRCh38
rs767409792 Chr.8: 10674690-10674690 on Build GRCh38
rs142359239 Chr.8: 10674388-10674388 on Build GRCh38
rs945394432 Chr.8: 10674538-10674538 on Build GRCh38
rs149071424 Chr.8: 10674493-10674493 on Build GRCh38
rs920817264 Chr.8: 10674594-10674594 on Build GRCh38
rs761639967 Chr.8: 10674680-10674680 on Build GRCh38
rs776844704 Chr.8: 10674627-10674627 on Build GRCh38
rs561041578 Chr.8: 10674669-10674669 on Build GRCh38
rs112614482 Chr.8: 10674284-10674284 on Build GRCh38
rs187372274 Chr.8: 10674435-10674435 on Build GRCh38
rs183538523 Chr.8: 10674511-10674511 on Build GRCh38
rs61729951 Chr.8: 10674678-10674678 on Build GRCh38
rs180940226 Chr.8: 10674347-10674347 on Build GRCh38
rs761604672 Chr.8: 10674651-10674651 on Build GRCh38
rs915252373 Chr.8: 10674536-10674536 on Build GRCh38
rs565879800 Chr.8: 10674249-10674249 on Build GRCh38
rs867577184 Chr.8: 10674390-10674390 on Build GRCh38
rs778382431 Chr.8: 10674664-10674664 on Build GRCh38
rs571297718 Chr.8: 10674504-10674504 on Build GRCh38
rs931393734 Chr.8: 10674260-10674260 on Build GRCh38
rs747337266 Chr.8: 10674665-10674665 on Build GRCh38
rs113106913 Chr.8: 10674578-10674578 on Build GRCh38
rs904398379 Chr.8: 10674663-10674663 on Build GRCh38
rs551552414 Chr.8: 10674460-10674460 on Build GRCh38
rs867322765 Chr.8: 10674339-10674339 on Build GRCh38
rs189926152 Chr.8: 10674652-10674652 on Build GRCh38
rs186464846 Chr.8: 10674535-10674535 on Build GRCh38
rs750348441 Chr.8: 10674577-10674577 on Build GRCh38
rs542735152 Chr.8: 10674635-10674635 on Build GRCh38
rs755713003 Chr.8: 10674656-10674656 on Build GRCh38
rs772671621 Chr.8: 10674607-10674607 on Build GRCh38
rs182048902 Chr.8: 10674428-10674428 on Build GRCh38
rs778329401 Chr.8: 10674610-10674610 on Build GRCh38
rs749330586 Chr.8: 10674676-10674676 on Build GRCh38
rs752543687 Chr.8: 10674623-10674623 on Build GRCh38
rs528230186 Chr.8: 10674433-10674433 on Build GRCh38
rs376097708 Chr.8: 10674606-10674606 on Build GRCh38
rs376100447 Chr.8: 10674685-10674685 on Build GRCh38
rs775516675 Chr.8: 10674644-10674644 on Build GRCh38
rs758181198 Chr.8: 10674632-10674633 on Build GRCh38
rs572694662 Chr.8: 10674670-10674670 on Build GRCh38
rs766024114 Chr.8: 10674658-10674658 on Build GRCh38
rs867962248 Chr.8: 10674533-10674533 on Build GRCh38
rs762926858 Chr.8: 10674645-10674645 on Build GRCh38
rs746065734 Chr.8: 10674628-10674628 on Build GRCh38
rs747349011 Chr.8: 10674619-10674619 on Build GRCh38
rs143126207 Chr.8: 10674496-10674496 on Build GRCh38
rs774167424 Chr.8: 10674649-10674649 on Build GRCh38
rs771087866 Chr.8: 10674624-10674624 on Build GRCh38
rs536725631 Chr.8: 10674531-10674531 on Build GRCh38
rs970378813 Chr.8: 10674653-10674653 on Build GRCh38
rs890554641 Chr.8: 10674617-10674617 on Build GRCh38
rs769930343 Chr.8: 10674631-10674631 on Build GRCh38
rs191070829 Chr.8: 10674427-10674427 on Build GRCh38
rs113424148 Chr.8: 10674659-10674659 on Build GRCh38
rs755589734 Chr.8: 10674604-10674604 on Build GRCh38
rs185124644 Chr.8: 10674389-10674389 on Build GRCh38
rs763973384 Chr.8: 10674647-10674647 on Build GRCh38
rs112161259 Chr.8: 10674587-10674587 on Build GRCh38
rs112969813 Chr.8: 10674281-10674281 on Build GRCh38

View Full Product Details