Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 53226845-53227355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPRK1
Entrez Gene ID
4986
Gene Name
opioid receptor kappa 1
Gene Aliases
K-OR-1, KOR, KOR-1, OPRK
UniGene
Hs.106795
Gene Chromosome Location
Chr.8: 53225716-53251697 on Build GRCh38
Transcripts
NM_000912.4 NM_001282904.1 NM_001318497.1

SNP Detail

SNP ID Chromosome Location
rs201380666 Chr.8: 53227278-53227278 on Build GRCh38
rs935907101 Chr.8: 53227244-53227244 on Build GRCh38
rs796954466 Chr.8: 53227003-53227003 on Build GRCh38
rs202200365 Chr.8: 53227075-53227075 on Build GRCh38
rs922320387 Chr.8: 53227154-53227154 on Build GRCh38
rs546062417 Chr.8: 53226938-53226938 on Build GRCh38
rs201977326 Chr.8: 53227306-53227306 on Build GRCh38
rs762998089 Chr.8: 53227176-53227176 on Build GRCh38
rs201466973 Chr.8: 53227258-53227258 on Build GRCh38
rs199715351 Chr.8: 53226957-53226957 on Build GRCh38
rs200610564 Chr.8: 53227214-53227214 on Build GRCh38
rs201878596 Chr.8: 53227023-53227023 on Build GRCh38
rs10094852 Chr.8: 53227114-53227114 on Build GRCh38
rs560207276 Chr.8: 53226869-53226869 on Build GRCh38
rs200050369 Chr.8: 53227227-53227227 on Build GRCh38
rs939744328 Chr.8: 53226929-53226929 on Build GRCh38
rs762990502 Chr.8: 53227115-53227115 on Build GRCh38
rs140284429 Chr.8: 53227139-53227139 on Build GRCh38
rs199928307 Chr.8: 53227260-53227260 on Build GRCh38
rs60954107 Chr.8: 53227276-53227278 on Build GRCh38
rs202050066 Chr.8: 53227220-53227220 on Build GRCh38
rs199971408 Chr.8: 53227014-53227014 on Build GRCh38
rs201356662 Chr.8: 53227130-53227130 on Build GRCh38
rs991497183 Chr.8: 53227020-53227020 on Build GRCh38
rs201836328 Chr.8: 53227148-53227148 on Build GRCh38
rs765186074 Chr.8: 53227097-53227097 on Build GRCh38
rs201383990 Chr.8: 53226984-53226984 on Build GRCh38
rs554431391 Chr.8: 53227172-53227172 on Build GRCh38
rs200729960 Chr.8: 53226976-53226976 on Build GRCh38
rs200762323 Chr.8: 53227022-53227022 on Build GRCh38
rs961670400 Chr.8: 53226993-53227005 on Build GRCh38
rs200945507 Chr.8: 53227298-53227298 on Build GRCh38
rs199536352 Chr.8: 53226927-53226927 on Build GRCh38
rs200255266 Chr.8: 53226880-53226880 on Build GRCh38
rs199558259 Chr.8: 53227131-53227131 on Build GRCh38
rs201958345 Chr.8: 53226977-53226977 on Build GRCh38
rs200269638 Chr.8: 53227123-53227123 on Build GRCh38
rs971575390 Chr.8: 53227087-53227087 on Build GRCh38
rs1049984592 Chr.8: 53227222-53227222 on Build GRCh38
rs200602437 Chr.8: 53226937-53226937 on Build GRCh38
rs201507979 Chr.8: 53226940-53226940 on Build GRCh38
rs889675411 Chr.8: 53226955-53226955 on Build GRCh38
rs199582493 Chr.8: 53227198-53227198 on Build GRCh38
rs149471172 Chr.8: 53226950-53226950 on Build GRCh38
rs576949771 Chr.8: 53226921-53226921 on Build GRCh38
rs199650585 Chr.8: 53226944-53226944 on Build GRCh38
rs112960681 Chr.8: 53226983-53226983 on Build GRCh38
rs969721073 Chr.8: 53227271-53227272 on Build GRCh38
rs200923671 Chr.8: 53227253-53227253 on Build GRCh38
rs201865379 Chr.8: 53226951-53226951 on Build GRCh38
rs200309521 Chr.8: 53227082-53227082 on Build GRCh38
rs200452553 Chr.8: 53227329-53227329 on Build GRCh38
rs764044129 Chr.8: 53227124-53227124 on Build GRCh38
rs754609061 Chr.8: 53227258-53227263 on Build GRCh38
rs181250246 Chr.8: 53227036-53227036 on Build GRCh38
rs967449051 Chr.8: 53226959-53226959 on Build GRCh38
rs200377550 Chr.8: 53227282-53227282 on Build GRCh38
rs1019538988 Chr.8: 53227170-53227170 on Build GRCh38
rs200936699 Chr.8: 53227067-53227067 on Build GRCh38
rs1036072459 Chr.8: 53227110-53227110 on Build GRCh38
rs201714047 Chr.8: 53227086-53227086 on Build GRCh38
rs201048635 Chr.8: 53226894-53226894 on Build GRCh38
rs940427338 Chr.8: 53227107-53227107 on Build GRCh38
rs55675556 Chr.8: 53226997-53226997 on Build GRCh38
rs200626983 Chr.8: 53227279-53227279 on Build GRCh38

View Full Product Details