Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 72033540-72034076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSC-AS1
Entrez Gene ID
100132891
Gene Name
MSC antisense RNA 1
Gene Aliases
-
UniGene
Hs.137674
Gene Chromosome Location
Chr.8: 71843123-72056312 on Build GRCh38
Transcripts
NR_033651.1 NR_033652.1 NM_007332.2 XM_011517624.2 XM_011517625.2 XM_017013946.1

Gene Symbol
TRPA1
Entrez Gene ID
8989
Gene Name
transient receptor potential cation channel subfamily A member 1
Gene Aliases
ANKTM1, FEPS
UniGene
Hs.716816
Gene Chromosome Location
Chr.8: 72021251-72094885 on Build GRCh38
Transcripts
NR_033651.1 NR_033652.1 NM_007332.2 XM_011517624.2 XM_011517625.2 XM_017013946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3925562 c.2788G>A p.E930K Chr.8: 72033724-72033724 on Build GRCh38
7333201 c.2760G>A p.E920E Chr.8: 72033752-72033752 on Build GRCh38
3983769 c.2686-6T>C p.? Chr.8: 72033832-72033832 on Build GRCh38
285981 c.2806C>A p.L936M Chr.8: 72033706-72033706 on Build GRCh38
6353840 c.2781G>C p.L927L Chr.8: 72033731-72033731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145505945 Chr.8: 72033756-72033756 on Build GRCh38
rs904749779 Chr.8: 72033834-72033834 on Build GRCh38
rs980040557 Chr.8: 72033969-72033969 on Build GRCh38
rs938763034 Chr.8: 72033732-72033732 on Build GRCh38
rs964943536 Chr.8: 72033808-72033808 on Build GRCh38
rs764021594 Chr.8: 72033653-72033653 on Build GRCh38
rs763420017 Chr.8: 72033871-72033871 on Build GRCh38
rs548646724 Chr.8: 72033992-72033992 on Build GRCh38
rs950499612 Chr.8: 72033886-72033886 on Build GRCh38
rs769003797 Chr.8: 72033928-72033928 on Build GRCh38
rs775684425 Chr.8: 72033638-72033638 on Build GRCh38
rs147706025 Chr.8: 72033757-72033757 on Build GRCh38
rs147736980 Chr.8: 72033838-72033838 on Build GRCh38
rs202111122 Chr.8: 72033845-72033845 on Build GRCh38
rs760812691 Chr.8: 72033651-72033651 on Build GRCh38
rs548077117 Chr.8: 72033839-72033839 on Build GRCh38
rs780522671 Chr.8: 72033783-72033783 on Build GRCh38
rs927305258 Chr.8: 72033970-72033970 on Build GRCh38
rs980094355 Chr.8: 72033644-72033644 on Build GRCh38
rs778949965 Chr.8: 72033864-72033865 on Build GRCh38
rs753787981 Chr.8: 72033648-72033648 on Build GRCh38
rs377411878 Chr.8: 72033971-72033971 on Build GRCh38
rs754942638 Chr.8: 72033809-72033810 on Build GRCh38
rs994799276 Chr.8: 72034023-72034023 on Build GRCh38
rs747339003 Chr.8: 72033909-72033909 on Build GRCh38
rs769654619 Chr.8: 72033609-72033609 on Build GRCh38
rs749140186 Chr.8: 72033826-72033826 on Build GRCh38
rs751108958 Chr.8: 72033762-72033762 on Build GRCh38
rs777666406 Chr.8: 72033820-72033820 on Build GRCh38
rs750863390 Chr.8: 72033716-72033716 on Build GRCh38
rs899138051 Chr.8: 72034006-72034006 on Build GRCh38
rs1007130009 Chr.8: 72033840-72033840 on Build GRCh38
rs923685860 Chr.8: 72033853-72033853 on Build GRCh38
rs372156749 Chr.8: 72033611-72033611 on Build GRCh38
rs762183740 Chr.8: 72033691-72033691 on Build GRCh38
rs563286178 Chr.8: 72033910-72033910 on Build GRCh38
rs554911941 Chr.8: 72033873-72033873 on Build GRCh38
rs756048374 Chr.8: 72033812-72033812 on Build GRCh38
rs780000915 Chr.8: 72033618-72033619 on Build GRCh38
rs772329985 Chr.8: 72033625-72033625 on Build GRCh38
rs913265569 Chr.8: 72033980-72033980 on Build GRCh38
rs200623988 Chr.8: 72033849-72033849 on Build GRCh38
rs768050608 Chr.8: 72033593-72033593 on Build GRCh38
rs914886271 Chr.8: 72033830-72033830 on Build GRCh38
rs748149700 Chr.8: 72033810-72033810 on Build GRCh38
rs371551336 Chr.8: 72033872-72033872 on Build GRCh38
rs939151313 Chr.8: 72033902-72033902 on Build GRCh38
rs267601986 Chr.8: 72033737-72033737 on Build GRCh38
rs975013460 Chr.8: 72033815-72033815 on Build GRCh38
rs963290696 Chr.8: 72033961-72033961 on Build GRCh38
rs765805074 Chr.8: 72033715-72033715 on Build GRCh38
rs749053240 Chr.8: 72033613-72033613 on Build GRCh38
rs1019144953 Chr.8: 72033790-72033790 on Build GRCh38
rs946181006 Chr.8: 72033767-72033767 on Build GRCh38
rs200810966 Chr.8: 72033858-72033858 on Build GRCh38
rs754533416 Chr.8: 72033780-72033780 on Build GRCh38
rs941936836 Chr.8: 72033629-72033629 on Build GRCh38
rs559497085 Chr.8: 72033684-72033684 on Build GRCh38
rs528929358 Chr.8: 72033973-72033973 on Build GRCh38
rs765900389 Chr.8: 72033857-72033857 on Build GRCh38
rs747980801 Chr.8: 72033607-72033607 on Build GRCh38
rs992858359 Chr.8: 72033977-72033977 on Build GRCh38
rs781198252 Chr.8: 72033596-72033596 on Build GRCh38

View Full Product Details