Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 18531105-18531601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSD3
Entrez Gene ID
23362
Gene Name
pleckstrin and Sec7 domain containing 3
Gene Aliases
EFA6D, EFA6R, HCA67
UniGene
Hs.434255
Gene Chromosome Location
Chr.8: 18527303-19086909 on Build GRCh38
Transcripts
NM_015310.3 NM_206909.2 XM_017013258.1 XM_017013259.1 XM_017013260.1 XM_017013261.1 XM_017013265.1

SNP Detail

SNP ID Chromosome Location
rs955232262 Chr.8: 18531447-18531447 on Build GRCh38
rs957401176 Chr.8: 18531165-18531165 on Build GRCh38
rs3739395 Chr.8: 18531537-18531537 on Build GRCh38
rs147562768 Chr.8: 18531526-18531526 on Build GRCh38
rs534135853 Chr.8: 18531480-18531480 on Build GRCh38
rs779365963 Chr.8: 18531154-18531154 on Build GRCh38
rs747716280 Chr.8: 18531329-18531329 on Build GRCh38
rs80335347 Chr.8: 18531471-18531471 on Build GRCh38
rs973585845 Chr.8: 18531530-18531530 on Build GRCh38
rs905808578 Chr.8: 18531426-18531426 on Build GRCh38
rs955413353 Chr.8: 18531212-18531212 on Build GRCh38
rs974728688 Chr.8: 18531323-18531323 on Build GRCh38
rs898114102 Chr.8: 18531179-18531179 on Build GRCh38
rs185220676 Chr.8: 18531185-18531185 on Build GRCh38
rs144532442 Chr.8: 18531361-18531361 on Build GRCh38
rs562934620 Chr.8: 18531378-18531378 on Build GRCh38
rs922202434 Chr.8: 18531325-18531325 on Build GRCh38
rs557936404 Chr.8: 18531385-18531385 on Build GRCh38
rs975920661 Chr.8: 18531182-18531182 on Build GRCh38
rs990570081 Chr.8: 18531560-18531560 on Build GRCh38
rs145273912 Chr.8: 18531500-18531500 on Build GRCh38
rs547539697 Chr.8: 18531493-18531493 on Build GRCh38
rs541184398 Chr.8: 18531284-18531284 on Build GRCh38
rs891583840 Chr.8: 18531444-18531444 on Build GRCh38
rs34795038 Chr.8: 18531468-18531469 on Build GRCh38
rs201864081 Chr.8: 18531467-18531467 on Build GRCh38
rs919597299 Chr.8: 18531321-18531321 on Build GRCh38
rs1033017175 Chr.8: 18531152-18531152 on Build GRCh38
rs767925851 Chr.8: 18531293-18531296 on Build GRCh38
rs1055298487 Chr.8: 18531332-18531332 on Build GRCh38
rs3832596 Chr.8: 18531467-18531468 on Build GRCh38
rs894632976 Chr.8: 18531360-18531360 on Build GRCh38
rs1002686609 Chr.8: 18531198-18531198 on Build GRCh38
rs535910896 Chr.8: 18531502-18531502 on Build GRCh38
rs72589245 Chr.8: 18531469-18531470 on Build GRCh38
rs1022156419 Chr.8: 18531466-18531467 on Build GRCh38
rs1044022947 Chr.8: 18531390-18531390 on Build GRCh38
rs987649872 Chr.8: 18531337-18531337 on Build GRCh38
rs933535532 Chr.8: 18531336-18531336 on Build GRCh38
rs999109340 Chr.8: 18531396-18531396 on Build GRCh38
rs769308518 Chr.8: 18531354-18531354 on Build GRCh38
rs775294417 Chr.8: 18531310-18531310 on Build GRCh38
rs117985629 Chr.8: 18531439-18531439 on Build GRCh38
rs908506077 Chr.8: 18531233-18531233 on Build GRCh38
rs908007305 Chr.8: 18531348-18531348 on Build GRCh38
rs1037967991 Chr.8: 18531292-18531292 on Build GRCh38
rs529044342 Chr.8: 18531511-18531511 on Build GRCh38
rs1236848 Chr.8: 18531445-18531445 on Build GRCh38
rs538243620 Chr.8: 18531535-18531535 on Build GRCh38
rs201044545 Chr.8: 18531465-18531465 on Build GRCh38
rs116930653 Chr.8: 18531429-18531429 on Build GRCh38
rs149305101 Chr.8: 18531247-18531247 on Build GRCh38
rs1032987228 Chr.8: 18531418-18531418 on Build GRCh38
rs17126756 Chr.8: 18531232-18531232 on Build GRCh38
rs893153999 Chr.8: 18531428-18531428 on Build GRCh38
rs543479749 Chr.8: 18531499-18531499 on Build GRCh38
rs191029215 Chr.8: 18531509-18531509 on Build GRCh38
rs377318094 Chr.8: 18531473-18531473 on Build GRCh38
rs549414136 Chr.8: 18531368-18531368 on Build GRCh38
rs560569701 Chr.8: 18531346-18531346 on Build GRCh38
rs908113947 Chr.8: 18531514-18531514 on Build GRCh38
rs397815662 Chr.8: 18531470-18531471 on Build GRCh38
rs139781876 Chr.8: 18531389-18531389 on Build GRCh38
rs756212929 Chr.8: 18531519-18531519 on Build GRCh38
rs545312531 Chr.8: 18531209-18531209 on Build GRCh38

View Full Product Details