Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27325401-27325902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTK2B
Entrez Gene ID
2185
Gene Name
protein tyrosine kinase 2 beta
Gene Aliases
CADTK, CAKB, FADK2, FAK2, PKB, PTK, PYK2, RAFTK
UniGene
Hs.491322
Gene Chromosome Location
Chr.8: 27311482-27459391 on Build GRCh38
Transcripts
NM_004103.4 NM_173174.2 NM_173175.2 NM_173176.2 XM_005273448.4 XM_011544442.2 XM_017013214.1 XM_017013216.1

SNP Detail

SNP ID Chromosome Location
rs912484037 Chr.8: 27325425-27325425 on Build GRCh38
rs372931497 Chr.8: 27325697-27325697 on Build GRCh38
rs75917942 Chr.8: 27325768-27325768 on Build GRCh38
rs950031056 Chr.8: 27325670-27325670 on Build GRCh38
rs553863389 Chr.8: 27325797-27325797 on Build GRCh38
rs1048847454 Chr.8: 27325712-27325712 on Build GRCh38
rs575026972 Chr.8: 27325755-27325755 on Build GRCh38
rs906790648 Chr.8: 27325594-27325594 on Build GRCh38
rs567189210 Chr.8: 27325672-27325672 on Build GRCh38
rs1007778353 Chr.8: 27325533-27325533 on Build GRCh38
rs896849270 Chr.8: 27325622-27325622 on Build GRCh38
rs755651364 Chr.8: 27325652-27325652 on Build GRCh38
rs867979493 Chr.8: 27325575-27325575 on Build GRCh38
rs897553402 Chr.8: 27325449-27325449 on Build GRCh38
rs995635311 Chr.8: 27325624-27325624 on Build GRCh38
rs896792049 Chr.8: 27325865-27325865 on Build GRCh38
rs1053891364 Chr.8: 27325541-27325541 on Build GRCh38
rs569071659 Chr.8: 27325659-27325659 on Build GRCh38
rs752075094 Chr.8: 27325616-27325616 on Build GRCh38
rs1046550439 Chr.8: 27325831-27325831 on Build GRCh38
rs1048731339 Chr.8: 27325472-27325472 on Build GRCh38
rs902076523 Chr.8: 27325778-27325778 on Build GRCh38
rs959485477 Chr.8: 27325876-27325876 on Build GRCh38
rs979504169 Chr.8: 27325603-27325603 on Build GRCh38
rs1032380395 Chr.8: 27325872-27325872 on Build GRCh38
rs754612883 Chr.8: 27325756-27325756 on Build GRCh38
rs990812062 Chr.8: 27325649-27325651 on Build GRCh38
rs895273943 Chr.8: 27325552-27325552 on Build GRCh38
rs546270837 Chr.8: 27325524-27325524 on Build GRCh38
rs1007225853 Chr.8: 27325751-27325756 on Build GRCh38
rs535488591 Chr.8: 27325734-27325734 on Build GRCh38
rs916670635 Chr.8: 27325807-27325807 on Build GRCh38
rs948174431 Chr.8: 27325826-27325826 on Build GRCh38
rs936724002 Chr.8: 27325482-27325482 on Build GRCh38
rs1026743124 Chr.8: 27325625-27325625 on Build GRCh38
rs998206677 Chr.8: 27325800-27325800 on Build GRCh38
rs76696915 Chr.8: 27325560-27325560 on Build GRCh38
rs978080925 Chr.8: 27325463-27325463 on Build GRCh38
rs906739770 Chr.8: 27325839-27325839 on Build GRCh38
rs571052218 Chr.8: 27325473-27325473 on Build GRCh38
rs1005124634 Chr.8: 27325596-27325596 on Build GRCh38
rs995186315 Chr.8: 27325873-27325873 on Build GRCh38
rs993277013 Chr.8: 27325464-27325464 on Build GRCh38
rs1046013596 Chr.8: 27325447-27325447 on Build GRCh38
rs943977177 Chr.8: 27325444-27325444 on Build GRCh38
rs971494058 Chr.8: 27325731-27325732 on Build GRCh38
rs1021097781 Chr.8: 27325853-27325853 on Build GRCh38
rs989935254 Chr.8: 27325801-27325801 on Build GRCh38
rs940928178 Chr.8: 27325845-27325845 on Build GRCh38
rs566499168 Chr.8: 27325574-27325574 on Build GRCh38
rs1036590685 Chr.8: 27325860-27325860 on Build GRCh38
rs140031457 Chr.8: 27325582-27325582 on Build GRCh38
rs41312168 Chr.8: 27325542-27325542 on Build GRCh38
rs549255958 Chr.8: 27325581-27325581 on Build GRCh38
rs775794484 Chr.8: 27325580-27325580 on Build GRCh38
rs753561486 Chr.8: 27325681-27325681 on Build GRCh38
rs765978132 Chr.8: 27325656-27325656 on Build GRCh38
rs936666828 Chr.8: 27325781-27325781 on Build GRCh38
rs555195396 Chr.8: 27325742-27325742 on Build GRCh38
rs923916495 Chr.8: 27325769-27325769 on Build GRCh38
rs531321210 Chr.8: 27325717-27325717 on Build GRCh38
rs919099581 Chr.8: 27325711-27325711 on Build GRCh38
rs112215847 Chr.8: 27325847-27325847 on Build GRCh38

View Full Product Details