Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144877003-144877550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF250
Entrez Gene ID
58500
Gene Name
zinc finger protein 250
Gene Aliases
ZFP647, ZNF647
UniGene
Hs.532277
Gene Chromosome Location
Chr.8: 144876951-144902311 on Build GRCh38
Transcripts
NM_001109689.3 NM_021061.4 XM_006716612.3 XM_006716613.3

SNP Detail

SNP ID Chromosome Location
rs950210505 Chr.8: 144877128-144877128 on Build GRCh38
rs984470990 Chr.8: 144877244-144877245 on Build GRCh38
rs370306211 Chr.8: 144877356-144877356 on Build GRCh38
rs936003273 Chr.8: 144877482-144877482 on Build GRCh38
rs1209853 Chr.8: 144877525-144877525 on Build GRCh38
rs939532491 Chr.8: 144877274-144877274 on Build GRCh38
rs753257319 Chr.8: 144877100-144877100 on Build GRCh38
rs887451157 Chr.8: 144877317-144877317 on Build GRCh38
rs1010889396 Chr.8: 144877072-144877072 on Build GRCh38
rs972261349 Chr.8: 144877183-144877183 on Build GRCh38
rs1054309903 Chr.8: 144877213-144877213 on Build GRCh38
rs910105256 Chr.8: 144877060-144877060 on Build GRCh38
rs1132534 Chr.8: 144877225-144877225 on Build GRCh38
rs1209854 Chr.8: 144877340-144877340 on Build GRCh38
rs536426985 Chr.8: 144877485-144877485 on Build GRCh38
rs754127803 Chr.8: 144877382-144877382 on Build GRCh38
rs572903212 Chr.8: 144877476-144877476 on Build GRCh38
rs550632236 Chr.8: 144877110-144877110 on Build GRCh38
rs915558736 Chr.8: 144877253-144877254 on Build GRCh38
rs557638887 Chr.8: 144877374-144877374 on Build GRCh38
rs189570911 Chr.8: 144877280-144877280 on Build GRCh38
rs764538798 Chr.8: 144877329-144877329 on Build GRCh38
rs528851548 Chr.8: 144877083-144877083 on Build GRCh38
rs1007934293 Chr.8: 144877233-144877233 on Build GRCh38
rs1028716439 Chr.8: 144877302-144877302 on Build GRCh38
rs993330008 Chr.8: 144877378-144877378 on Build GRCh38
rs568555755 Chr.8: 144877271-144877271 on Build GRCh38
rs984796197 Chr.8: 144877052-144877052 on Build GRCh38
rs980607436 Chr.8: 144877431-144877431 on Build GRCh38
rs180805871 Chr.8: 144877296-144877296 on Build GRCh38
rs889081654 Chr.8: 144877344-144877344 on Build GRCh38
rs961659101 Chr.8: 144877337-144877337 on Build GRCh38
rs570344620 Chr.8: 144877174-144877174 on Build GRCh38
rs965897255 Chr.8: 144877143-144877143 on Build GRCh38
rs552246390 Chr.8: 144877112-144877113 on Build GRCh38
rs961543859 Chr.8: 144877106-144877106 on Build GRCh38
rs1022222945 Chr.8: 144877095-144877095 on Build GRCh38
rs748782373 Chr.8: 144877383-144877383 on Build GRCh38
rs980194096 Chr.8: 144877145-144877145 on Build GRCh38
rs756535424 Chr.8: 144877109-144877109 on Build GRCh38
rs1022647613 Chr.8: 144877480-144877480 on Build GRCh38
rs79881333 Chr.8: 144877190-144877190 on Build GRCh38
rs535465120 Chr.8: 144877277-144877277 on Build GRCh38
rs990150058 Chr.8: 144877488-144877488 on Build GRCh38
rs553162578 Chr.8: 144877238-144877238 on Build GRCh38
rs927304961 Chr.8: 144877146-144877146 on Build GRCh38
rs561844859 Chr.8: 144877074-144877074 on Build GRCh38
rs746503544 Chr.8: 144877445-144877445 on Build GRCh38
rs1132533 Chr.8: 144877254-144877254 on Build GRCh38
rs1040216848 Chr.8: 144877263-144877263 on Build GRCh38
rs78716787 Chr.8: 144877514-144877514 on Build GRCh38
rs184752294 Chr.8: 144877193-144877193 on Build GRCh38
rs1007463524 Chr.8: 144877354-144877354 on Build GRCh38
rs535225043 Chr.8: 144877236-144877236 on Build GRCh38
rs1005850130 Chr.8: 144877323-144877323 on Build GRCh38
rs951725813 Chr.8: 144877048-144877048 on Build GRCh38
rs927367331 Chr.8: 144877473-144877473 on Build GRCh38
rs981061281 Chr.8: 144877526-144877526 on Build GRCh38
rs570917211 Chr.8: 144877231-144877231 on Build GRCh38
rs369309962 Chr.8: 144877518-144877518 on Build GRCh38
rs948288807 Chr.8: 144877254-144877254 on Build GRCh38

View Full Product Details