Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 1055591-1056138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRT2
Entrez Gene ID
10655
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
UniGene
Hs.59506
Gene Chromosome Location
Chr.9: 1003237-1057554 on Build GRCh38
Transcripts
NM_001130865.2 NM_006557.6 NM_181872.4 XM_011517687.1 XM_011517690.2 XM_011517694.2 XM_017014213.1 XM_017014214.1 XM_017014215.1 XM_017014216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7147349 c.676C>G p.L226V Chr.9: 1055769-1055769 on Build GRCh38
3367477 c.653A>G p.Y218C Chr.9: 1055746-1055746 on Build GRCh38
3167428 c.674A>G p.H225R Chr.9: 1055767-1055767 on Build GRCh38
5805840 c.677T>A p.L226H Chr.9: 1055770-1055770 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776835597 Chr.9: 1055710-1055710 on Build GRCh38
rs1000556180 Chr.9: 1055878-1055878 on Build GRCh38
rs986710318 Chr.9: 1055625-1055625 on Build GRCh38
rs779575754 Chr.9: 1055782-1055782 on Build GRCh38
rs753089394 Chr.9: 1055961-1055961 on Build GRCh38
rs1018201341 Chr.9: 1055894-1055894 on Build GRCh38
rs753664115 Chr.9: 1055638-1055638 on Build GRCh38
rs115628248 Chr.9: 1056052-1056052 on Build GRCh38
rs767425170 Chr.9: 1055705-1055705 on Build GRCh38
rs867910424 Chr.9: 1056041-1056041 on Build GRCh38
rs1053460848 Chr.9: 1056061-1056061 on Build GRCh38
rs747929955 Chr.9: 1055787-1055787 on Build GRCh38
rs761984453 Chr.9: 1056102-1056102 on Build GRCh38
rs764103857 Chr.9: 1056005-1056005 on Build GRCh38
rs998558025 Chr.9: 1055864-1055864 on Build GRCh38
rs970637881 Chr.9: 1055756-1055756 on Build GRCh38
rs755166695 Chr.9: 1055928-1055928 on Build GRCh38
rs976119024 Chr.9: 1055930-1055930 on Build GRCh38
rs976026079 Chr.9: 1055659-1055659 on Build GRCh38
rs200747181 Chr.9: 1055802-1055802 on Build GRCh38
rs6474554 Chr.9: 1055833-1055833 on Build GRCh38
rs545363391 Chr.9: 1056077-1056077 on Build GRCh38
rs949905362 Chr.9: 1056023-1056023 on Build GRCh38
rs966265368 Chr.9: 1055909-1055909 on Build GRCh38
rs1045531792 Chr.9: 1055852-1055852 on Build GRCh38
rs774186247 Chr.9: 1055785-1055785 on Build GRCh38
rs1033617798 Chr.9: 1055957-1055957 on Build GRCh38
rs781763532 Chr.9: 1055747-1055747 on Build GRCh38
rs982868346 Chr.9: 1056019-1056019 on Build GRCh38
rs1056054227 Chr.9: 1055818-1055818 on Build GRCh38
rs927269738 Chr.9: 1055788-1055788 on Build GRCh38
rs761786159 Chr.9: 1055704-1055704 on Build GRCh38
rs752888143 Chr.9: 1055718-1055718 on Build GRCh38
rs750522995 Chr.9: 1055690-1055691 on Build GRCh38
rs773032376 Chr.9: 1055792-1055792 on Build GRCh38
rs764399352 Chr.9: 1055823-1055823 on Build GRCh38
rs530324849 Chr.9: 1055666-1055666 on Build GRCh38
rs752039179 Chr.9: 1055825-1055825 on Build GRCh38
rs759535088 Chr.9: 1055713-1055713 on Build GRCh38
rs893357247 Chr.9: 1056065-1056065 on Build GRCh38
rs893379124 Chr.9: 1055891-1055891 on Build GRCh38
rs112564390 Chr.9: 1055746-1055746 on Build GRCh38
rs751953407 Chr.9: 1055740-1055740 on Build GRCh38
rs527977472 Chr.9: 1056103-1056103 on Build GRCh38
rs902851404 Chr.9: 1055854-1055854 on Build GRCh38
rs375024785 Chr.9: 1055964-1055968 on Build GRCh38
rs1032479709 Chr.9: 1055882-1055882 on Build GRCh38
rs542434676 Chr.9: 1056050-1056050 on Build GRCh38
rs760615461 Chr.9: 1055793-1055793 on Build GRCh38
rs57558840 Chr.9: 1055975-1055975 on Build GRCh38
rs567898162 Chr.9: 1055655-1055655 on Build GRCh38
rs144855636 Chr.9: 1055631-1055631 on Build GRCh38
rs957555732 Chr.9: 1055621-1055621 on Build GRCh38
rs974114094 Chr.9: 1055976-1055976 on Build GRCh38
rs950712213 Chr.9: 1055936-1055936 on Build GRCh38
rs944286982 Chr.9: 1055866-1055866 on Build GRCh38
rs777706209 Chr.9: 1055685-1055686 on Build GRCh38
rs553542064 Chr.9: 1055892-1055892 on Build GRCh38
rs576494807 Chr.9: 1056075-1056075 on Build GRCh38
rs980669935 Chr.9: 1055951-1055951 on Build GRCh38
rs756143560 Chr.9: 1055696-1055699 on Build GRCh38
rs1046962730 Chr.9: 1055768-1055768 on Build GRCh38
rs748920345 Chr.9: 1055678-1055678 on Build GRCh38
rs749556839 Chr.9: 1055811-1055811 on Build GRCh38
rs755600953 Chr.9: 1055776-1055776 on Build GRCh38
rs549014161 Chr.9: 1055680-1055680 on Build GRCh38
rs1039963337 Chr.9: 1055868-1055871 on Build GRCh38
rs879211032 Chr.9: 1056062-1056062 on Build GRCh38
rs746320247 Chr.9: 1055765-1055765 on Build GRCh38
rs758567170 Chr.9: 1055721-1055721 on Build GRCh38
rs78320923 Chr.9: 1055846-1055846 on Build GRCh38
rs892166636 Chr.9: 1056067-1056067 on Build GRCh38
rs568883963 Chr.9: 1055681-1055681 on Build GRCh38
rs537878305 Chr.9: 1055660-1055661 on Build GRCh38
rs749744094 Chr.9: 1056036-1056036 on Build GRCh38
rs770540681 Chr.9: 1056055-1056055 on Build GRCh38
rs778355444 Chr.9: 1055979-1055979 on Build GRCh38
rs909776942 Chr.9: 1055960-1055960 on Build GRCh38
rs371107724 Chr.9: 1055679-1055679 on Build GRCh38
rs567611497 Chr.9: 1055789-1055789 on Build GRCh38
rs902016911 Chr.9: 1056109-1056109 on Build GRCh38
rs775611126 Chr.9: 1055805-1055805 on Build GRCh38
rs912718250 Chr.9: 1056034-1056034 on Build GRCh38
rs915713701 Chr.9: 1055999-1055999 on Build GRCh38
rs12350001 Chr.9: 1055870-1055870 on Build GRCh38
rs956722261 Chr.9: 1055667-1055667 on Build GRCh38
rs763319388 Chr.9: 1055810-1055810 on Build GRCh38
rs780075531 Chr.9: 1055810-1055810 on Build GRCh38
rs902944954 Chr.9: 1056042-1056042 on Build GRCh38
rs1026396885 Chr.9: 1055933-1055933 on Build GRCh38
rs748931401 Chr.9: 1055784-1055784 on Build GRCh38
rs758657376 Chr.9: 1055723-1055723 on Build GRCh38
rs562571430 Chr.9: 1055724-1055724 on Build GRCh38
rs936303512 Chr.9: 1056060-1056060 on Build GRCh38
rs528018527 Chr.9: 1055622-1055622 on Build GRCh38
rs781339152 Chr.9: 1055948-1055948 on Build GRCh38
rs1001010560 Chr.9: 1055661-1055661 on Build GRCh38
rs7854435 Chr.9: 1056101-1056101 on Build GRCh38
rs533882403 Chr.9: 1055884-1055884 on Build GRCh38

View Full Product Details