Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4576478-4576981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC1A1
Entrez Gene ID
6505
Gene Name
solute carrier family 1 member 1
Gene Aliases
DCBXA, EAAC1, EAAT3, SCZD18
UniGene
Hs.444915
Gene Chromosome Location
Chr.9: 4490427-4587469 on Build GRCh38
Transcripts
NM_004170.5 XM_011518007.1 XM_011518008.2 XM_011518009.2 XM_011518010.1 XM_017015042.1 XM_017015043.1 XM_017014882.1

Gene Symbol
SPATA6L
Entrez Gene ID
55064
Gene Name
spermatogenesis associated 6 like
Gene Aliases
C9orf68, bA6J24.2
UniGene
Hs.179615
Gene Chromosome Location
Chr.9: 4540092-4666674 on Build GRCh38
Transcripts
NM_004170.5 XM_011518007.1 XM_011518008.2 XM_011518009.2 XM_011518010.1 XM_017015042.1 XM_017015043.1 XM_017014882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5539048 c.1013C>T p.P338L Chr.9: 4576583-4576583 on Build GRCh38
6868920 c.1050G>A p.V350V Chr.9: 4576620-4576620 on Build GRCh38
1462533 c.1071C>T p.F357F Chr.9: 4576641-4576641 on Build GRCh38
2777055 c.1026C>T p.R342R Chr.9: 4576596-4576596 on Build GRCh38
487480 c.1044C>G p.N348K Chr.9: 4576614-4576614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966788409 Chr.9: 4576890-4576890 on Build GRCh38
rs757318003 Chr.9: 4576731-4576731 on Build GRCh38
rs1471786 Chr.9: 4576774-4576774 on Build GRCh38
rs545612752 Chr.9: 4576937-4576937 on Build GRCh38
rs939779561 Chr.9: 4576959-4576959 on Build GRCh38
rs759267334 Chr.9: 4576706-4576706 on Build GRCh38
rs1032767607 Chr.9: 4576905-4576905 on Build GRCh38
rs369270825 Chr.9: 4576594-4576594 on Build GRCh38
rs954725459 Chr.9: 4576841-4576841 on Build GRCh38
rs760130029 Chr.9: 4576786-4576786 on Build GRCh38
rs774159792 Chr.9: 4576681-4576681 on Build GRCh38
rs750865075 Chr.9: 4576742-4576742 on Build GRCh38
rs760713773 Chr.9: 4576703-4576703 on Build GRCh38
rs756790302 Chr.9: 4576883-4576883 on Build GRCh38
rs757336485 Chr.9: 4576657-4576657 on Build GRCh38
rs373554585 Chr.9: 4576650-4576650 on Build GRCh38
rs938289788 Chr.9: 4576698-4576698 on Build GRCh38
rs370380719 Chr.9: 4576570-4576570 on Build GRCh38
rs543643470 Chr.9: 4576855-4576855 on Build GRCh38
rs780907696 Chr.9: 4576892-4576892 on Build GRCh38
rs775964685 Chr.9: 4576679-4576679 on Build GRCh38
rs779000661 Chr.9: 4576545-4576545 on Build GRCh38
rs761430315 Chr.9: 4576637-4576637 on Build GRCh38
rs763472138 Chr.9: 4576625-4576625 on Build GRCh38
rs369261520 Chr.9: 4576547-4576547 on Build GRCh38
rs780033370 Chr.9: 4576920-4576920 on Build GRCh38
rs927716214 Chr.9: 4576953-4576953 on Build GRCh38
rs969552682 Chr.9: 4576936-4576936 on Build GRCh38
rs773968205 Chr.9: 4576636-4576636 on Build GRCh38
rs1010360168 Chr.9: 4576504-4576504 on Build GRCh38
rs763672191 Chr.9: 4576519-4576519 on Build GRCh38
rs771402016 Chr.9: 4576560-4576560 on Build GRCh38
rs143926021 Chr.9: 4576651-4576651 on Build GRCh38
rs372708972 Chr.9: 4576641-4576641 on Build GRCh38
rs890965833 Chr.9: 4576852-4576852 on Build GRCh38
rs147755482 Chr.9: 4576909-4576909 on Build GRCh38
rs538800675 Chr.9: 4576719-4576719 on Build GRCh38
rs587777697 Chr.9: 4576754-4576756 on Build GRCh38
rs301430 Chr.9: 4576680-4576680 on Build GRCh38
rs140093899 Chr.9: 4576527-4576527 on Build GRCh38
rs908279377 Chr.9: 4576944-4576944 on Build GRCh38
rs1008049120 Chr.9: 4576863-4576863 on Build GRCh38
rs1020991638 Chr.9: 4576885-4576885 on Build GRCh38
rs770034584 Chr.9: 4576670-4576670 on Build GRCh38
rs374679151 Chr.9: 4576552-4576552 on Build GRCh38
rs781300108 Chr.9: 4576739-4576739 on Build GRCh38
rs763550394 Chr.9: 4576815-4576815 on Build GRCh38
rs746224512 Chr.9: 4576581-4576581 on Build GRCh38
rs756032741 Chr.9: 4576539-4576539 on Build GRCh38
rs150295646 Chr.9: 4576812-4576812 on Build GRCh38
rs376223134 Chr.9: 4576642-4576642 on Build GRCh38
rs756604649 Chr.9: 4576744-4576744 on Build GRCh38
rs745561503 Chr.9: 4576901-4576901 on Build GRCh38
rs866673192 Chr.9: 4576876-4576876 on Build GRCh38
rs750221771 Chr.9: 4576528-4576528 on Build GRCh38
rs201587651 Chr.9: 4576595-4576595 on Build GRCh38
rs570228124 Chr.9: 4576704-4576704 on Build GRCh38
rs561950227 Chr.9: 4576710-4576710 on Build GRCh38
rs564902026 Chr.9: 4576785-4576785 on Build GRCh38
rs1014873097 Chr.9: 4576858-4576858 on Build GRCh38
rs772885883 Chr.9: 4576775-4576775 on Build GRCh38
rs780447467 Chr.9: 4576746-4576746 on Build GRCh38
rs554916480 Chr.9: 4576840-4576840 on Build GRCh38
rs375261823 Chr.9: 4576788-4576788 on Build GRCh38
rs774554523 Chr.9: 4576626-4576626 on Build GRCh38
rs372540843 Chr.9: 4576548-4576548 on Build GRCh38
rs770847366 Chr.9: 4576799-4576799 on Build GRCh38
rs563383019 Chr.9: 4576904-4576904 on Build GRCh38
rs749608311 Chr.9: 4576759-4576759 on Build GRCh38
rs925538090 Chr.9: 4576695-4576695 on Build GRCh38
rs767200763 Chr.9: 4576683-4576683 on Build GRCh38
rs752836977 Chr.9: 4576648-4576648 on Build GRCh38
rs968820336 Chr.9: 4576509-4576509 on Build GRCh38
rs374031340 Chr.9: 4576664-4576664 on Build GRCh38
rs747479410 Chr.9: 4576554-4576554 on Build GRCh38
rs755531811 Chr.9: 4576790-4576790 on Build GRCh38
rs1003916694 Chr.9: 4576606-4576606 on Build GRCh38
rs920366823 Chr.9: 4576617-4576617 on Build GRCh38
rs766450812 Chr.9: 4576688-4576689 on Build GRCh38
rs80238687 Chr.9: 4576688-4576688 on Build GRCh38
rs751848390 Chr.9: 4576720-4576720 on Build GRCh38
rs768905004 Chr.9: 4576760-4576760 on Build GRCh38
rs301979 Chr.9: 4576851-4576851 on Build GRCh38
rs567586426 Chr.9: 4576585-4576585 on Build GRCh38
rs199834296 Chr.9: 4576565-4576565 on Build GRCh38
rs761339722 Chr.9: 4576682-4576682 on Build GRCh38
rs761087718 Chr.9: 4576662-4576662 on Build GRCh38
rs572378464 Chr.9: 4576809-4576809 on Build GRCh38
rs778701407 Chr.9: 4576768-4576768 on Build GRCh38
rs1022154653 Chr.9: 4576505-4576505 on Build GRCh38
rs751493278 Chr.9: 4576522-4576522 on Build GRCh38
rs987350407 Chr.9: 4576566-4576566 on Build GRCh38
rs747858391 Chr.9: 4576769-4576769 on Build GRCh38
rs920402588 Chr.9: 4576918-4576918 on Build GRCh38

View Full Product Details