Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4697546-4698078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC37L1
Entrez Gene ID
55664
Gene Name
cell division cycle 37 like 1
Gene Aliases
CDC37B, HARC
UniGene
Hs.666439
Gene Chromosome Location
Chr.9: 4679553-4708399 on Build GRCh38
Transcripts
NM_017913.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6427728 c.643C>T p.Q215* Chr.9: 4697775-4697775 on Build GRCh38
2777145 c.670C>T p.Q224* Chr.9: 4697802-4697802 on Build GRCh38
4440172 c.691delA p.N232fs*39 Chr.9: 4697823-4697823 on Build GRCh38
337199 c.653A>G p.H218R Chr.9: 4697785-4697785 on Build GRCh38
4956606 c.735C>T p.F245F Chr.9: 4697867-4697867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759540796 Chr.9: 4697726-4697726 on Build GRCh38
rs912444494 Chr.9: 4697925-4697925 on Build GRCh38
rs1024018169 Chr.9: 4697694-4697694 on Build GRCh38
rs954000905 Chr.9: 4697851-4697851 on Build GRCh38
rs958473828 Chr.9: 4697668-4697668 on Build GRCh38
rs192448371 Chr.9: 4698034-4698034 on Build GRCh38
rs777441416 Chr.9: 4697901-4697901 on Build GRCh38
rs1002451496 Chr.9: 4697591-4697591 on Build GRCh38
rs780299848 Chr.9: 4697861-4697861 on Build GRCh38
rs368151824 Chr.9: 4697929-4697929 on Build GRCh38
rs945394123 Chr.9: 4697998-4697998 on Build GRCh38
rs1044490647 Chr.9: 4697988-4697988 on Build GRCh38
rs979870036 Chr.9: 4697729-4697729 on Build GRCh38
rs201155602 Chr.9: 4697919-4697919 on Build GRCh38
rs892715137 Chr.9: 4697597-4697598 on Build GRCh38
rs759324804 Chr.9: 4697927-4697927 on Build GRCh38
rs764882435 Chr.9: 4697748-4697748 on Build GRCh38
rs903993647 Chr.9: 4697611-4697611 on Build GRCh38
rs756219911 Chr.9: 4697857-4697857 on Build GRCh38
rs960868977 Chr.9: 4697618-4697618 on Build GRCh38
rs71496488 Chr.9: 4697589-4697589 on Build GRCh38
rs376013391 Chr.9: 4697996-4697996 on Build GRCh38
rs1028315658 Chr.9: 4697760-4697760 on Build GRCh38
rs562180640 Chr.9: 4697912-4697912 on Build GRCh38
rs905473710 Chr.9: 4697577-4697577 on Build GRCh38
rs886116751 Chr.9: 4697830-4697830 on Build GRCh38
rs771690085 Chr.9: 4697898-4697898 on Build GRCh38
rs780345021 Chr.9: 4698033-4698034 on Build GRCh38
rs750511751 Chr.9: 4697827-4697827 on Build GRCh38
rs1009704814 Chr.9: 4697635-4697635 on Build GRCh38
rs1051126460 Chr.9: 4697950-4697950 on Build GRCh38
rs377091767 Chr.9: 4697914-4697914 on Build GRCh38
rs377175699 Chr.9: 4697653-4697653 on Build GRCh38
rs931067268 Chr.9: 4697983-4697983 on Build GRCh38
rs919696123 Chr.9: 4697971-4697971 on Build GRCh38
rs968141098 Chr.9: 4697699-4697699 on Build GRCh38
rs1002805218 Chr.9: 4697645-4697645 on Build GRCh38
rs397831599 Chr.9: 4697605-4697606 on Build GRCh38
rs1056740179 Chr.9: 4698020-4698020 on Build GRCh38
rs572932550 Chr.9: 4697943-4697943 on Build GRCh38
rs73643843 Chr.9: 4698017-4698017 on Build GRCh38
rs188910311 Chr.9: 4697964-4697964 on Build GRCh38
rs1009737269 Chr.9: 4697597-4697597 on Build GRCh38
rs542686952 Chr.9: 4697700-4697700 on Build GRCh38
rs749696827 Chr.9: 4697875-4697875 on Build GRCh38
rs3215723 Chr.9: 4697595-4697596 on Build GRCh38
rs951034182 Chr.9: 4697744-4697744 on Build GRCh38
rs113526026 Chr.9: 4697653-4697654 on Build GRCh38
rs184358515 Chr.9: 4698043-4698043 on Build GRCh38
rs776512488 Chr.9: 4697722-4697722 on Build GRCh38
rs757419292 Chr.9: 4697812-4697812 on Build GRCh38
rs748603313 Chr.9: 4697896-4697896 on Build GRCh38
rs879440031 Chr.9: 4697834-4697834 on Build GRCh38
rs371933343 Chr.9: 4697890-4697890 on Build GRCh38
rs1035227577 Chr.9: 4697613-4697613 on Build GRCh38
rs767912433 Chr.9: 4697813-4697813 on Build GRCh38
rs553847782 Chr.9: 4697616-4697616 on Build GRCh38
rs535132383 Chr.9: 4697650-4697650 on Build GRCh38
rs183365350 Chr.9: 4697932-4697932 on Build GRCh38
rs992487254 Chr.9: 4697673-4697673 on Build GRCh38
rs762635436 Chr.9: 4697579-4697579 on Build GRCh38
rs779140352 Chr.9: 4697891-4697891 on Build GRCh38

View Full Product Details