Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4710963-4711476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK3
Entrez Gene ID
50808
Gene Name
adenylate kinase 3
Gene Aliases
AK3L1, AK6, AKL3L, AKL3L1, FIX
UniGene
Hs.732022
Gene Chromosome Location
Chr.9: 4709556-4742043 on Build GRCh38
Transcripts
NM_001199852.1 NM_001199853.1 NM_001199855.1 NM_001199856.1 NM_016282.3

SNP Detail

SNP ID Chromosome Location
rs12256 Chr.9: 4711367-4711367 on Build GRCh38
rs138386660 Chr.9: 4711362-4711362 on Build GRCh38
rs1026394817 Chr.9: 4711421-4711421 on Build GRCh38
rs760612846 Chr.9: 4711094-4711094 on Build GRCh38
rs547106050 Chr.9: 4711386-4711386 on Build GRCh38
rs35711809 Chr.9: 4711046-4711046 on Build GRCh38
rs867083062 Chr.9: 4711055-4711055 on Build GRCh38
rs984686241 Chr.9: 4711436-4711436 on Build GRCh38
rs1033403815 Chr.9: 4711405-4711405 on Build GRCh38
rs148099967 Chr.9: 4711091-4711091 on Build GRCh38
rs896144556 Chr.9: 4711410-4711410 on Build GRCh38
rs1021745089 Chr.9: 4711199-4711199 on Build GRCh38
rs368620967 Chr.9: 4711003-4711003 on Build GRCh38
rs182521651 Chr.9: 4711341-4711341 on Build GRCh38
rs1039081628 Chr.9: 4711056-4711056 on Build GRCh38
rs920042020 Chr.9: 4711107-4711107 on Build GRCh38
rs755115998 Chr.9: 4711281-4711281 on Build GRCh38
rs950691644 Chr.9: 4711261-4711261 on Build GRCh38
rs531668230 Chr.9: 4711228-4711228 on Build GRCh38
rs1038461645 Chr.9: 4711102-4711102 on Build GRCh38
rs191099398 Chr.9: 4711155-4711155 on Build GRCh38
rs186664995 Chr.9: 4711411-4711411 on Build GRCh38
rs943455325 Chr.9: 4711315-4711315 on Build GRCh38
rs964399754 Chr.9: 4711007-4711007 on Build GRCh38
rs568916441 Chr.9: 4711447-4711448 on Build GRCh38
rs186355982 Chr.9: 4711058-4711058 on Build GRCh38
rs974471773 Chr.9: 4711017-4711017 on Build GRCh38
rs553748784 Chr.9: 4711000-4711000 on Build GRCh38
rs1033326090 Chr.9: 4711233-4711233 on Build GRCh38
rs551382184 Chr.9: 4711260-4711260 on Build GRCh38
rs909031383 Chr.9: 4711303-4711303 on Build GRCh38
rs923277886 Chr.9: 4711330-4711330 on Build GRCh38
rs6915 Chr.9: 4711440-4711440 on Build GRCh38
rs933300875 Chr.9: 4711340-4711340 on Build GRCh38
rs967801108 Chr.9: 4711221-4711221 on Build GRCh38
rs7839 Chr.9: 4711224-4711224 on Build GRCh38
rs1047703003 Chr.9: 4711157-4711157 on Build GRCh38
rs998193484 Chr.9: 4711132-4711132 on Build GRCh38
rs948082138 Chr.9: 4711101-4711101 on Build GRCh38
rs960930977 Chr.9: 4710995-4710995 on Build GRCh38
rs929123588 Chr.9: 4711028-4711028 on Build GRCh38
rs892086348 Chr.9: 4711344-4711344 on Build GRCh38
rs1011661821 Chr.9: 4711191-4711191 on Build GRCh38
rs964510335 Chr.9: 4711448-4711448 on Build GRCh38
rs371016175 Chr.9: 4711367-4711367 on Build GRCh38
rs916595139 Chr.9: 4711098-4711098 on Build GRCh38
rs1047899 Chr.9: 4711218-4711218 on Build GRCh38
rs776345046 Chr.9: 4711406-4711406 on Build GRCh38
rs867400208 Chr.9: 4711294-4711294 on Build GRCh38
rs974799311 Chr.9: 4711325-4711325 on Build GRCh38
rs978362104 Chr.9: 4711041-4711041 on Build GRCh38
rs1022042794 Chr.9: 4711018-4711018 on Build GRCh38
rs147050579 Chr.9: 4711001-4711001 on Build GRCh38
rs778347508 Chr.9: 4711423-4711423 on Build GRCh38
rs139372796 Chr.9: 4711358-4711358 on Build GRCh38
rs992495609 Chr.9: 4711239-4711239 on Build GRCh38
rs773117120 Chr.9: 4711029-4711029 on Build GRCh38
rs746071268 Chr.9: 4711415-4711416 on Build GRCh38
rs140932221 Chr.9: 4711095-4711095 on Build GRCh38

View Full Product Details