Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5469058-5469605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD274
Entrez Gene ID
29126
Gene Name
CD274 molecule
Gene Aliases
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1
UniGene
Hs.521989
Gene Chromosome Location
Chr.9: 5450503-5470567 on Build GRCh38
Transcripts
NM_001267706.1 NM_014143.3 NR_052005.1

SNP Detail

SNP ID Chromosome Location
rs943676932 Chr.9: 5469166-5469166 on Build GRCh38
rs769054238 Chr.9: 5469363-5469363 on Build GRCh38
rs1026801874 Chr.9: 5469367-5469367 on Build GRCh38
rs1044415425 Chr.9: 5469497-5469497 on Build GRCh38
rs765308977 Chr.9: 5469093-5469093 on Build GRCh38
rs967443755 Chr.9: 5469241-5469241 on Build GRCh38
rs920961038 Chr.9: 5469454-5469454 on Build GRCh38
rs1329944 Chr.9: 5469504-5469504 on Build GRCh38
rs991258478 Chr.9: 5469333-5469333 on Build GRCh38
rs975014112 Chr.9: 5469451-5469451 on Build GRCh38
rs796460414 Chr.9: 5469318-5469318 on Build GRCh38
rs998554261 Chr.9: 5469291-5469291 on Build GRCh38
rs763324924 Chr.9: 5469355-5469355 on Build GRCh38
rs899187950 Chr.9: 5469507-5469507 on Build GRCh38
rs947151735 Chr.9: 5469464-5469464 on Build GRCh38
rs982648981 Chr.9: 5469416-5469416 on Build GRCh38
rs965257255 Chr.9: 5469571-5469571 on Build GRCh38
rs915747279 Chr.9: 5469341-5469341 on Build GRCh38
rs944929924 Chr.9: 5469496-5469496 on Build GRCh38
rs1012884497 Chr.9: 5469302-5469302 on Build GRCh38
rs770179979 Chr.9: 5469279-5469279 on Build GRCh38
rs996756739 Chr.9: 5469578-5469578 on Build GRCh38
rs903461539 Chr.9: 5469520-5469520 on Build GRCh38
rs368481749 Chr.9: 5469554-5469554 on Build GRCh38
rs981758242 Chr.9: 5469398-5469398 on Build GRCh38
rs971254294 Chr.9: 5469364-5469364 on Build GRCh38
rs114579116 Chr.9: 5469424-5469424 on Build GRCh38
rs372328773 Chr.9: 5469480-5469480 on Build GRCh38
rs139324756 Chr.9: 5469581-5469581 on Build GRCh38
rs555215327 Chr.9: 5469488-5469488 on Build GRCh38
rs539361136 Chr.9: 5469181-5469181 on Build GRCh38
rs1057511459 Chr.9: 5469229-5469229 on Build GRCh38
rs564189967 Chr.9: 5469114-5469114 on Build GRCh38
rs903386934 Chr.9: 5469212-5469212 on Build GRCh38
rs912217271 Chr.9: 5469153-5469153 on Build GRCh38
rs375689198 Chr.9: 5469124-5469124 on Build GRCh38
rs908495035 Chr.9: 5469415-5469415 on Build GRCh38
rs781463850 Chr.9: 5469220-5469220 on Build GRCh38
rs1032708362 Chr.9: 5469299-5469299 on Build GRCh38
rs1027267540 Chr.9: 5469137-5469137 on Build GRCh38
rs746064465 Chr.9: 5469221-5469221 on Build GRCh38
rs1023033786 Chr.9: 5469100-5469100 on Build GRCh38
rs188880293 Chr.9: 5469092-5469092 on Build GRCh38
rs113029061 Chr.9: 5469115-5469115 on Build GRCh38
rs1004412898 Chr.9: 5469567-5469567 on Build GRCh38
rs1026789132 Chr.9: 5469550-5469550 on Build GRCh38
rs965267241 Chr.9: 5469441-5469441 on Build GRCh38
rs760168795 Chr.9: 5469472-5469472 on Build GRCh38
rs1057324862 Chr.9: 5469483-5469483 on Build GRCh38
rs775860169 Chr.9: 5469346-5469346 on Build GRCh38
rs1012936879 Chr.9: 5469098-5469098 on Build GRCh38
rs920650527 Chr.9: 5469458-5469458 on Build GRCh38
rs951320107 Chr.9: 5469371-5469371 on Build GRCh38
rs947146090 Chr.9: 5469194-5469194 on Build GRCh38
rs185359196 Chr.9: 5469467-5469467 on Build GRCh38
rs571833216 Chr.9: 5469152-5469152 on Build GRCh38
rs201324948 Chr.9: 5469351-5469351 on Build GRCh38
rs1043220447 Chr.9: 5469205-5469205 on Build GRCh38
rs951327527 Chr.9: 5469151-5469151 on Build GRCh38
rs1032656043 Chr.9: 5469535-5469535 on Build GRCh38
rs998896906 Chr.9: 5469534-5469534 on Build GRCh38
rs994918916 Chr.9: 5469540-5469540 on Build GRCh38
rs573692330 Chr.9: 5469493-5469493 on Build GRCh38
rs975066203 Chr.9: 5469172-5469172 on Build GRCh38
rs1019413774 Chr.9: 5469568-5469568 on Build GRCh38
rs182006040 Chr.9: 5469253-5469253 on Build GRCh38

View Full Product Details