Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175989182-175989685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFWD2
Entrez Gene ID
64326
Gene Name
ring finger and WD repeat domain 2
Gene Aliases
COP1, RNF200
UniGene
Hs.523744
Gene Chromosome Location
Chr.1: 175944826-176207279 on Build GRCh38
Transcripts
NM_001001740.3 NM_001286644.1 NM_022457.6 XM_005245447.2 XM_005245448.2 XM_006711487.2 XM_011509881.1 XM_017002059.1 XM_017002060.1 XM_017002061.1 XM_017002062.1 XM_017002063.1 XM_017002064.1 XM_017002065.1 XM_017002066.1 XM_017002067.1 XM_017002068.1 XM_017002069.1 XM_017002070.1 XM_017002071.1 XM_017002072.1 XM_017002073.1 XM_017002074.1 XM_017002075.1 XM_017002076.1 XM_017002077.1 XM_017002078.1 XM_017002079.1 XM_017002080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7281765 c.1825A>G p.S609G Chr.1: 175989384-175989384 on Build GRCh38
6915075 c.1757G>C p.R586P Chr.1: 175989452-175989452 on Build GRCh38
3478741 c.1756C>T p.R586C Chr.1: 175989453-175989453 on Build GRCh38
676959 c.1740C>G p.V580V Chr.1: 175989469-175989469 on Build GRCh38
6923842 c.1796G>A p.R599H Chr.1: 175989413-175989413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763302837 Chr.1: 175989510-175989511 on Build GRCh38
rs763471827 Chr.1: 175989311-175989311 on Build GRCh38
rs769230149 Chr.1: 175989435-175989435 on Build GRCh38
rs1051473789 Chr.1: 175989618-175989618 on Build GRCh38
rs773722602 Chr.1: 175989343-175989344 on Build GRCh38
rs201204985 Chr.1: 175989359-175989359 on Build GRCh38
rs772783961 Chr.1: 175989314-175989314 on Build GRCh38
rs777842149 Chr.1: 175989397-175989397 on Build GRCh38
rs750646229 Chr.1: 175989530-175989530 on Build GRCh38
rs764011836 Chr.1: 175989505-175989505 on Build GRCh38
rs536134887 Chr.1: 175989554-175989554 on Build GRCh38
rs144811828 Chr.1: 175989461-175989461 on Build GRCh38
rs575937778 Chr.1: 175989571-175989571 on Build GRCh38
rs950782683 Chr.1: 175989219-175989219 on Build GRCh38
rs561356705 Chr.1: 175989278-175989278 on Build GRCh38
rs202000325 Chr.1: 175989491-175989491 on Build GRCh38
rs78482766 Chr.1: 175989232-175989232 on Build GRCh38
rs781236782 Chr.1: 175989415-175989415 on Build GRCh38
rs769816033 Chr.1: 175989341-175989343 on Build GRCh38
rs887007660 Chr.1: 175989608-175989608 on Build GRCh38
rs956604797 Chr.1: 175989569-175989569 on Build GRCh38
rs749453236 Chr.1: 175989238-175989238 on Build GRCh38
rs956635154 Chr.1: 175989222-175989223 on Build GRCh38
rs770747539 Chr.1: 175989475-175989475 on Build GRCh38
rs760419611 Chr.1: 175989315-175989315 on Build GRCh38
rs112827369 Chr.1: 175989313-175989313 on Build GRCh38
rs940317522 Chr.1: 175989446-175989446 on Build GRCh38
rs1056587107 Chr.1: 175989380-175989380 on Build GRCh38
rs990962633 Chr.1: 175989349-175989349 on Build GRCh38
rs757539169 Chr.1: 175989406-175989406 on Build GRCh38
rs768862554 Chr.1: 175989259-175989259 on Build GRCh38
rs200323555 Chr.1: 175989409-175989409 on Build GRCh38
rs935033726 Chr.1: 175989464-175989464 on Build GRCh38
rs762433396 Chr.1: 175989376-175989376 on Build GRCh38
rs139905169 Chr.1: 175989426-175989426 on Build GRCh38
rs372372343 Chr.1: 175989338-175989338 on Build GRCh38
rs759220584 Chr.1: 175989346-175989346 on Build GRCh38
rs765032631 Chr.1: 175989347-175989347 on Build GRCh38
rs780429584 Chr.1: 175989433-175989433 on Build GRCh38
rs768106659 Chr.1: 175989463-175989463 on Build GRCh38
rs753734710 Chr.1: 175989343-175989343 on Build GRCh38
rs1004916649 Chr.1: 175989642-175989642 on Build GRCh38
rs762932926 Chr.1: 175989502-175989502 on Build GRCh38
rs181761493 Chr.1: 175989566-175989566 on Build GRCh38
rs373279693 Chr.1: 175989424-175989424 on Build GRCh38
rs939498680 Chr.1: 175989373-175989373 on Build GRCh38
rs376474310 Chr.1: 175989583-175989583 on Build GRCh38
rs558185084 Chr.1: 175989607-175989607 on Build GRCh38
rs537174310 Chr.1: 175989243-175989243 on Build GRCh38
rs770082017 Chr.1: 175989427-175989427 on Build GRCh38
rs368607299 Chr.1: 175989327-175989327 on Build GRCh38
rs148861358 Chr.1: 175989310-175989310 on Build GRCh38
rs915009269 Chr.1: 175989492-175989492 on Build GRCh38
rs36053042 Chr.1: 175989516-175989516 on Build GRCh38
rs988129099 Chr.1: 175989223-175989223 on Build GRCh38
rs749820946 Chr.1: 175989434-175989434 on Build GRCh38
rs1039558392 Chr.1: 175989544-175989544 on Build GRCh38
rs751623215 Chr.1: 175989403-175989403 on Build GRCh38
rs201071402 Chr.1: 175989402-175989402 on Build GRCh38
rs752477506 Chr.1: 175989501-175989501 on Build GRCh38
rs988017475 Chr.1: 175989592-175989592 on Build GRCh38
rs111531372 Chr.1: 175989442-175989442 on Build GRCh38
rs766118243 Chr.1: 175989340-175989340 on Build GRCh38
rs751674525 Chr.1: 175989514-175989514 on Build GRCh38
rs915284390 Chr.1: 175989258-175989258 on Build GRCh38
rs764943754 Chr.1: 175989497-175989497 on Build GRCh38
rs776169758 Chr.1: 175989484-175989484 on Build GRCh38

View Full Product Details