Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178283441-178283959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASAL2
Entrez Gene ID
9462
Gene Name
RAS protein activator like 2
Gene Aliases
NGAP
UniGene
Hs.496139
Gene Chromosome Location
Chr.1: 178093729-178479514 on Build GRCh38
Transcripts
NM_170692.2 XM_011510166.2 XM_011510167.2 XM_017002849.1 XM_017002850.1 XM_017002851.1 XM_017002852.1 XM_017002853.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
900115 c.220C>T p.R74W Chr.1: 178283635-178283635 on Build GRCh38
3418391 c.230G>A p.C77Y Chr.1: 178283645-178283645 on Build GRCh38
6763117 c.297C>G p.D99E Chr.1: 178283658-178283658 on Build GRCh38
3802979 c.276G>T p.E92D Chr.1: 178283691-178283691 on Build GRCh38
677914 c.213G>C p.T71T Chr.1: 178283628-178283628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748045048 Chr.1: 178283688-178283688 on Build GRCh38
rs879478185 Chr.1: 178283781-178283781 on Build GRCh38
rs755664680 Chr.1: 178283665-178283665 on Build GRCh38
rs775258572 Chr.1: 178283521-178283521 on Build GRCh38
rs969580831 Chr.1: 178283543-178283543 on Build GRCh38
rs56181616 Chr.1: 178283762-178283762 on Build GRCh38
rs1042477100 Chr.1: 178283848-178283848 on Build GRCh38
rs760935063 Chr.1: 178283590-178283591 on Build GRCh38
rs780569696 Chr.1: 178283606-178283606 on Build GRCh38
rs765706747 Chr.1: 178283635-178283635 on Build GRCh38
rs901832630 Chr.1: 178283823-178283823 on Build GRCh38
rs769708256 Chr.1: 178283610-178283610 on Build GRCh38
rs765694364 Chr.1: 178283814-178283814 on Build GRCh38
rs949071143 Chr.1: 178283852-178283852 on Build GRCh38
rs995661097 Chr.1: 178283890-178283890 on Build GRCh38
rs756852015 Chr.1: 178283657-178283657 on Build GRCh38
rs748996332 Chr.1: 178283483-178283483 on Build GRCh38
rs774022113 Chr.1: 178283683-178283683 on Build GRCh38
rs779930574 Chr.1: 178283671-178283671 on Build GRCh38
rs764282138 Chr.1: 178283535-178283535 on Build GRCh38
rs1043962267 Chr.1: 178283754-178283754 on Build GRCh38
rs775459193 Chr.1: 178283613-178283613 on Build GRCh38
rs371933668 Chr.1: 178283658-178283658 on Build GRCh38
rs368618531 Chr.1: 178283587-178283587 on Build GRCh38
rs990850759 Chr.1: 178283607-178283607 on Build GRCh38
rs904969868 Chr.1: 178283879-178283879 on Build GRCh38
rs767568879 Chr.1: 178283620-178283620 on Build GRCh38
rs563642381 Chr.1: 178283573-178283573 on Build GRCh38
rs755433366 Chr.1: 178283581-178283581 on Build GRCh38
rs745447952 Chr.1: 178283608-178283608 on Build GRCh38
rs753149594 Chr.1: 178283905-178283905 on Build GRCh38
rs896145257 Chr.1: 178283840-178283840 on Build GRCh38
rs546030844 Chr.1: 178283588-178283588 on Build GRCh38
rs767518304 Chr.1: 178283571-178283571 on Build GRCh38
rs201094626 Chr.1: 178283701-178283701 on Build GRCh38
rs751823505 Chr.1: 178283545-178283545 on Build GRCh38
rs780763409 Chr.1: 178283577-178283577 on Build GRCh38
rs935236752 Chr.1: 178283816-178283816 on Build GRCh38
rs374211213 Chr.1: 178283627-178283627 on Build GRCh38
rs1052066045 Chr.1: 178283835-178283835 on Build GRCh38
rs908846307 Chr.1: 178283853-178283853 on Build GRCh38
rs1044802635 Chr.1: 178283876-178283876 on Build GRCh38
rs376848012 Chr.1: 178283664-178283664 on Build GRCh38
rs774681852 Chr.1: 178283515-178283516 on Build GRCh38
rs762797200 Chr.1: 178283534-178283534 on Build GRCh38
rs764124482 Chr.1: 178283620-178283622 on Build GRCh38
rs148830015 Chr.1: 178283676-178283676 on Build GRCh38
rs369135291 Chr.1: 178283822-178283822 on Build GRCh38
rs921390383 Chr.1: 178283698-178283698 on Build GRCh38
rs577021348 Chr.1: 178283580-178283580 on Build GRCh38
rs773521862 Chr.1: 178283710-178283710 on Build GRCh38
rs751153209 Chr.1: 178283636-178283636 on Build GRCh38
rs368634100 Chr.1: 178283628-178283628 on Build GRCh38
rs138728063 Chr.1: 178283906-178283906 on Build GRCh38
rs374450439 Chr.1: 178283513-178283513 on Build GRCh38
rs199969518 Chr.1: 178283616-178283616 on Build GRCh38
rs182619464 Chr.1: 178283615-178283615 on Build GRCh38
rs561483301 Chr.1: 178283891-178283891 on Build GRCh38
rs369397594 Chr.1: 178283559-178283559 on Build GRCh38
rs878945397 Chr.1: 178283498-178283498 on Build GRCh38
rs768383867 Chr.1: 178283679-178283679 on Build GRCh38
rs371638409 Chr.1: 178283617-178283617 on Build GRCh38

View Full Product Details