Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 88544292-88544794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXNL2
Entrez Gene ID
158046
Gene Name
nucleoredoxin-like 2
Gene Aliases
C9orf121, RDCVF2
UniGene
Hs.734507
Gene Chromosome Location
Chr.9: 88535088-88584510 on Build GRCh38
Transcripts
NM_001161625.1 NM_145283.2 XM_005251727.3 XM_011518275.2 XM_011518276.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1598557 c.319T>A p.Y107N Chr.9: 88544395-88544395 on Build GRCh38
5003229 c.381C>A p.N127K Chr.9: 88544457-88544457 on Build GRCh38
456209 c.333C>G p.A111A Chr.9: 88544409-88544409 on Build GRCh38
1737767 c.440C>T p.A147V Chr.9: 88544516-88544516 on Build GRCh38
7288898 c.319T>C p.Y107H Chr.9: 88544395-88544395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115978059 Chr.9: 88544703-88544703 on Build GRCh38
rs761046671 Chr.9: 88544511-88544511 on Build GRCh38
rs201353888 Chr.9: 88544426-88544426 on Build GRCh38
rs908825994 Chr.9: 88544339-88544339 on Build GRCh38
rs7032252 Chr.9: 88544679-88544679 on Build GRCh38
rs770769943 Chr.9: 88544483-88544483 on Build GRCh38
rs775868982 Chr.9: 88544506-88544506 on Build GRCh38
rs974296953 Chr.9: 88544598-88544598 on Build GRCh38
rs908529243 Chr.9: 88544581-88544581 on Build GRCh38
rs751445572 Chr.9: 88544500-88544500 on Build GRCh38
rs533646670 Chr.9: 88544516-88544516 on Build GRCh38
rs992590766 Chr.9: 88544576-88544576 on Build GRCh38
rs553411501 Chr.9: 88544541-88544541 on Build GRCh38
rs939314149 Chr.9: 88544323-88544323 on Build GRCh38
rs754077421 Chr.9: 88544392-88544392 on Build GRCh38
rs539255598 Chr.9: 88544596-88544596 on Build GRCh38
rs866253863 Chr.9: 88544697-88544697 on Build GRCh38
rs920092510 Chr.9: 88544599-88544599 on Build GRCh38
rs943177197 Chr.9: 88544714-88544714 on Build GRCh38
rs570667217 Chr.9: 88544595-88544595 on Build GRCh38
rs529601721 Chr.9: 88544338-88544338 on Build GRCh38
rs1008343850 Chr.9: 88544434-88544434 on Build GRCh38
rs150542040 Chr.9: 88544542-88544542 on Build GRCh38
rs1034212619 Chr.9: 88544521-88544521 on Build GRCh38
rs753080590 Chr.9: 88544340-88544340 on Build GRCh38
rs991626951 Chr.9: 88544355-88544355 on Build GRCh38
rs763733033 Chr.9: 88544602-88544602 on Build GRCh38
rs1038575007 Chr.9: 88544370-88544370 on Build GRCh38
rs371921367 Chr.9: 88544492-88544492 on Build GRCh38
rs745989226 Chr.9: 88544413-88544413 on Build GRCh38
rs376120424 Chr.9: 88544517-88544517 on Build GRCh38
rs749899269 Chr.9: 88544464-88544464 on Build GRCh38
rs754269531 Chr.9: 88544520-88544520 on Build GRCh38
rs778961052 Chr.9: 88544401-88544401 on Build GRCh38
rs748931914 Chr.9: 88544479-88544479 on Build GRCh38
rs371045307 Chr.9: 88544362-88544362 on Build GRCh38
rs553328100 Chr.9: 88544462-88544462 on Build GRCh38
rs1053227423 Chr.9: 88544706-88544706 on Build GRCh38
rs887791863 Chr.9: 88544566-88544566 on Build GRCh38
rs531243605 Chr.9: 88544448-88544448 on Build GRCh38
rs781013337 Chr.9: 88544428-88544428 on Build GRCh38
rs187905666 Chr.9: 88544646-88544646 on Build GRCh38
rs867525547 Chr.9: 88544716-88544716 on Build GRCh38
rs891433774 Chr.9: 88544751-88544751 on Build GRCh38
rs200418094 Chr.9: 88544487-88544487 on Build GRCh38
rs62000369 Chr.9: 88544477-88544477 on Build GRCh38
rs552871536 Chr.9: 88544319-88544319 on Build GRCh38
rs959559955 Chr.9: 88544342-88544342 on Build GRCh38
rs761992428 Chr.9: 88544526-88544526 on Build GRCh38
rs992371798 Chr.9: 88544619-88544619 on Build GRCh38
rs867172593 Chr.9: 88544341-88544341 on Build GRCh38
rs935881506 Chr.9: 88544464-88544464 on Build GRCh38
rs569444287 Chr.9: 88544400-88544400 on Build GRCh38
rs199693382 Chr.9: 88544356-88544356 on Build GRCh38
rs376257686 Chr.9: 88544451-88544451 on Build GRCh38
rs193193177 Chr.9: 88544752-88544752 on Build GRCh38
rs910511586 Chr.9: 88544707-88544707 on Build GRCh38
rs932406446 Chr.9: 88544406-88544406 on Build GRCh38

View Full Product Details