Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 90610870-90611383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIRAS2
Entrez Gene ID
54769
Gene Name
DIRAS family GTPase 2
Gene Aliases
Di-Ras2
UniGene
Hs.165636
Gene Chromosome Location
Chr.9: 90609832-90643105 on Build GRCh38
Transcripts
NM_017594.4

SNP Detail

SNP ID Chromosome Location
rs906192640 Chr.9: 90611130-90611130 on Build GRCh38
rs1023773598 Chr.9: 90611309-90611309 on Build GRCh38
rs914557393 Chr.9: 90611146-90611147 on Build GRCh38
rs947968689 Chr.9: 90611102-90611102 on Build GRCh38
rs1017600190 Chr.9: 90610933-90610933 on Build GRCh38
rs776974393 Chr.9: 90611193-90611193 on Build GRCh38
rs142447288 Chr.9: 90611192-90611192 on Build GRCh38
rs915058941 Chr.9: 90611319-90611319 on Build GRCh38
rs934081653 Chr.9: 90611010-90611010 on Build GRCh38
rs1044977378 Chr.9: 90611331-90611331 on Build GRCh38
rs957409145 Chr.9: 90611278-90611278 on Build GRCh38
rs984370175 Chr.9: 90610897-90610897 on Build GRCh38
rs111351389 Chr.9: 90610951-90610951 on Build GRCh38
rs981728267 Chr.9: 90611069-90611069 on Build GRCh38
rs539671486 Chr.9: 90611044-90611044 on Build GRCh38
rs16906711 Chr.9: 90610996-90610996 on Build GRCh38
rs570695526 Chr.9: 90611028-90611028 on Build GRCh38
rs574148329 Chr.9: 90611108-90611108 on Build GRCh38
rs572490210 Chr.9: 90611277-90611277 on Build GRCh38
rs563054204 Chr.9: 90611222-90611222 on Build GRCh38
rs997085150 Chr.9: 90611203-90611203 on Build GRCh38
rs997476556 Chr.9: 90610936-90610936 on Build GRCh38
rs947991230 Chr.9: 90611328-90611328 on Build GRCh38
rs540196495 Chr.9: 90611075-90611075 on Build GRCh38
rs576499035 Chr.9: 90611174-90611174 on Build GRCh38
rs1037354828 Chr.9: 90611276-90611276 on Build GRCh38
rs990512804 Chr.9: 90611030-90611030 on Build GRCh38
rs955343631 Chr.9: 90611215-90611215 on Build GRCh38
rs977543168 Chr.9: 90611229-90611229 on Build GRCh38
rs947489581 Chr.9: 90611153-90611153 on Build GRCh38
rs970256440 Chr.9: 90611318-90611318 on Build GRCh38
rs964656269 Chr.9: 90611194-90611194 on Build GRCh38
rs556614755 Chr.9: 90611159-90611159 on Build GRCh38
rs935150475 Chr.9: 90611042-90611042 on Build GRCh38
rs975825610 Chr.9: 90610986-90610986 on Build GRCh38
rs1004930013 Chr.9: 90610953-90610953 on Build GRCh38
rs928842404 Chr.9: 90611070-90611070 on Build GRCh38
rs748125175 Chr.9: 90611115-90611115 on Build GRCh38
rs1004744570 Chr.9: 90611269-90611269 on Build GRCh38
rs541320226 Chr.9: 90611294-90611294 on Build GRCh38
rs769630149 Chr.9: 90611017-90611017 on Build GRCh38
rs759664927 Chr.9: 90611357-90611357 on Build GRCh38
rs939016283 Chr.9: 90611359-90611359 on Build GRCh38
rs181380913 Chr.9: 90611214-90611214 on Build GRCh38
rs1003277495 Chr.9: 90611158-90611158 on Build GRCh38
rs927668956 Chr.9: 90611343-90611343 on Build GRCh38

View Full Product Details