Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93319021-93319523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf129
Entrez Gene ID
445577
Gene Name
chromosome 9 open reading frame 129
Gene Aliases
bA165J3.3
UniGene
Hs.575636
Gene Chromosome Location
Chr.9: 93318199-93346414 on Build GRCh38
Transcripts
NM_001098808.1 XM_006717117.3 NM_001282394.1 NM_006648.3 XM_005252137.3 XM_005252140.2 XM_005252141.2 XM_011518926.2 XM_011518927.2 XM_011518928.2 XM_011518929.2 XM_011518930.2 XM_011518931.2 XM_011518932.2 XM_011518935.2 XM_011518936.2 XM_011518937.2 XM_011518939.2 XM_011518940.2 XM_017015045.1 XM_017015046.1 XM_017015047.1 XM_017015048.1 XM_017015049.1 XM_017015050.1 XM_017015051.1 XM_017015052.1 XM_017015053.1 XM_017015054.1 XM_017015055.1

Gene Symbol
WNK2
Entrez Gene ID
65268
Gene Name
WNK lysine deficient protein kinase 2
Gene Aliases
NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43
UniGene
Hs.654856
Gene Chromosome Location
Chr.9: 93184156-93327581 on Build GRCh38
Transcripts
NM_001098808.1 XM_006717117.3 NM_001282394.1 NM_006648.3 XM_005252137.3 XM_005252140.2 XM_005252141.2 XM_011518926.2 XM_011518927.2 XM_011518928.2 XM_011518929.2 XM_011518930.2 XM_011518931.2 XM_011518932.2 XM_011518935.2 XM_011518936.2 XM_011518937.2 XM_011518939.2 XM_011518940.2 XM_017015045.1 XM_017015046.1 XM_017015047.1 XM_017015048.1 XM_017015049.1 XM_017015050.1 XM_017015051.1 XM_017015052.1 XM_017015053.1 XM_017015054.1 XM_017015055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
117667 c.370A>C p.T124P Chr.9: 93319170-93319170 on Build GRCh38
6473342 c.416G>A p.R139K Chr.9: 93319124-93319124 on Build GRCh38
6473343 c.389G>A p.R130H Chr.9: 93319151-93319151 on Build GRCh38
4589044 c.377_378delAT p.Y126fs*7 Chr.9: 93319162-93319163 on Build GRCh38
290238 c.388C>T p.R130C Chr.9: 93319152-93319152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929388262 Chr.9: 93319394-93319394 on Build GRCh38
rs746205595 Chr.9: 93319143-93319143 on Build GRCh38
rs117465023 Chr.9: 93319151-93319151 on Build GRCh38
rs1007745000 Chr.9: 93319230-93319230 on Build GRCh38
rs959824895 Chr.9: 93319361-93319361 on Build GRCh38
rs755658521 Chr.9: 93319087-93319087 on Build GRCh38
rs376337249 Chr.9: 93319122-93319122 on Build GRCh38
rs766297514 Chr.9: 93319185-93319185 on Build GRCh38
rs753321777 Chr.9: 93319223-93319223 on Build GRCh38
rs577987563 Chr.9: 93319294-93319294 on Build GRCh38
rs751314524 Chr.9: 93319070-93319070 on Build GRCh38
rs971976077 Chr.9: 93319344-93319344 on Build GRCh38
rs373255290 Chr.9: 93319158-93319158 on Build GRCh38
rs540163220 Chr.9: 93319162-93319163 on Build GRCh38
rs762815114 Chr.9: 93319169-93319169 on Build GRCh38
rs1005886892 Chr.9: 93319242-93319242 on Build GRCh38
rs774582885 Chr.9: 93319132-93319132 on Build GRCh38
rs982027574 Chr.9: 93319059-93319059 on Build GRCh38
rs947499657 Chr.9: 93319422-93319422 on Build GRCh38
rs915994586 Chr.9: 93319372-93319372 on Build GRCh38
rs555576938 Chr.9: 93319245-93319245 on Build GRCh38
rs777331438 Chr.9: 93319100-93319100 on Build GRCh38
rs758356145 Chr.9: 93319060-93319060 on Build GRCh38
rs754654379 Chr.9: 93319074-93319074 on Build GRCh38
rs780771440 Chr.9: 93319080-93319080 on Build GRCh38
rs934960205 Chr.9: 93319063-93319063 on Build GRCh38
rs777158722 Chr.9: 93319210-93319210 on Build GRCh38
rs897825283 Chr.9: 93319156-93319156 on Build GRCh38
rs199832464 Chr.9: 93319085-93319085 on Build GRCh38
rs958202074 Chr.9: 93319237-93319237 on Build GRCh38
rs532336280 Chr.9: 93319149-93319149 on Build GRCh38
rs751331371 Chr.9: 93319190-93319190 on Build GRCh38
rs114534350 Chr.9: 93319268-93319268 on Build GRCh38
rs770288896 Chr.9: 93319450-93319450 on Build GRCh38
rs149476600 Chr.9: 93319371-93319371 on Build GRCh38
rs1037768891 Chr.9: 93319152-93319152 on Build GRCh38
rs201220086 Chr.9: 93319163-93319163 on Build GRCh38
rs767265403 Chr.9: 93319203-93319203 on Build GRCh38
rs1041260821 Chr.9: 93319451-93319451 on Build GRCh38
rs749818439 Chr.9: 93319104-93319104 on Build GRCh38
rs201504435 Chr.9: 93319204-93319204 on Build GRCh38
rs373508900 Chr.9: 93319084-93319084 on Build GRCh38
rs759371907 Chr.9: 93319193-93319193 on Build GRCh38
rs981813185 Chr.9: 93319342-93319342 on Build GRCh38
rs1020662411 Chr.9: 93319308-93319308 on Build GRCh38
rs781046397 Chr.9: 93319274-93319274 on Build GRCh38
rs888903412 Chr.9: 93319167-93319167 on Build GRCh38
rs369036642 Chr.9: 93319052-93319052 on Build GRCh38
rs139100598 Chr.9: 93319284-93319284 on Build GRCh38
rs80221697 Chr.9: 93319333-93319333 on Build GRCh38
rs1036032919 Chr.9: 93319357-93319357 on Build GRCh38
rs1007777522 Chr.9: 93319440-93319440 on Build GRCh38
rs374384034 Chr.9: 93319352-93319353 on Build GRCh38
rs545053802 Chr.9: 93319312-93319312 on Build GRCh38
rs932799040 Chr.9: 93319470-93319470 on Build GRCh38
rs567303457 Chr.9: 93319252-93319252 on Build GRCh38
rs531783114 Chr.9: 93319484-93319484 on Build GRCh38
rs768736191 Chr.9: 93319135-93319135 on Build GRCh38
rs554368379 Chr.9: 93319109-93319109 on Build GRCh38
rs542319242 Chr.9: 93319430-93319430 on Build GRCh38
rs1018216328 Chr.9: 93319235-93319235 on Build GRCh38
rs979159993 Chr.9: 93319429-93319429 on Build GRCh38
rs1051736530 Chr.9: 93319407-93319407 on Build GRCh38
rs571326914 Chr.9: 93319102-93319102 on Build GRCh38
rs934676409 Chr.9: 93319439-93319439 on Build GRCh38
rs561533623 Chr.9: 93319456-93319456 on Build GRCh38
rs766147613 Chr.9: 93319061-93319061 on Build GRCh38
rs888275042 Chr.9: 93319134-93319134 on Build GRCh38

View Full Product Details