Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96928663-96929161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUTM2G
Entrez Gene ID
441457
Gene Name
NUT family member 2G
Gene Aliases
FAM22G, NUTMG
UniGene
Hs.652383
Gene Chromosome Location
Chr.9: 96929004-96942290 on Build GRCh38
Transcripts
NM_001045477.2 NM_001170741.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
403103 c.16+2T>A p.? Chr.9: 96929042-96929042 on Build GRCh38
3659890 c.4G>A p.A2T Chr.9: 96929028-96929028 on Build GRCh38
3659889 c.4G>A p.A2T Chr.9: 96929028-96929028 on Build GRCh38
3780245 c.14G>C p.G5A Chr.9: 96929038-96929038 on Build GRCh38
3659891 c.13G>A p.G5R Chr.9: 96929037-96929037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550270856 Chr.9: 96928740-96928740 on Build GRCh38
rs757953188 Chr.9: 96929021-96929021 on Build GRCh38
rs777073988 Chr.9: 96929084-96929084 on Build GRCh38
rs961203810 Chr.9: 96928994-96928994 on Build GRCh38
rs532348533 Chr.9: 96928795-96928795 on Build GRCh38
rs45584840 Chr.9: 96928839-96928839 on Build GRCh38
rs751301001 Chr.9: 96929040-96929040 on Build GRCh38
rs775687448 Chr.9: 96928974-96928974 on Build GRCh38
rs779821104 Chr.9: 96929027-96929027 on Build GRCh38
rs754837261 Chr.9: 96929042-96929042 on Build GRCh38
rs922586612 Chr.9: 96929118-96929118 on Build GRCh38
rs373790984 Chr.9: 96928831-96928831 on Build GRCh38
rs536530938 Chr.9: 96929019-96929019 on Build GRCh38
rs906738720 Chr.9: 96928806-96928806 on Build GRCh38
rs2479277 Chr.9: 96929069-96929069 on Build GRCh38
rs201497106 Chr.9: 96928983-96928983 on Build GRCh38
rs771086629 Chr.9: 96929050-96929050 on Build GRCh38
rs754347267 Chr.9: 96928991-96928991 on Build GRCh38
rs764745652 Chr.9: 96929010-96929010 on Build GRCh38
rs569647340 Chr.9: 96929075-96929075 on Build GRCh38
rs566028066 Chr.9: 96928935-96928935 on Build GRCh38
rs1003585066 Chr.9: 96928819-96928819 on Build GRCh38
rs548239655 Chr.9: 96929043-96929043 on Build GRCh38
rs959510693 Chr.9: 96928868-96928868 on Build GRCh38
rs1016948725 Chr.9: 96928969-96928969 on Build GRCh38
rs778837091 Chr.9: 96929052-96929052 on Build GRCh38
rs756715957 Chr.9: 96929004-96929004 on Build GRCh38
rs762281305 Chr.9: 96929086-96929086 on Build GRCh38
rs571604828 Chr.9: 96928783-96928783 on Build GRCh38
rs1034086724 Chr.9: 96928823-96928823 on Build GRCh38
rs984150897 Chr.9: 96928931-96928931 on Build GRCh38
rs531573455 Chr.9: 96928719-96928719 on Build GRCh38
rs972502960 Chr.9: 96929113-96929113 on Build GRCh38
rs761157246 Chr.9: 96928982-96928982 on Build GRCh38
rs374101719 Chr.9: 96929061-96929061 on Build GRCh38
rs45530231 Chr.9: 96928850-96928850 on Build GRCh38
rs746000877 Chr.9: 96929059-96929059 on Build GRCh38
rs769038838 Chr.9: 96929079-96929079 on Build GRCh38

View Full Product Details