Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97662804-97663331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCBP1
Entrez Gene ID
4686
Gene Name
nuclear cap binding protein subunit 1
Gene Aliases
CBP80, NCBP, Sto1
UniGene
Hs.595669
Gene Chromosome Location
Chr.9: 97633423-97673748 on Build GRCh38
Transcripts
NM_002486.4 XM_017014735.1 XM_017014736.1 XM_017014737.1 XM_017014738.1 XM_006717278.1

Gene Symbol
XPA
Entrez Gene ID
7507
Gene Name
XPA, DNA damage recognition and repair factor
Gene Aliases
XP1, XPAC
UniGene
Hs.654364
Gene Chromosome Location
Chr.9: 97654398-97697409 on Build GRCh38
Transcripts
NM_002486.4 XM_017014735.1 XM_017014736.1 XM_017014737.1 XM_017014738.1 XM_006717278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1241472 c.1794A>C p.P598P Chr.9: 97663044-97663044 on Build GRCh38
3902440 c.1709A>G p.H570R Chr.9: 97662959-97662959 on Build GRCh38
4933023 c.1788C>G p.N596K Chr.9: 97663038-97663038 on Build GRCh38
5027106 c.1741G>A p.E581K Chr.9: 97662991-97662991 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs946804909 Chr.9: 97663272-97663272 on Build GRCh38
rs751269308 Chr.9: 97662934-97662934 on Build GRCh38
rs751912193 Chr.9: 97663073-97663074 on Build GRCh38
rs1024393365 Chr.9: 97663095-97663095 on Build GRCh38
rs773658380 Chr.9: 97663018-97663018 on Build GRCh38
rs759896177 Chr.9: 97662923-97662926 on Build GRCh38
rs372207879 Chr.9: 97662942-97662942 on Build GRCh38
rs116865109 Chr.9: 97662837-97662837 on Build GRCh38
rs759638692 Chr.9: 97663130-97663130 on Build GRCh38
rs775656113 Chr.9: 97662981-97662981 on Build GRCh38
rs754636371 Chr.9: 97662938-97662938 on Build GRCh38
rs1044100272 Chr.9: 97663276-97663276 on Build GRCh38
rs117257381 Chr.9: 97663136-97663136 on Build GRCh38
rs200307901 Chr.9: 97663038-97663038 on Build GRCh38
rs762372000 Chr.9: 97662903-97662903 on Build GRCh38
rs900797208 Chr.9: 97662986-97662986 on Build GRCh38
rs376816057 Chr.9: 97662944-97662944 on Build GRCh38
rs754582942 Chr.9: 97663070-97663070 on Build GRCh38
rs767175785 Chr.9: 97663074-97663074 on Build GRCh38
rs557963365 Chr.9: 97662978-97662978 on Build GRCh38
rs749927680 Chr.9: 97662920-97662920 on Build GRCh38
rs998741369 Chr.9: 97662907-97662907 on Build GRCh38
rs576031865 Chr.9: 97662987-97662987 on Build GRCh38
rs765744243 Chr.9: 97663008-97663008 on Build GRCh38
rs564783835 Chr.9: 97663054-97663054 on Build GRCh38
rs777615947 Chr.9: 97662946-97662946 on Build GRCh38
rs904465833 Chr.9: 97662898-97662898 on Build GRCh38
rs763497901 Chr.9: 97663055-97663058 on Build GRCh38
rs745960045 Chr.9: 97662957-97662957 on Build GRCh38
rs1055317627 Chr.9: 97663297-97663297 on Build GRCh38
rs944628203 Chr.9: 97663231-97663234 on Build GRCh38
rs985940848 Chr.9: 97663157-97663157 on Build GRCh38
rs769033892 Chr.9: 97662989-97662989 on Build GRCh38
rs377241977 Chr.9: 97663100-97663100 on Build GRCh38
rs764722317 Chr.9: 97663069-97663070 on Build GRCh38
rs62560502 Chr.9: 97662939-97662939 on Build GRCh38
rs772105621 Chr.9: 97662964-97662964 on Build GRCh38
rs755933890 Chr.9: 97663084-97663084 on Build GRCh38
rs1040796632 Chr.9: 97662861-97662861 on Build GRCh38
rs866941411 Chr.9: 97663283-97663283 on Build GRCh38
rs749412376 Chr.9: 97662915-97662918 on Build GRCh38
rs187823646 Chr.9: 97663108-97663108 on Build GRCh38
rs938047233 Chr.9: 97663293-97663293 on Build GRCh38
rs775714615 Chr.9: 97662936-97662937 on Build GRCh38
rs951786782 Chr.9: 97663145-97663145 on Build GRCh38
rs757885845 Chr.9: 97662925-97662925 on Build GRCh38
rs960957100 Chr.9: 97663176-97663176 on Build GRCh38
rs550563336 Chr.9: 97663301-97663301 on Build GRCh38
rs903937280 Chr.9: 97663009-97663009 on Build GRCh38
rs762511481 Chr.9: 97663019-97663019 on Build GRCh38
rs752462064 Chr.9: 97663080-97663080 on Build GRCh38
rs776329196 Chr.9: 97663267-97663267 on Build GRCh38
rs777757200 Chr.9: 97663086-97663086 on Build GRCh38
rs975993237 Chr.9: 97663238-97663238 on Build GRCh38
rs747965974 Chr.9: 97662943-97662943 on Build GRCh38
rs779562115 Chr.9: 97662885-97662885 on Build GRCh38
rs945438639 Chr.9: 97663264-97663264 on Build GRCh38
rs913802251 Chr.9: 97663259-97663259 on Build GRCh38
rs112268381 Chr.9: 97663215-97663215 on Build GRCh38
rs927971589 Chr.9: 97663155-97663155 on Build GRCh38
rs139290785 Chr.9: 97663058-97663058 on Build GRCh38
rs972936667 Chr.9: 97663137-97663137 on Build GRCh38
rs762318862 Chr.9: 97663005-97663005 on Build GRCh38
rs765685708 Chr.9: 97662905-97662905 on Build GRCh38
rs765402687 Chr.9: 97662930-97662930 on Build GRCh38
rs386736630 Chr.9: 97663295-97663297 on Build GRCh38
rs370231054 Chr.9: 97662836-97662836 on Build GRCh38
rs990830488 Chr.9: 97663180-97663180 on Build GRCh38
rs879572632 Chr.9: 97663305-97663305 on Build GRCh38
rs777089319 Chr.9: 97662999-97662999 on Build GRCh38
rs766049902 Chr.9: 97662930-97662930 on Build GRCh38

View Full Product Details