Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98734922-98735429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS6
Entrez Gene ID
203286
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases
ANKRD14, NPHP16, PKDR1, SAMD6
UniGene
Hs.406890
Gene Chromosome Location
Chr.9: 98732009-98796542 on Build GRCh38
Transcripts
NM_173551.4 XM_005251793.4 XM_005251794.4 XM_006716998.3 XM_006716999.2 XM_011518357.2 XM_017014445.1 XM_017014446.1 XM_017014447.1

SNP Detail

SNP ID Chromosome Location
rs555566449 Chr.9: 98735170-98735170 on Build GRCh38
rs944816716 Chr.9: 98735292-98735292 on Build GRCh38
rs985102952 Chr.9: 98735093-98735093 on Build GRCh38
rs962434498 Chr.9: 98735129-98735129 on Build GRCh38
rs989017670 Chr.9: 98735009-98735009 on Build GRCh38
rs546594512 Chr.9: 98735014-98735014 on Build GRCh38
rs905844177 Chr.9: 98735277-98735277 on Build GRCh38
rs337588 Chr.9: 98735219-98735219 on Build GRCh38
rs945480087 Chr.9: 98735347-98735347 on Build GRCh38
rs1010374431 Chr.9: 98735333-98735333 on Build GRCh38
rs1027759185 Chr.9: 98735271-98735271 on Build GRCh38
rs71501875 Chr.9: 98735400-98735400 on Build GRCh38
rs146059953 Chr.9: 98735276-98735276 on Build GRCh38
rs1043712139 Chr.9: 98735394-98735394 on Build GRCh38
rs1024008648 Chr.9: 98735113-98735113 on Build GRCh38
rs745554147 Chr.9: 98735130-98735130 on Build GRCh38
rs951764700 Chr.9: 98735272-98735272 on Build GRCh38
rs753345816 Chr.9: 98734987-98734987 on Build GRCh38
rs1034285389 Chr.9: 98735022-98735022 on Build GRCh38
rs778737766 Chr.9: 98735076-98735076 on Build GRCh38
rs1045530141 Chr.9: 98735037-98735037 on Build GRCh38
rs918553505 Chr.9: 98735139-98735139 on Build GRCh38
rs756918612 Chr.9: 98735063-98735063 on Build GRCh38
rs892555543 Chr.9: 98734964-98734964 on Build GRCh38
rs554677681 Chr.9: 98734965-98734965 on Build GRCh38
rs903825184 Chr.9: 98735395-98735395 on Build GRCh38
rs538050086 Chr.9: 98735255-98735255 on Build GRCh38
rs956396184 Chr.9: 98735008-98735008 on Build GRCh38
rs117846104 Chr.9: 98735040-98735040 on Build GRCh38
rs758221815 Chr.9: 98735266-98735266 on Build GRCh38
rs1044148506 Chr.9: 98734988-98734988 on Build GRCh38
rs558466940 Chr.9: 98735264-98735264 on Build GRCh38
rs779865176 Chr.9: 98735302-98735302 on Build GRCh38
rs142793751 Chr.9: 98735069-98735069 on Build GRCh38
rs1000233155 Chr.9: 98735303-98735303 on Build GRCh38
rs910861558 Chr.9: 98735099-98735099 on Build GRCh38
rs929774451 Chr.9: 98735154-98735154 on Build GRCh38
rs1055114325 Chr.9: 98735304-98735304 on Build GRCh38
rs534445795 Chr.9: 98735307-98735307 on Build GRCh38
rs1056237869 Chr.9: 98735160-98735160 on Build GRCh38

View Full Product Details