Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 99980765-99981288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERP44
Entrez Gene ID
23071
Gene Name
endoplasmic reticulum protein 44
Gene Aliases
PDIA10, TXNDC4
UniGene
Hs.154023
Gene Chromosome Location
Chr.9: 99979179-100099052 on Build GRCh38
Transcripts
NM_015051.2

SNP Detail

SNP ID Chromosome Location
rs561538257 Chr.9: 99980834-99980834 on Build GRCh38
rs868076387 Chr.9: 99980993-99980993 on Build GRCh38
rs972216667 Chr.9: 99981186-99981186 on Build GRCh38
rs187200481 Chr.9: 99981086-99981086 on Build GRCh38
rs987478412 Chr.9: 99980818-99980818 on Build GRCh38
rs1011206631 Chr.9: 99981070-99981070 on Build GRCh38
rs1031250073 Chr.9: 99981037-99981037 on Build GRCh38
rs144013232 Chr.9: 99980828-99980828 on Build GRCh38
rs559515709 Chr.9: 99981181-99981185 on Build GRCh38
rs1029293262 Chr.9: 99981101-99981101 on Build GRCh38
rs1005835900 Chr.9: 99981079-99981079 on Build GRCh38
rs948043069 Chr.9: 99980864-99980864 on Build GRCh38
rs564010363 Chr.9: 99980957-99980957 on Build GRCh38
rs977163911 Chr.9: 99981145-99981145 on Build GRCh38
rs987447382 Chr.9: 99981115-99981115 on Build GRCh38
rs118143312 Chr.9: 99980870-99980870 on Build GRCh38
rs916766443 Chr.9: 99981188-99981188 on Build GRCh38
rs762345579 Chr.9: 99981258-99981258 on Build GRCh38
rs139791061 Chr.9: 99981207-99981207 on Build GRCh38
rs964277527 Chr.9: 99981088-99981088 on Build GRCh38
rs777436714 Chr.9: 99980879-99980881 on Build GRCh38
rs962421884 Chr.9: 99981163-99981163 on Build GRCh38
rs916177291 Chr.9: 99981259-99981259 on Build GRCh38
rs145936623 Chr.9: 99980901-99980901 on Build GRCh38
rs553889191 Chr.9: 99980799-99980799 on Build GRCh38
rs113918299 Chr.9: 99981151-99981151 on Build GRCh38
rs533089080 Chr.9: 99980969-99980969 on Build GRCh38
rs372302631 Chr.9: 99981184-99981184 on Build GRCh38
rs542804074 Chr.9: 99980830-99980830 on Build GRCh38
rs953357311 Chr.9: 99981104-99981104 on Build GRCh38
rs779281589 Chr.9: 99980877-99980878 on Build GRCh38
rs957931479 Chr.9: 99981063-99981063 on Build GRCh38
rs746028209 Chr.9: 99980966-99980966 on Build GRCh38
rs549139741 Chr.9: 99981218-99981218 on Build GRCh38
rs998121939 Chr.9: 99981089-99981093 on Build GRCh38
rs540586493 Chr.9: 99980796-99980796 on Build GRCh38
rs150975024 Chr.9: 99981166-99981169 on Build GRCh38
rs906268764 Chr.9: 99980984-99980984 on Build GRCh38
rs948281759 Chr.9: 99981189-99981189 on Build GRCh38
rs775160975 Chr.9: 99981240-99981240 on Build GRCh38
rs1036926006 Chr.9: 99980853-99980853 on Build GRCh38
rs930890366 Chr.9: 99980893-99980893 on Build GRCh38
rs1056056838 Chr.9: 99980945-99980945 on Build GRCh38
rs925819757 Chr.9: 99981127-99981127 on Build GRCh38
rs151201819 Chr.9: 99981087-99981088 on Build GRCh38
rs150630834 Chr.9: 99980818-99980819 on Build GRCh38
rs999305024 Chr.9: 99980988-99980988 on Build GRCh38
rs894771147 Chr.9: 99980952-99980952 on Build GRCh38

View Full Product Details