Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 99981251-99981763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERP44
Entrez Gene ID
23071
Gene Name
endoplasmic reticulum protein 44
Gene Aliases
PDIA10, TXNDC4
UniGene
Hs.154023
Gene Chromosome Location
Chr.9: 99979179-100099052 on Build GRCh38
Transcripts
NM_015051.2

SNP Detail

SNP ID Chromosome Location
rs80274877 Chr.9: 99981662-99981662 on Build GRCh38
rs1052197939 Chr.9: 99981357-99981357 on Build GRCh38
rs928258051 Chr.9: 99981683-99981684 on Build GRCh38
rs879848003 Chr.9: 99981688-99981693 on Build GRCh38
rs953284182 Chr.9: 99981439-99981439 on Build GRCh38
rs972525705 Chr.9: 99981537-99981537 on Build GRCh38
rs1052591205 Chr.9: 99981741-99981741 on Build GRCh38
rs957318050 Chr.9: 99981480-99981480 on Build GRCh38
rs867550407 Chr.9: 99981502-99981502 on Build GRCh38
rs1028838880 Chr.9: 99981412-99981412 on Build GRCh38
rs1010758464 Chr.9: 99981373-99981373 on Build GRCh38
rs45598240 Chr.9: 99981629-99981629 on Build GRCh38
rs529042847 Chr.9: 99981463-99981463 on Build GRCh38
rs14297 Chr.9: 99981726-99981726 on Build GRCh38
rs906236219 Chr.9: 99981322-99981322 on Build GRCh38
rs952280999 Chr.9: 99981556-99981556 on Build GRCh38
rs576149215 Chr.9: 99981605-99981605 on Build GRCh38
rs184971201 Chr.9: 99981344-99981344 on Build GRCh38
rs200765208 Chr.9: 99981720-99981720 on Build GRCh38
rs751134727 Chr.9: 99981596-99981596 on Build GRCh38
rs543093937 Chr.9: 99981554-99981554 on Build GRCh38
rs758163255 Chr.9: 99981386-99981386 on Build GRCh38
rs779549699 Chr.9: 99981551-99981551 on Build GRCh38
rs950277033 Chr.9: 99981710-99981710 on Build GRCh38
rs556264864 Chr.9: 99981568-99981568 on Build GRCh38
rs41273939 Chr.9: 99981302-99981302 on Build GRCh38
rs899670538 Chr.9: 99981325-99981328 on Build GRCh38
rs762260682 Chr.9: 99981308-99981311 on Build GRCh38
rs1018900092 Chr.9: 99981473-99981473 on Build GRCh38
rs563946568 Chr.9: 99981657-99981657 on Build GRCh38
rs962105490 Chr.9: 99981481-99981481 on Build GRCh38
rs1039798335 Chr.9: 99981733-99981733 on Build GRCh38
rs77514765 Chr.9: 99981361-99981361 on Build GRCh38
rs981723358 Chr.9: 99981712-99981712 on Build GRCh38
rs998952696 Chr.9: 99981389-99981389 on Build GRCh38
rs180930353 Chr.9: 99981325-99981325 on Build GRCh38
rs999674122 Chr.9: 99981347-99981347 on Build GRCh38
rs916105075 Chr.9: 99981704-99981704 on Build GRCh38
rs75182987 Chr.9: 99981329-99981329 on Build GRCh38
rs1027755255 Chr.9: 99981543-99981543 on Build GRCh38
rs541358485 Chr.9: 99981684-99981684 on Build GRCh38

View Full Product Details