Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 100577494-100577997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSANTD3-TMEFF1
Entrez Gene ID
100526694
Gene Name
MSANTD3-TMEFF1 readthrough
Gene Aliases
C9orf3-TMEFF1, C9orf30-TMEFF1
UniGene
Hs.598100
Gene Chromosome Location
Chr.9: 100441906-100577636 on Build GRCh38
Transcripts
NM_001198812.1 NM_003692.4

Gene Symbol
TMEFF1
Entrez Gene ID
8577
Gene Name
transmembrane protein with EGF like and two follistatin like domains 1
Gene Aliases
C9orf2, CT120.1, H7365, TR-1
UniGene
Hs.598100
Gene Chromosome Location
Chr.9: 100473238-100577636 on Build GRCh38
Transcripts
NM_001198812.1 NM_003692.4

SNP Detail

SNP ID Chromosome Location
rs73495724 Chr.9: 100577640-100577640 on Build GRCh38
rs541783853 Chr.9: 100577920-100577923 on Build GRCh38
rs1033869333 Chr.9: 100577614-100577618 on Build GRCh38
rs945296558 Chr.9: 100577864-100577864 on Build GRCh38
rs764212165 Chr.9: 100577795-100577795 on Build GRCh38
rs962453128 Chr.9: 100577749-100577749 on Build GRCh38
rs566349113 Chr.9: 100577942-100577942 on Build GRCh38
rs991832723 Chr.9: 100577820-100577820 on Build GRCh38
rs182403567 Chr.9: 100577610-100577610 on Build GRCh38
rs1226592 Chr.9: 100577665-100577665 on Build GRCh38
rs981002127 Chr.9: 100577611-100577611 on Build GRCh38
rs1049693816 Chr.9: 100577666-100577666 on Build GRCh38
rs903660572 Chr.9: 100577895-100577895 on Build GRCh38
rs905545983 Chr.9: 100577811-100577811 on Build GRCh38
rs1007626932 Chr.9: 100577562-100577562 on Build GRCh38
rs943442420 Chr.9: 100577950-100577950 on Build GRCh38
rs938980022 Chr.9: 100577806-100577806 on Build GRCh38
rs1017631033 Chr.9: 100577569-100577569 on Build GRCh38
rs917789527 Chr.9: 100577634-100577634 on Build GRCh38
rs901811158 Chr.9: 100577614-100577614 on Build GRCh38
rs1039099691 Chr.9: 100577952-100577952 on Build GRCh38
rs970093490 Chr.9: 100577584-100577584 on Build GRCh38
rs919984124 Chr.9: 100577972-100577972 on Build GRCh38
rs1001179979 Chr.9: 100577833-100577833 on Build GRCh38
rs960927064 Chr.9: 100577621-100577621 on Build GRCh38
rs1035305546 Chr.9: 100577860-100577860 on Build GRCh38
rs775366626 Chr.9: 100577583-100577583 on Build GRCh38
rs148371890 Chr.9: 100577953-100577953 on Build GRCh38
rs570266340 Chr.9: 100577667-100577667 on Build GRCh38
rs926297430 Chr.9: 100577799-100577799 on Build GRCh38
rs1052977481 Chr.9: 100577741-100577741 on Build GRCh38
rs552611378 Chr.9: 100577874-100577874 on Build GRCh38
rs554543601 Chr.9: 100577959-100577959 on Build GRCh38
rs754112234 Chr.9: 100577852-100577852 on Build GRCh38
rs891074706 Chr.9: 100577712-100577712 on Build GRCh38
rs1043556015 Chr.9: 100577523-100577523 on Build GRCh38
rs143362907 Chr.9: 100577731-100577731 on Build GRCh38
rs541010457 Chr.9: 100577594-100577594 on Build GRCh38
rs891582090 Chr.9: 100577758-100577758 on Build GRCh38
rs1027023416 Chr.9: 100577638-100577638 on Build GRCh38
rs16919211 Chr.9: 100577558-100577558 on Build GRCh38

View Full Product Details