Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 100586048-100586548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MURC
Entrez Gene ID
347273
Gene Name
muscle related coiled-coil protein
Gene Aliases
CAVIN4, cavin-4
UniGene
Hs.733374
Gene Chromosome Location
Chr.9: 100578079-100588389 on Build GRCh38
Transcripts
NM_001018116.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3763491 c.996G>A p.R332R Chr.9: 100586352-100586352 on Build GRCh38
7268540 c.1011C>T p.P337P Chr.9: 100586367-100586367 on Build GRCh38
1258326 c.1072T>C p.L358L Chr.9: 100586428-100586428 on Build GRCh38
7341491 c.882_883delGG p.D295fs*13 Chr.9: 100586238-100586239 on Build GRCh38
3652331 c.790C>T p.P264S Chr.9: 100586146-100586146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758961734 Chr.9: 100586475-100586475 on Build GRCh38
rs746462546 Chr.9: 100586071-100586091 on Build GRCh38
rs765035440 Chr.9: 100586434-100586434 on Build GRCh38
rs763489899 Chr.9: 100586491-100586491 on Build GRCh38
rs768828025 Chr.9: 100586112-100586112 on Build GRCh38
rs368039947 Chr.9: 100586465-100586465 on Build GRCh38
rs769864035 Chr.9: 100586291-100586291 on Build GRCh38
rs765728679 Chr.9: 100586131-100586131 on Build GRCh38
rs149770384 Chr.9: 100586135-100586135 on Build GRCh38
rs762951809 Chr.9: 100586439-100586439 on Build GRCh38
rs147712803 Chr.9: 100586313-100586313 on Build GRCh38
rs565207218 Chr.9: 100586199-100586199 on Build GRCh38
rs943127632 Chr.9: 100586139-100586139 on Build GRCh38
rs557045047 Chr.9: 100586136-100586136 on Build GRCh38
rs766006712 Chr.9: 100586146-100586146 on Build GRCh38
rs768357648 Chr.9: 100586407-100586407 on Build GRCh38
rs901566413 Chr.9: 100586501-100586501 on Build GRCh38
rs771579521 Chr.9: 100586504-100586504 on Build GRCh38
rs147742000 Chr.9: 100586327-100586327 on Build GRCh38
rs771495511 Chr.9: 100586246-100586246 on Build GRCh38
rs748500255 Chr.9: 100586212-100586212 on Build GRCh38
rs746449850 Chr.9: 100586405-100586405 on Build GRCh38
rs775739263 Chr.9: 100586109-100586109 on Build GRCh38
rs772113280 Chr.9: 100586092-100586092 on Build GRCh38
rs748425240 Chr.9: 100586290-100586290 on Build GRCh38
rs774281723 Chr.9: 100586513-100586513 on Build GRCh38
rs768816843 Chr.9: 100586204-100586204 on Build GRCh38
rs779613737 Chr.9: 100586371-100586371 on Build GRCh38
rs141175381 Chr.9: 100586341-100586341 on Build GRCh38
rs554874636 Chr.9: 100586079-100586079 on Build GRCh38
rs751070367 Chr.9: 100586149-100586149 on Build GRCh38
rs781317175 Chr.9: 100586289-100586289 on Build GRCh38
rs371552277 Chr.9: 100586214-100586214 on Build GRCh38
rs761606996 Chr.9: 100586367-100586367 on Build GRCh38
rs371617157 Chr.9: 100586480-100586480 on Build GRCh38
rs757506400 Chr.9: 100586463-100586463 on Build GRCh38
rs190889727 Chr.9: 100586468-100586468 on Build GRCh38
rs369823650 Chr.9: 100586329-100586329 on Build GRCh38
rs752878354 Chr.9: 100586447-100586447 on Build GRCh38
rs755935790 Chr.9: 100586171-100586171 on Build GRCh38
rs546031058 Chr.9: 100586322-100586322 on Build GRCh38
rs781273271 Chr.9: 100586085-100586085 on Build GRCh38
rs773396389 Chr.9: 100586231-100586231 on Build GRCh38
rs139714525 Chr.9: 100586111-100586111 on Build GRCh38
rs375099309 Chr.9: 100586190-100586190 on Build GRCh38
rs1045740890 Chr.9: 100586450-100586450 on Build GRCh38
rs752652787 Chr.9: 100586343-100586343 on Build GRCh38
rs146549670 Chr.9: 100586070-100586070 on Build GRCh38
rs774818167 Chr.9: 100586319-100586319 on Build GRCh38
rs767137800 Chr.9: 100586159-100586159 on Build GRCh38
rs1042001140 Chr.9: 100586511-100586511 on Build GRCh38
rs761642781 Chr.9: 100586428-100586428 on Build GRCh38
rs930468369 Chr.9: 100586427-100586427 on Build GRCh38
rs746598336 Chr.9: 100586485-100586485 on Build GRCh38
rs370376859 Chr.9: 100586257-100586257 on Build GRCh38
rs752495543 Chr.9: 100586170-100586170 on Build GRCh38
rs754679919 Chr.9: 100586152-100586152 on Build GRCh38
rs774996606 Chr.9: 100586300-100586300 on Build GRCh38
rs766169152 Chr.9: 100586445-100586445 on Build GRCh38
rs199888023 Chr.9: 100586194-100586194 on Build GRCh38
rs536624756 Chr.9: 100586128-100586128 on Build GRCh38
rs774008744 Chr.9: 100586248-100586248 on Build GRCh38
rs754091589 Chr.9: 100586456-100586456 on Build GRCh38
rs376964215 Chr.9: 100586286-100586286 on Build GRCh38
rs778284038 Chr.9: 100586295-100586295 on Build GRCh38
rs756371499 Chr.9: 100586346-100586346 on Build GRCh38
rs146893930 Chr.9: 100586328-100586328 on Build GRCh38
rs148374985 Chr.9: 100586336-100586336 on Build GRCh38
rs2780956 Chr.9: 100586352-100586352 on Build GRCh38
rs115947662 Chr.9: 100586497-100586497 on Build GRCh38
rs899257053 Chr.9: 100586124-100586124 on Build GRCh38
rs781471187 Chr.9: 100586210-100586210 on Build GRCh38
rs148950537 Chr.9: 100586269-100586269 on Build GRCh38
rs1036130331 Chr.9: 100586093-100586093 on Build GRCh38
rs375965738 Chr.9: 100586251-100586251 on Build GRCh38
rs775398666 Chr.9: 100586226-100586231 on Build GRCh38
rs775073275 Chr.9: 100586398-100586398 on Build GRCh38
rs772947291 Chr.9: 100586435-100586435 on Build GRCh38
rs767144523 Chr.9: 100586254-100586254 on Build GRCh38
rs371645389 Chr.9: 100586518-100586518 on Build GRCh38
rs763581383 Chr.9: 100586233-100586233 on Build GRCh38
rs769458575 Chr.9: 100586096-100586097 on Build GRCh38
rs770238683 Chr.9: 100586448-100586448 on Build GRCh38
rs1014259103 Chr.9: 100586186-100586186 on Build GRCh38
rs757138375 Chr.9: 100586188-100586188 on Build GRCh38
rs772229637 Chr.9: 100586319-100586319 on Build GRCh38
rs200688664 Chr.9: 100586082-100586082 on Build GRCh38
rs770571523 Chr.9: 100586083-100586086 on Build GRCh38
rs745878563 Chr.9: 100586090-100586091 on Build GRCh38
rs776232912 Chr.9: 100586424-100586424 on Build GRCh38
rs777615745 Chr.9: 100586179-100586179 on Build GRCh38
rs751703046 Chr.9: 100586276-100586276 on Build GRCh38
rs772422202 Chr.9: 100586382-100586382 on Build GRCh38
rs145774333 Chr.9: 100586256-100586256 on Build GRCh38
rs565406194 Chr.9: 100586361-100586361 on Build GRCh38
rs957720883 Chr.9: 100586314-100586314 on Build GRCh38
rs876657509 Chr.9: 100586224-100586224 on Build GRCh38
rs746006234 Chr.9: 100586091-100586091 on Build GRCh38
rs183034024 Chr.9: 100586519-100586519 on Build GRCh38
rs746409931 Chr.9: 100586378-100586378 on Build GRCh38
rs762683955 Chr.9: 100586298-100586298 on Build GRCh38
rs753757744 Chr.9: 100586184-100586184 on Build GRCh38
rs771622110 Chr.9: 100586299-100586299 on Build GRCh38
rs749603456 Chr.9: 100586296-100586296 on Build GRCh38
rs757191826 Chr.9: 100586266-100586266 on Build GRCh38
rs764452556 Chr.9: 100586452-100586452 on Build GRCh38
rs776866140 Chr.9: 100586116-100586116 on Build GRCh38
rs756021968 Chr.9: 100586083-100586083 on Build GRCh38
rs753591105 Chr.9: 100586260-100586260 on Build GRCh38
rs749739234 Chr.9: 100586359-100586359 on Build GRCh38

View Full Product Details