Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105464082-105464585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSD1L
Entrez Gene ID
83856
Gene Name
fibronectin type III and SPRY domain containing 1 like
Gene Aliases
CCDC10, CSDUFD1, FSD1CL, FSD1NL, MIR1
UniGene
Hs.136901
Gene Chromosome Location
Chr.9: 105442183-105552433 on Build GRCh38
Transcripts
NM_001145313.2 NM_001287191.1 NM_001287192.1 NM_031919.4 XM_005252254.2 XM_005252257.3 XM_005252260.3 XM_011519077.2 XM_011519078.2 XM_011519079.2 XM_017015182.1 XM_017015183.1 XM_017015185.1 XM_017015187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6266617 c.207+9G>T p.? Chr.9: 105464340-105464340 on Build GRCh38
6266618 c.111+9G>T p.? Chr.9: 105464340-105464340 on Build GRCh38
455098 c.172A>T p.I58F Chr.9: 105464296-105464296 on Build GRCh38
4448628 c.16-1G>A p.? Chr.9: 105464235-105464235 on Build GRCh38
4473861 c.91C>T p.H31Y Chr.9: 105464311-105464311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768563159 Chr.9: 105464137-105464137 on Build GRCh38
rs949016828 Chr.9: 105464479-105464479 on Build GRCh38
rs557408266 Chr.9: 105464150-105464150 on Build GRCh38
rs527414528 Chr.9: 105464166-105464166 on Build GRCh38
rs916109791 Chr.9: 105464467-105464467 on Build GRCh38
rs1011446757 Chr.9: 105464207-105464207 on Build GRCh38
rs768844207 Chr.9: 105464244-105464244 on Build GRCh38
rs894762325 Chr.9: 105464117-105464117 on Build GRCh38
rs773986049 Chr.9: 105464373-105464373 on Build GRCh38
rs927361559 Chr.9: 105464375-105464375 on Build GRCh38
rs188989563 Chr.9: 105464357-105464357 on Build GRCh38
rs945768364 Chr.9: 105464110-105464110 on Build GRCh38
rs1045681841 Chr.9: 105464498-105464498 on Build GRCh38
rs34789325 Chr.9: 105464134-105464135 on Build GRCh38
rs41277771 Chr.9: 105464338-105464338 on Build GRCh38
rs984985062 Chr.9: 105464461-105464461 on Build GRCh38
rs934686619 Chr.9: 105464558-105464558 on Build GRCh38
rs919697367 Chr.9: 105464377-105464377 on Build GRCh38
rs1026938061 Chr.9: 105464209-105464209 on Build GRCh38
rs998910699 Chr.9: 105464154-105464154 on Build GRCh38
rs763243174 Chr.9: 105464345-105464345 on Build GRCh38
rs748451838 Chr.9: 105464206-105464206 on Build GRCh38
rs748573579 Chr.9: 105464268-105464268 on Build GRCh38
rs41277773 Chr.9: 105464397-105464397 on Build GRCh38
rs575765409 Chr.9: 105464199-105464199 on Build GRCh38
rs531255492 Chr.9: 105464459-105464459 on Build GRCh38
rs907201457 Chr.9: 105464542-105464542 on Build GRCh38
rs770295157 Chr.9: 105464286-105464286 on Build GRCh38
rs541272804 Chr.9: 105464135-105464135 on Build GRCh38
rs972462769 Chr.9: 105464366-105464366 on Build GRCh38
rs549658944 Chr.9: 105464494-105464494 on Build GRCh38
rs370718247 Chr.9: 105464448-105464448 on Build GRCh38
rs1041951218 Chr.9: 105464545-105464545 on Build GRCh38
rs1028648299 Chr.9: 105464224-105464224 on Build GRCh38
rs930800498 Chr.9: 105464394-105464394 on Build GRCh38
rs986422705 Chr.9: 105464254-105464254 on Build GRCh38
rs747645305 Chr.9: 105464281-105464281 on Build GRCh38
rs776821757 Chr.9: 105464249-105464249 on Build GRCh38

View Full Product Details